In primary care, managing cow’s milk and egg allergies effectively can be a challenge. With similar and overlapping symptoms, it can be difficult to accurately identify the triggers and develop an effective management plan.
This article explores ways to optimize the diagnosis and management of these common food allergies, spotlighting the pivotal role of specific IgE tests, and utilizing cutting-edge ImmunoCAP™ technology, to deliver more accurate and informed care.
The ImmunoCAP test is a precise aid in diagnosing allergies, especially useful for food allergies like cow's milk or egg. It identifies specific allergens triggering allergic reactions, aiding in accurate diagnosis and personalized management plans.
In conjunction with the patient's detailed clinical history and physical examination, testing with the ImmunoCAP Specific IgE test will assist you in the following:
Epidemiology
Cow’s milk allergy, CMA, stands as the most common childhood allergy, affecting 2-3% of children under the age of 3.1-3
The early onset of sensitization, typically occurring within the first year of life, underscores the significant health concern posed by CMA.3 Studies indicate a resolution rate of around 50% by the age of 10.1-3 CMA can manifest as either IgE or non-IgE mediated reactions.
How to diagnose CMA?
Like any food allergy, diagnosing CMA entails a comprehensive approach, relying on clinical history, physical examinations, and targeted inquiries about food allergies.4, 5
The diagnostic journey is intricate due to the vague nature of symptoms, leading to potential misdiagnoses and subsequent health risks for infants and caregivers.6
Learn more about the common symptoms of cow’s milk allergy.
Clinical suspicion of IgE-mediated CMA is confirmed by demonstrating the presence of allergen-specific IgE antibodies through in vivo (skin tests) or in vitro blood testing. Anyone presenting with allergy symptoms can be a candidate for specific IgE testing.
The ImmunoCAP Specific IgE blood test aids in identifying candidates for an oral food challenge, offering opportunities for personalized care: more precise guidance to broaden dietary options and enhance patient’s quality of life.7, 8
Access the step-by-step guide to cow's milk allergy diagnosis for additional information.
How to test for CMA?
Epidemiology
Egg allergy is the second most common food allergy in infants and young children, second only to cow’s milk in prevalence, affecting 0,5-2,5% of them.9-11
It usually presents in the second half of the first year of life, with a median age of presentation of 10 months.9
Resolution of egg allergy is highly associated with lower IgE levels and the absence of systemic reactions beyond skin presentation. Although approximately 50 % of pediatric patients are likely to outgrow their egg allergy by age 6, research suggests that children are outgrowing their egg allergy more slowly than before.11
How to diagnose egg allergy?
Making an accurate diagnosis when evaluating a patient with a possible egg allergy is important to avoid unnecessary dietary restrictions and prevent life-threatening reactions.
The typical diagnostic routine for egg allergy begins with a physical examination and detailed clinical history.4, 5, 12
Find out about common symptoms of egg allergy
In adults and children, subsequent steps of primary diagnosis may involve skin-prick testing or specific IgE testing, such as the ImmunoCAP test, to confirm the diagnosis or assess the need for an oral food challenge. The Specific IgE test leverages egg white extract containing major allergens recognized by egg-allergic individuals, allowing it to be a highly accurate initial diagnostic tool.13
Access the step-by-step guide to egg allergy diagnosis for additional information.
How to test for egg allergy?
In some children allergic to eggs, the allergy may disappear, allowing for the reintroduction of foods containing this allergen into their diet. Therefore, it is recommended to undergo periodic evaluations and oral food challenges to monitor any progress toward the resolution of egg allergy.
As a General Practitioner, you play an essential role in the early detection, initial testing for allergen triggers in cow’s milk or egg allergies. Patients want answers and relief, and you want the best tools to deliver. By prescribing a specific IgE blood test, you can identify food allergens impacting patients, making it easier for them to avoid their triggers and live healthier.
Regular follow-ups are recommended, conducting clinical reassessments, and performing testing for example every 6-12 months with the same technique.
By employing these measures, you can tailor personalized management plans, including customized dairy elimination strategies and age-appropriate milk substitutes. This approach not only improves patient outcomes but also reinforces the quality of care provided.
In cases where food allergy diagnosis remains uncertain or requires specialized attention, your collaboration with allergists and clinical immunology professionals ensures that patients receive tailored treatment plans. This collaborative effort contributes to the comprehensive management and well-being of individuals affected by cow’s milk or egg allergies
You have the power to make a difference.
For additional info on ImmunoCAP testing, allergies, and/or educational tools and resources, contact our allergy experts.
Tools to aid in your diagnosis and patient management.