Case Study
Michael R. Broyles
PharmD – Director of Medical Affairs, Thermo Fisher Scientific
Explore similar topics
Approximately 30% of PCT occurs renally.1
The elevated PCT level on admission was consistent with bacterial pneumonia. The patient was successfully treated with antibiotics.
The difference in PCT decrease was higher on dialysis days (approximately 50%) vs. non-dialysis days (approximately 30%). This is to be expected when there is renal impairment, PCT will only decrease approximately 30%.1
Submit your own PCT Case Study and you may be selected to be featured on procalcitonin.com.