The overuse and misuse of antibiotics accelerates the natural process of antibiotic resistance.7 Resistant bacteria are then spread via healthcare-acquired infections, which hospitals and clinics struggle to prevent and control. It’s a treacherous cycle that has led to increased resistance to life-saving antibiotics around the world, greatly reducing treatment options. Some bacterial strains have even become resistant to both first- and second-line antibiotics. These multidrug-resistant (MDR) strains can only be treated with last-resort antibiotics, if they can be treated at all.
For example, among antibiotic-resistant pathogens, one of the most concerning is Klebsiella pneumoniae (K. pneumoniae). It accounts for about one third of all Gram-negative infections, including urinary tract infections, cystitis, pneumonia, surgical wound infections, endocarditis and sepsis.8
K. pneumoniae has a high resistance against the four major antibiotic classes: the third-generation cephalosporins, aminoglycosides, fluoroquinolones and carbapenems.9-11 This resistance is resulting in a growing, worldwide problem in which clinicians have fewer effective antibiotic treatment options for hospital-acquired infections.12