PCT and blood cultures
Blood cultures are performed to identify bacteria, yeast, or other microorganisms that may be causing a patient’s clinical condition. Physicians then use the information to prescribe targeted treatments. Traditional blood culture takes several days (usually 2-3, but sometimes 10 days or longer, depending on the germ10-12), although more recent methods can provide a result in several hours.13
Given the slow turnaround time for cultures, physicians need to prescribe antibiotics empirically, as delaying antibiotic treatment in cases of true bacterial infection may increase the patient’s risk of progressing to a severe disease state. However, this decision comes at the cost of possibly increasing the chances of antibiotic overuse and resistance.
Another challenge with relying on blood cultures is interpreting the result correctly, as blood culture sensitivity is only moderate and may be further decreased in patients already on antibiotics. In addition, there is the potential for false-positive results due to sample contamination. Consequently, it can be difficult to determine whether the bacteria that grew in the culture are the cause of the clinical condition of the patient or not.10-12
With each PCT test taking only 20 minutes, results may be available in just 1-2 hours, especially if the hospital has good logistics in place. This information allows treating physicians to complement their clinical assessment in cases of suspected bacterial infection, including first decisions on the initiation of empiric, broad-spectrum antibiotic therapy. In addition to the PCT test, a blood culture helps identify the pathogen(s) and aid in narrowing the antibiotic spectrum.
During the further treatment course, PCT serial testing—combined with medical history and patient presentation—can be used for monitoring antibiotic treatment efficacy.