Select the pipette that’s right for your lab and application
Meet the demands of daily pipetting with the right pipette for your workflow. When you are looking for improved accuracy and precision, we have a selection of manual, electronic, single channel and multichannel pipettes.
Manual pipettes
F1-ClipTip Pipettes
Finnpipette F1 Pipettes
Finnpipette F2 Pipettes
Description
The F1-ClipTip pipetting system provides a complete seal on every channel for precise sample volumes, saving valuable time to accelerate your discoveries. No longer bang tips onto your pipette. With a light touch each tip is clipped securely on the pipette and remains sealed until you decide to eject the tips. F1-ClipTip supports consistent and reproducible pipetting. Enjoy the enhanced volume adjustment on Thermo Scientific Finnpipette F1 system pipette, which is silent and effortless with a textured adjustment knob for a firmer feel when setting volumes. The adjustable finger rest and lightweight design maximize comfort without sacrificing performance. Get optimal precision and ergonomics, all in a lightweight design that maximizes user comfort and reduces strain. Finnpipette F2 pipettes meet the challenges of high demanding applications where robustness and durability are essential. With a sleek and rugged design the variable volume pipettes are built with tough PVDF components that stand up to harsh chemicals and damaging effects of UV light. These variable volume pipettes are fully autoclavable without disassembling. They feature a wide selection of expressly designed tips for optimal performance, precision and accuracy.
Channels
Single, 8 - , 12 -channel
Single, 8-, 12- 16-channel
Single, 8-, 12- 16-channel
Format
Variable and Fixed Volume Variable and Fixed Volume Variable and Fixed Volume
Volume Range
0.1-1000 µl (variable)
10-1000 µl  (fixed)
0.2-10,000 µl (variable)
10-10,000 µl  (fixed)
0.2-10,000 µl (variable)
10-10,000 µl  (fixed)
Electronic pipettes
E1-ClipTip Pipettes
E1-ClipTip Equalizer Pipettes
Finnpipette Novus Pipettes
Description
E1-ClipTip Pipettes are designed for accurate and efficient liquid handling. They feature an electronic control system that supports precise pipetting volume with a simple push of a button. The ClipTip technology secures leak-free sample pickup and ejection. With it's ergonomic design and lightweight construction, the pipette offers comfortable and fatigue-free pipetting. The Equalizer Pipette is a state-of-the-art laboratory instrument that revolutionizes liquid handling. Designed to transfer multiple samples between different labware formats using adjustable tip spacing, the Equalizer helps save time and repetitions compared to the single channel manual pipette. The Cliptip technology supports secure and leak-free sample pick up and ejection, minimizing risk of cross-contamination. The Finnpipette Novus Pipette is a reliable and versatile laboratory tool designed for accurate and efficient pipetting. It features a lightweight and ergonomic design that supports comfortable and fatigue-free pipetting, even during extended use. With its user-friendly interface and intuitive operation, the pipette simplifies pipetting tasks and enhances productivity in the laboratory. Its durable construction and high-quality materials make it a durable and long-lasting pipette choice for scientists and researchers.
Channels
Single, 8-, 12-, 16- channel
6-, 8-, 12- channel
8-, 12-, 16- channel
Volume Range
0.5 – 1250 µl 0.5 – 1250 µl
1 – 5000 µl
Transfer pipettes
General Purpose
Graduated
Narrow Stem
Fine Tip Special purpose Exra Long Exact Volume Capillary
Options          
Bulb draw (mL): 0.3, 0.9, 1.8, 2.2, 2.4, 2.8, 3.0, 4.7, 6.2, 7.4
Bulb draw (mL): 0.8, 1.8, 2.1, 3.2, 3.5, 4.3, 7.4
Stem diameter (mm):
2.5, 3.0, 8.0
Stem diameter (mm):
1.2, 3.0, 4.6, 6.1
Bulb draw (mL): 0.2, 3.3
Length:
9 in (22.5 cm)
12 in (30.0 cm)
Dispense Volume (µl):
20, 40, 50, 60, 80, 100, 150, 200, 250, 300, 400
Volume (µl):
25, 50
Applications          
Rapid transfer, controlled drop size, calibrated transfer
Remove separate liquids, dispense small drops
Genomic and proteomic, reagent prep, drop additions to culture plates
Sample preparation, layering in columns, transfers in columns, loading gels
Assay development, research lab
Assay development, research lab
Assay development, research in microbiology, agriculture, veterinary, food
Liquid transfer, rapid tests, lateral flow assay development
Recommended Use          
Larger aperture for creams and gels, oils, industrial slurries, paints, perfumes
Ideal for measure transfers, drop by drop work, aliquoting
Loading gels, PCR work, DNA sample removal, feed hybridomas, lipid extraction, atomic absorption, spectroscopy
Loading gels, PCR, lipid extractions, DNA samples removal, protein assays
Grear for viscous fluid, paddle for mixing
Sub-culturing and harvesting cells and tissues; Transferring from large bottle; Removal of supernatant fluids from centrifugation
Exact volume dispensing, analytical testing
Pathogen testing
Liquid transfer, point of care tests, rapid tests, lateral flow assay development
For Laboratory Use. It is the customer’s responsibility to ensure that the performance of the product is suitable for customers’ specific uses or applications.
1x1 image pixel for data collection