Get a secure pipette tip seal, every time
ClipTip pipette system is a revolutionary tool designed to enhance precision and ease in pipetting tasks. With its unique ClipTip technology, the system allows secure attachments of tips, reducing the risk of tip ejection and sample loss. Move over, its ergonomic design guarantees a comfortable pipetting experience even during extended use. This innovative system facilitates accuracy, reliability, and user comfort to deliver exceptional results in a wide range of applications.
Benefits of ClipTip Pipetting System
Precise sample volumes in every channel regardless of touch-off
Generic tips (a) and ClipTip tips (b) after dispensing into two 96-well microplates. ClipTip pipette tips have an airtight seal in every channel and 68% less variation in dispensed liquid vs. traditional friction based systems. The tips attachment force used was 3.8 kg.
Generic Tips (a)
ClipTip Tips (b)
Download application note
Tips only eject when you want them to
When ClipTip pipette tips are on, they are sealed and will not loosen. The ClipTip pipette system remains sealed until you decide to eject the tips, even in the most comprehensive applications, including multiple tip touches and mixing cycles. Yes those tips are supporting 2 lb weights.
cliptip weights
Safer and more comfortable pipetting
When all ergonomic elements are combined, pipetting is a comfortable, safe, and enjoyable experience. Average results from the tip attachment force measurements. The error bars show standard deviation.
Tip attachment forces of the E1-ClipTip Pipettes* were up to 87% lower compared to the other electroic pipettes tested.
Tip ejection forces of the E1-ClipTip Pipettes* are up to 93% lower compared to the other electronic pipettes tested.
*Testing was done with E1-ClipTip Equalizer pipettes
  • Low tip attachment, ejection, and plunger forces required
  • Remarkably lightweight
  • Electronic index finger pipetting action minimizes thumb strain
  • Adjustable finger rest supports optimal pipetting position for both left and right-handed users
  • Electronic tip ejection makes tip ejection effortless
  • An intuitive user interface to help prevent any accidental tip ejection whenever there is liquid inside tips
  • Rotating display, unique multichannel ski boot design, and short single-channel barrel maximize comfort
Save up to 80% of your time with the E1-ClipTip Equalizer pipette
Sample transfer made easy. Use E1-ClipTip Adjustable Tip Spacing Equalizer pipettes instead of manual pipettes and transfer multiple samples between different labware formats at once, with one pipette and one movement. 
Available ClipTip Pipetting Systems
For Laboratory Use. It is the customer’s responsibility to ensure that the performance of the product is suitable for customers’ specific uses or applications.