Hot-start PCR is a modified version of the traditional polymerase chain reaction (PCR) technique that provides higher specificity and sensitivity by minimizing non-specific amplification. In hot-start PCR, the polymerase stays inactive at lower temperatures due to an inhibitor or modification of the enzyme to prevent extension of primers at lower temperatures. Upon heating, the inhibitor is released and amplification begins.
Video Player is loading.
Current Time 0:00
Duration 2:25
Loaded: 6.77%
Stream Type LIVE
Remaining Time 2:25
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Watch the video on common issues in traditional PCR amplification and how you can resolve them with hot-start PCR.
    Highlights of Platinum II Taq Hot-Start DNA Polymerase
    • Universal primer annealing at 60°C—No optimization of primer annealing temperatures and multiple PCR runs can be co-cycled together
    • 4x faster DNA synthesis—Faster synthesis rates than traditional Taq polymerases
    • High inhibitor tolerance—Utilizes an engineered Taq polymerase that is extremely robust
    • Platinum hot-start technology—Enhances PCR specificity, sensitivity, and yields; allows for room temperature reaction setup
    • Green buffer formats—Help reduce pipetting errors and enable direct gel loading
    Advantages of Platinum II Taq Hot-Start DNA Polymerase
    The innovative Platinum II PCR buffer enables universal primer annealing protocol by isostabilizing primer-template duplex structures. When amplifying with Platinum II Taq DNA Polymerase, a single 60°C annealing temperature can be used for any primer pair.  
    Two panels stacked vertically; top panel shows partial binding of primers are 60°C with standard buffer and bottom panel shows perfect primer annealing at 60°C with Platinum II PCR buffer
    Figure 1. Isostabilization effect of Platinum II PCR buffer compared to standard PCR buffer.
    Video Player is loading.
    Current Time 0:00
    Duration 0:30
    Loaded: 32.81%
    Stream Type LIVE
    Remaining Time 0:30
    1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
      • en (Main), selected
      Watch the benefits of universal annealing feature.
      Video Player is loading.
      Current Time 0:00
      Duration 0:30
      Loaded: 32.81%
      Stream Type LIVE
      Remaining Time 0:30
      1x
        • Chapters
        • descriptions off, selected
        • captions off, selected
        • en (Main), selected
        Platinum II Taq Hot-Start DNA Polymerase is four times faster than conventional Taq DNA polymerase. Watch video.
        Video Player is loading.
        Current Time 0:00
        Duration 0:30
        Loaded: 33.33%
        Stream Type LIVE
        Remaining Time 0:30
        1x
          • Chapters
          • descriptions off, selected
          • captions off, selected
          • en (Main), selected
          Platinum II Taq Hot-Start DNA Polymerase exhibits tolerance to common PCR inhibitors. Watch video.
          Green buffer options allow for direct gel loading and help reduce pipetting errors. DNA migration is easily tracked with two dyes that are readily visible during electrophoresis.
          Schematic depicts direct loading of PCR products from tube to electrophoresis machine
          Figure 2. Direct gel loading simplifies PCR to electrophoresis.
          The innovative Platinum II PCR buffer enables universal primer annealing protocol by isostabilizing primer-template duplex structures. When amplifying with Platinum II Taq DNA Polymerase, a single 60°C annealing temperature can be used for any primer pair.  
          Two panels stacked vertically; top panel shows partial binding of primers are 60°C with standard buffer and bottom panel shows perfect primer annealing at 60°C with Platinum II PCR buffer
          Figure 1. Isostabilization effect of Platinum II PCR buffer compared to standard PCR buffer.
          Video Player is loading.
          Current Time 0:00
          Duration 0:30
          Loaded: 32.81%
          Stream Type LIVE
          Remaining Time 0:30
          1x
            • Chapters
            • descriptions off, selected
            • captions off, selected
            • en (Main), selected
            Watch the benefits of universal annealing feature.
            Video Player is loading.
            Current Time 0:00
            Duration 0:30
            Loaded: 33.33%
            Stream Type LIVE
            Remaining Time 0:30
            1x
              • Chapters
              • descriptions off, selected
              • captions off, selected
              • en (Main), selected
              Platinum II Taq Hot-Start DNA Polymerase is four times faster than conventional Taq DNA polymerase. Watch video.
              Video Player is loading.
              Current Time 0:00
              Duration 0:30
              Loaded: 33.33%
              Stream Type LIVE
              Remaining Time 0:30
              1x
                • Chapters
                • descriptions off, selected
                • captions off, selected
                • en (Main), selected
                Platinum II Taq Hot-Start DNA Polymerase exhibits tolerance to common PCR inhibitors. Watch video.
                Green buffer options allow for direct gel loading and help reduce pipetting errors. DNA migration is easily tracked with two dyes that are readily visible during electrophoresis.
                Schematic depicts direct loading of PCR products from tube to electrophoresis machine
                Figure 2. Direct gel loading simplifies PCR to electrophoresis.
                Platinum Taq DNA polymerases: Selection guide
                product package and three tubes of reagents
                Platinum II Taq Hot-Start DNA Polymerase
                product package and four tubes of reagents
                Platinum Taq DNA Polymerase
                Universal annealing protocolYesNo
                Speed15 sec/kb1 min/kb
                Flexible extension step*YesNo
                Inhibitor toleranceYesNo
                Target lengthUp to 5 kbUp to 5 kb
                Hot-start modificationAntibody-mediatedAntibody-mediated
                Fidelity versus Taq DNA Polymerase1x1x
                Amplicon overhangs3’A3’A
                Benchtop stability of assembled PCR reactions24 h24 h
                GC-rich amplificationYesYes
                Certified low level of residual human and bacterial DNAYes
                (≤1 copy of bacterial gDNA/enzyme unit)
                No
                Master mix formats
                Colorless
                Order now
                Green**
                Order now
                Colorless
                Order now
                Green**
                Order now
                Stand-alone enzyme formats
                Colorless†
                Order now
                Colorless
                Order now
                Green**
                Order now
                *The extension step can be extended up to 60 sec/kb without the effect on specificity.
                **Direct gel loading with green buffer options.
                Green buffer available as separate item for use with stand-alone enzyme for direct loading gel.
                ‡During manufacturing of Platinum II Taq Hot-Start DNA Polymerase, strict measures are taken to control and verify by qPCR that no more than one copy of residual bacterial genomic DNA is present per unit of the polymerase.
                Benchmarking data on Platinum II Taq Hot-Start DNA Polymerase
                Engineered Platinum II Taq Hot-Start DNA Polymerase enables fast cycling of assays in as few as 30 minutes. The universal primer annealing temperature enables cycling of shorter and longer amplicons together using the same protocol.
                Process flowchart depicts time savings co-cycling multiple targets in a single protocol
                Figure 3. Time saving enabled by assay co-cycling. PCR assays using conventional PCR reagents require specific protocols for amplification of each DNA fragment because of the different primer annealing temperatures and extension steps. Therefore, with traditional PCR reagents, multiple targets often cannot be amplified together in the same PCR run. With Platinum II Taq Hot-Start DNA Polymerase, different PCR assays can be cycled together using one protocol with a universal primer annealing temperature and the extension step selected for the longest fragment to be amplified. Moreover, Platinum II Taq Hot-Start DNA Polymerase is a fast DNA polymerase, delivering PCR results in as little as 30 minutes.
                Gel image of multiple targets amplified from human genomic DNA in a single protocol
                Figure 4. Platinum II Taq Hot-Start DNA Polymerase enables cycling of shorter and longer amplicons together. 132 bp, 251 bp, 1,005 bp, and 3.9 kb fragments were amplified from 50 ng of human genomic DNA in 50 μL reactions using Platinum II Taq Hot-Start DNA Polymerase or other hot-start DNA polymerases: (A) NEB OneTaq™ Hot Start DNA Polymerase, (B) Qiagen Fast Cycling™ PCR Kit, (C) Roche FastStart™ Taq DNA Polymerase. The same protocol was used for all four targets with the annealing and extension settings indicated. The size marker used is Thermo Scientific ZipRuler Express DNA Ladder 2.
                The higher synthesis rate of Platinum II Taq Hot-Start DNA Polymerase allows 2 times faster PCR results compared to other hot-start Taq DNA polymerases.
                Stacked bar graph depicting ramping time in red and cycling time in purple for each polymerase tested. The corresponding PCR product is presented below the graph
                Figure 5. Fast cycling reduces PCR run time. Amplification of a 529 bp fragment from 50 ng of human genomic DNA in 50 μL reactions for 35 cycles was carried out using Platinum II Taq Hot-Start DNA Polymerase and hot-start DNA polymerases from other suppliers: (A) Sigma-Aldrich KAPA2G™ Fast HotStart PCR Kit, (B) NEB OneTaq™ Hot Start DNA Polymerase, (C) Promega GoTaq™ G2 DNA Polymerase, (D) Toyobo Quick Taq™ HS DyeMix, (E) Roche FastStart™ Taq DNA Polymerase, and (F) Sigma-Aldrich JumpStart™ Taq DNA Polymerase. Cycling times for each polymerase are shown in purple, while ramping times on the ProFlex PCR System (6°C/sec peak block ramp rate) are shown in red. PCR product analysis in 1% TAE agarose gels is presented below the graph. The size marker used is the ZipRuler Express DNA Ladder 2.
                The sensitivity of Platinum II Taq Hot-Start DNA Polymerase enables successful amplification of specific product in experiments where there is a limited amount of starting material, or the target DNA is in low concentration. . 
                Gel images of DNA amplified from low inputs; Platinum II Taq Hot-Start DNA Polymerase shows superior performance compared to competitor products
                Figure 6. High sensitivity and reliable amplification from low amounts of input DNA. Amplification of a 529 bp fragment from 0 (no template control); 0.016; 0.08; 0.4; 2; 10; 50; 250 ng of human genomic DNA were amplified in 50 μL PCR reactions using Platinum II Taq Hot-Start DNA Polymerase or competitor DNA polymerases (A: KAPA2G™ Fast HotStart, B: NEB OneTaq™ Hot Start, C: Promega GoTaq™ G2, D: Sigma JumpStart™ Taq, and E: Takara™ Taq HS Perfect Mix). The estimated copy number is ~5 copies per 0.016 ng of human genomic DNA. The molecular weight marker is ZipRuler Express DNA Ladder 2.
                Platinum II Taq Hot-Start DNA Polymerase exhibits resistance to inhibitors and helps enable successful amplification of samples with suboptimal purity.
                Gel depicting robust amplification by Platinum II Taq Hot-Start DNA Polymerase in the presence of inhibitors
                Figure 7. Resistance to inhibitors. Amplification of a 1 kb fragment from human genomic DNA using Platinum II Taq Hot-Start DNA Polymerase or competitor DNA polymerases (A: KAPA2G™ Robust HotStart, B: NEB OneTaq™ Hot Start, C: Promega GoTaq™ G2, and D: Takara™ Taq Hot Start Version) in reaction mixtures containing: 1: humic acid (up to final concentration of 1.3 µg/mL), 2: hemin (up to final concentration of 6 µM), 3: xylan (up to final concentration of 0.26 mg/mL), or 4: no inhibitor control. The molecular weight marker used is ZipRuler Express DNA Ladder 2.
                Amplification of DNA extracted from FFPE tissue samples
                Figure 8. Amplification of DNA extracted from FFPE tissue samples. Amplification of a 527 bp fragment from varying amounts of DNA extracted from mouse FFPE tissue samples using Platinum II Taq Hot-Start DNA polymerase. RecoverAll Total Nucleic Acid Isolation Kit for FFPE was used for DNA extraction. NTC: no template control. PC: positive control from 1 ng of purified mouse genomic DNA. The molecular weight marker used is ZipRuler Express DNA Ladder 2.
                The formulation of Platinum II Taq Hot-Start DNA Polymerase and 2X Master Mixes allows for amplification of versatile range of targets, from AT-rich to GC-rich. A separate vial of Platinum GC Enhancer is provided for specific amplification and improved yields of targets with high-GC content.
                Gel image of GC-rich targets
                Figure 9. Robust amplification of AT-rich and GC-rich targets. Various DNA fragments of increasing GC content (indicated above the corresponding lanes) were amplified from 100 ng of human genomic DNA in 50 µL PCR reactions. Platinum GC Enhancer was used for targets with >65% GC. The molecular weight marker used is ZipRuler Express DNA Ladder 2.
                PCR fragments generated by Platinum II Taq Hot-Start DNA Polymerase work well for Sanger sequencing. The enzyme’s superior performance, universal primer annealing, and fast synthesis enable generation of PCR amplicons for Sanger sequencing, with ease and simplicity.
                Sanger sequencing results indicating distinct sequencing peaks using Platinum II Taq Hot-start DNA Polymerase
                Figure 10. High-quality Sanger sequencing results. A 1.6 kb PCR fragment amplified by Platinum II Taq Hot-Start DNA Polymerase was Sanger sequenced using Applied Biosystems 3130xl Genetic Analyzer. Data reported by the KB basecaller of the built-in sequencing analysis software is shown. Clear range read length (CRL) is defined as the longest uninterrupted segment of bases at a given Quality Value (QV). QV20 corresponds to 1% probability of a base call error and QV30 corresponds to 0.1%. QV>20 is considered high quality and acceptable in most cases.
                Engineered Platinum II Taq Hot-Start DNA Polymerase enables fast cycling of assays in as few as 30 minutes. The universal primer annealing temperature enables cycling of shorter and longer amplicons together using the same protocol.
                Process flowchart depicts time savings co-cycling multiple targets in a single protocol
                Figure 3. Time saving enabled by assay co-cycling. PCR assays using conventional PCR reagents require specific protocols for amplification of each DNA fragment because of the different primer annealing temperatures and extension steps. Therefore, with traditional PCR reagents, multiple targets often cannot be amplified together in the same PCR run. With Platinum II Taq Hot-Start DNA Polymerase, different PCR assays can be cycled together using one protocol with a universal primer annealing temperature and the extension step selected for the longest fragment to be amplified. Moreover, Platinum II Taq Hot-Start DNA Polymerase is a fast DNA polymerase, delivering PCR results in as little as 30 minutes.
                Gel image of multiple targets amplified from human genomic DNA in a single protocol
                Figure 4. Platinum II Taq Hot-Start DNA Polymerase enables cycling of shorter and longer amplicons together. 132 bp, 251 bp, 1,005 bp, and 3.9 kb fragments were amplified from 50 ng of human genomic DNA in 50 μL reactions using Platinum II Taq Hot-Start DNA Polymerase or other hot-start DNA polymerases: (A) NEB OneTaq™ Hot Start DNA Polymerase, (B) Qiagen Fast Cycling™ PCR Kit, (C) Roche FastStart™ Taq DNA Polymerase. The same protocol was used for all four targets with the annealing and extension settings indicated. The size marker used is Thermo Scientific ZipRuler Express DNA Ladder 2.
                The higher synthesis rate of Platinum II Taq Hot-Start DNA Polymerase allows 2 times faster PCR results compared to other hot-start Taq DNA polymerases.
                Stacked bar graph depicting ramping time in red and cycling time in purple for each polymerase tested. The corresponding PCR product is presented below the graph
                Figure 5. Fast cycling reduces PCR run time. Amplification of a 529 bp fragment from 50 ng of human genomic DNA in 50 μL reactions for 35 cycles was carried out using Platinum II Taq Hot-Start DNA Polymerase and hot-start DNA polymerases from other suppliers: (A) Sigma-Aldrich KAPA2G™ Fast HotStart PCR Kit, (B) NEB OneTaq™ Hot Start DNA Polymerase, (C) Promega GoTaq™ G2 DNA Polymerase, (D) Toyobo Quick Taq™ HS DyeMix, (E) Roche FastStart™ Taq DNA Polymerase, and (F) Sigma-Aldrich JumpStart™ Taq DNA Polymerase. Cycling times for each polymerase are shown in purple, while ramping times on the ProFlex PCR System (6°C/sec peak block ramp rate) are shown in red. PCR product analysis in 1% TAE agarose gels is presented below the graph. The size marker used is the ZipRuler Express DNA Ladder 2.
                The sensitivity of Platinum II Taq Hot-Start DNA Polymerase enables successful amplification of specific product in experiments where there is a limited amount of starting material, or the target DNA is in low concentration. . 
                Gel images of DNA amplified from low inputs; Platinum II Taq Hot-Start DNA Polymerase shows superior performance compared to competitor products
                Figure 6. High sensitivity and reliable amplification from low amounts of input DNA. Amplification of a 529 bp fragment from 0 (no template control); 0.016; 0.08; 0.4; 2; 10; 50; 250 ng of human genomic DNA were amplified in 50 μL PCR reactions using Platinum II Taq Hot-Start DNA Polymerase or competitor DNA polymerases (A: KAPA2G™ Fast HotStart, B: NEB OneTaq™ Hot Start, C: Promega GoTaq™ G2, D: Sigma JumpStart™ Taq, and E: Takara™ Taq HS Perfect Mix). The estimated copy number is ~5 copies per 0.016 ng of human genomic DNA. The molecular weight marker is ZipRuler Express DNA Ladder 2.
                Platinum II Taq Hot-Start DNA Polymerase exhibits resistance to inhibitors and helps enable successful amplification of samples with suboptimal purity.
                Gel depicting robust amplification by Platinum II Taq Hot-Start DNA Polymerase in the presence of inhibitors
                Figure 7. Resistance to inhibitors. Amplification of a 1 kb fragment from human genomic DNA using Platinum II Taq Hot-Start DNA Polymerase or competitor DNA polymerases (A: KAPA2G™ Robust HotStart, B: NEB OneTaq™ Hot Start, C: Promega GoTaq™ G2, and D: Takara™ Taq Hot Start Version) in reaction mixtures containing: 1: humic acid (up to final concentration of 1.3 µg/mL), 2: hemin (up to final concentration of 6 µM), 3: xylan (up to final concentration of 0.26 mg/mL), or 4: no inhibitor control. The molecular weight marker used is ZipRuler Express DNA Ladder 2.
                Amplification of DNA extracted from FFPE tissue samples
                Figure 8. Amplification of DNA extracted from FFPE tissue samples. Amplification of a 527 bp fragment from varying amounts of DNA extracted from mouse FFPE tissue samples using Platinum II Taq Hot-Start DNA polymerase. RecoverAll Total Nucleic Acid Isolation Kit for FFPE was used for DNA extraction. NTC: no template control. PC: positive control from 1 ng of purified mouse genomic DNA. The molecular weight marker used is ZipRuler Express DNA Ladder 2.
                The formulation of Platinum II Taq Hot-Start DNA Polymerase and 2X Master Mixes allows for amplification of versatile range of targets, from AT-rich to GC-rich. A separate vial of Platinum GC Enhancer is provided for specific amplification and improved yields of targets with high-GC content.
                Gel image of GC-rich targets
                Figure 9. Robust amplification of AT-rich and GC-rich targets. Various DNA fragments of increasing GC content (indicated above the corresponding lanes) were amplified from 100 ng of human genomic DNA in 50 µL PCR reactions. Platinum GC Enhancer was used for targets with >65% GC. The molecular weight marker used is ZipRuler Express DNA Ladder 2.
                PCR fragments generated by Platinum II Taq Hot-Start DNA Polymerase work well for Sanger sequencing. The enzyme’s superior performance, universal primer annealing, and fast synthesis enable generation of PCR amplicons for Sanger sequencing, with ease and simplicity.
                Sanger sequencing results indicating distinct sequencing peaks using Platinum II Taq Hot-start DNA Polymerase
                Figure 10. High-quality Sanger sequencing results. A 1.6 kb PCR fragment amplified by Platinum II Taq Hot-Start DNA Polymerase was Sanger sequenced using Applied Biosystems 3130xl Genetic Analyzer. Data reported by the KB basecaller of the built-in sequencing analysis software is shown. Clear range read length (CRL) is defined as the longest uninterrupted segment of bases at a given Quality Value (QV). QV20 corresponds to 1% probability of a base call error and QV30 corresponds to 0.1%. QV>20 is considered high quality and acceptable in most cases.
                Platinum II Taq Hot-Start DNA Polymerase: Cited and quoted
                Microbiota references
                UsagePublications
                Ion Torrent sequencing of bacterial 16S rRNA genesBasic M, Bolsega S, Smoczek A et al. (2021) Monitoring and contamination incidence of gnotobiotic experiments performed in microisolator cages. Int J Med Microbiol 311(3):151482. 
                Metabarcoding of sedimentary ancient DNA samplesCapo E, Giguet-Covex C, Rouillard A et al. (2021) Lake sedimentary DNA research on past terrestrial and aquatic biodiversity: overview and recommendations. Quaternary 4:6. 
                Restriction enzyme site-directed amplification PCR (REDA-PCR) and cloning with microalga DNALee JW, Lee MW, Ha JS (2020) Development of a species-specific transformation system using the novel endogenous promoter calreticulin from oleaginous microalgae Ettlia sp. Sci Rep 10(1):13947. 
                Diatom DNA amplification with degenerate primers for Illumina sequencingNistal A, Garcia P, Garcia J et al. (2021) DNA metabarcoding and morphological methods show complementary patterns in the metacommunity organization of lentic epiphytic diatoms. ARPHA Conference Abstracts 4: e63672. 
                Ion Torrent sequencing of bacterial 16S rRNA genesSchwarz SR, Hirsch S, Hiergeist A et al (2020) Limited antimicrobial efficacy of oral care antiseptics in microcosm biofilms and phenotypic adaptation of bacteria upon repeated exposure. Clin Oral Investig Epub ahead of print 
                Amplification of bacterial 16S rRNA gene using barcoded primers for Illumina sequencingWallis A, Yannuzzi IM, Choi MW (2021) Investigating the distribution of strains of Erwinia amylovora and streptomycin resistance in apple orchards in New York using CRISPR profiles: a six-year follow-up. Plant Dis Epub ahead of print. 
                Cancer research references
                UsagePublications
                PCR with genomic DNA of primary human cancer cells for mutation detectionAnwar SL, Hasemeier B, Schipper E et al. (2019) LINE-1 hypomethylation in human hepatocellular carcinomas correlates with shorter overall survival and CIMP phenotype. PLoS One 14(5):e0216374. 
                PCR with genomic DNA from whole blood samples, followed by Sanger sequencingArévalo-Jaramillo P, Idrobo A, Salcedo L (2019) Biochemical and genotoxic effects in women exposed to pesticides in Southern Ecuador. Environ Sci Pollut Res Int 26(24):24911–24921. 
                PCR of bisulfite-treated DNA from carcinoma cell lines, followed by Sanger sequencingIbrahim ML, Klement JD, Lu C et al. (2018) Myeloid-Derived Suppressor Cells Produce IL-10 to Elicit DNMT3b-Dependent IRF8 Silencing to Promote Colitis-Associated Colon Tumorigenesis. Cell Rep 25(11):3036–3046. 
                PCR with bisulfite-treated DNA from human tissues and cells, followed by Ion Torrent sequencingMa Y, Chai N, Jiang Q (2020) DNA methyltransferase mediates the hypermethylation of the microRNA 34a promoter and enhances the resistance of patient-derived pancreatic cancer cells to molecular targeting agents. Pharmacol Res 160:105071. 
                Touchdown PCR with DNA from FFPE samples, followed by Sanger SequencingMaier AD, Stenman A, Svahn F et al. (2021) TERT promoter mutations in primary and secondary WHO grade III meningioma. Brain Pathol 31(1):61–69. 
                Two-step RT-PCR with DNA from human cells for detection of transgene expressionMcCormick CA, Samuels TL, Battle MA (2021) H+/K+ATPase Expression in the Larynx of Laryngopharyngeal Reflux and Laryngeal Cancer Patients. Laryngoscope 131(1):130–135. 
                Amplification of an siRNA template from human cancer cell linesWang YL, Chang LC, Chen KB et al. (2021) Aptamer-guided targeting of the intracellular long-noncoding RNA HOTAIR. Am J Cancer Res 11(3):945–954. 
                Microbiota references
                UsagePublications
                Ion Torrent sequencing of bacterial 16S rRNA genesBasic M, Bolsega S, Smoczek A et al. (2021) Monitoring and contamination incidence of gnotobiotic experiments performed in microisolator cages. Int J Med Microbiol 311(3):151482. 
                Metabarcoding of sedimentary ancient DNA samplesCapo E, Giguet-Covex C, Rouillard A et al. (2021) Lake sedimentary DNA research on past terrestrial and aquatic biodiversity: overview and recommendations. Quaternary 4:6. 
                Restriction enzyme site-directed amplification PCR (REDA-PCR) and cloning with microalga DNALee JW, Lee MW, Ha JS (2020) Development of a species-specific transformation system using the novel endogenous promoter calreticulin from oleaginous microalgae Ettlia sp. Sci Rep 10(1):13947. 
                Diatom DNA amplification with degenerate primers for Illumina sequencingNistal A, Garcia P, Garcia J et al. (2021) DNA metabarcoding and morphological methods show complementary patterns in the metacommunity organization of lentic epiphytic diatoms. ARPHA Conference Abstracts 4: e63672. 
                Ion Torrent sequencing of bacterial 16S rRNA genesSchwarz SR, Hirsch S, Hiergeist A et al (2020) Limited antimicrobial efficacy of oral care antiseptics in microcosm biofilms and phenotypic adaptation of bacteria upon repeated exposure. Clin Oral Investig Epub ahead of print 
                Amplification of bacterial 16S rRNA gene using barcoded primers for Illumina sequencingWallis A, Yannuzzi IM, Choi MW (2021) Investigating the distribution of strains of Erwinia amylovora and streptomycin resistance in apple orchards in New York using CRISPR profiles: a six-year follow-up. Plant Dis Epub ahead of print. 
                Cancer research references
                UsagePublications
                PCR with genomic DNA of primary human cancer cells for mutation detectionAnwar SL, Hasemeier B, Schipper E et al. (2019) LINE-1 hypomethylation in human hepatocellular carcinomas correlates with shorter overall survival and CIMP phenotype. PLoS One 14(5):e0216374. 
                PCR with genomic DNA from whole blood samples, followed by Sanger sequencingArévalo-Jaramillo P, Idrobo A, Salcedo L (2019) Biochemical and genotoxic effects in women exposed to pesticides in Southern Ecuador. Environ Sci Pollut Res Int 26(24):24911–24921. 
                PCR of bisulfite-treated DNA from carcinoma cell lines, followed by Sanger sequencingIbrahim ML, Klement JD, Lu C et al. (2018) Myeloid-Derived Suppressor Cells Produce IL-10 to Elicit DNMT3b-Dependent IRF8 Silencing to Promote Colitis-Associated Colon Tumorigenesis. Cell Rep 25(11):3036–3046. 
                PCR with bisulfite-treated DNA from human tissues and cells, followed by Ion Torrent sequencingMa Y, Chai N, Jiang Q (2020) DNA methyltransferase mediates the hypermethylation of the microRNA 34a promoter and enhances the resistance of patient-derived pancreatic cancer cells to molecular targeting agents. Pharmacol Res 160:105071. 
                Touchdown PCR with DNA from FFPE samples, followed by Sanger SequencingMaier AD, Stenman A, Svahn F et al. (2021) TERT promoter mutations in primary and secondary WHO grade III meningioma. Brain Pathol 31(1):61–69. 
                Two-step RT-PCR with DNA from human cells for detection of transgene expressionMcCormick CA, Samuels TL, Battle MA (2021) H+/K+ATPase Expression in the Larynx of Laryngopharyngeal Reflux and Laryngeal Cancer Patients. Laryngoscope 131(1):130–135. 
                Amplification of an siRNA template from human cancer cell linesWang YL, Chang LC, Chen KB et al. (2021) Aptamer-guided targeting of the intracellular long-noncoding RNA HOTAIR. Am J Cancer Res 11(3):945–954. 
                “We had a very troublesome genotyping primer set that gave us bands in the NTC. Then we tried the Platinum II Taq [DNA Polymerase] and it worked the first time! We also save a ton of time on the PCR for this primer set since we no longer have to do two step PCR!”
                —Research Associate
                Large research university, US
                Education icon
                 PCR enzymes education resources
                Access PCR learning resources—a knowledge base of all things PCR.
                Webinar icon
                Molecular biology webinars
                View our webinars for current date to learn more about molecular biology topics.
                Video icon
                PCR enzymes videos
                Discover product features and tips to optimize your PCR experiments.
                Tools icon
                Support center
                Find tips, troubleshooting help, and resources for your end-point PCR and cDNA applications.
                Computer mouse icon
                Contact us
                Email or call our Technical Application Scientists for questions regarding PCR enzymes and master mixes.
                Check icon
                OEM and custom PCR products
                Discover custom manufacturing solutions, product labeling, and packaging capabilities tailored to your specifications.
                Stylesheet for Classic Wide Template adjustments
                For Research Use Only. Not for use in diagnostic procedures.