Enhancing capabilities and lifetime of existing instrument to drive innovation
Apreo Scanning Electron Microscope upgrades
Key benefits of upgrading Apreo 1 SEM to Apreo 2 SEM
  • Eliminate slow, tedious, manual processes with SmartAlign Technology, saving  up to 4 hours per week
  • Simplify the image tuning procedure with Flash technology and enhance the user experience
  • For low-vacuum models, the automated PLA loader enables switching between high vacuum and low vacuum without the need to open the chamber
  • Faster performance with new Windows 10 microscope PC
  • SEM IT infrastructure suited to latest updates
  • Improved cybersecurity and integration to MS OneDrive
Additional upgrades for Apreo scanning electron microscopes
ChemiSEM
  • In-depth elemental analysis (EDS), providing deeper insights into otherwise unnoticed elements
  • Live chemical quantification for the shortest time to elemental information
High-Res Gun aperture
  • Resolution boost with smaller gun aperture
Avizo 2D software
  • Advanced image processing, quantification, analysis, and reporting
  • Pre-built workflows and powerful image processing tools to get the desired answers form the imaging data
AutoScript 4 software
  • Powerful Python scripting (API) for DualBeams, allowing repetitive work to be done faster, more accurately, and with fewer mistakes
Maps 3 software
  • Completely automated solution for generating high-quality, multi-scale image of large overview while preserving the context of observations
  • Correlation of images from different microscopes, including imported images from third-party instruments using the integrated Bio-Formats Library
Verios scanning electron microscope upgrades
Key benefits of upgrading Verios G4 to Verios G5
Key benefits of upgrading Verios G4 to Verios G5
  • Eliminate slow, tedious, manual processes with SmartAlign Technology, saving  up to 4 hours per week
  • Simplify the image tuning procedure with Flash technology and enhance the user experience
  • Faster performance with new Windows 10 microscope PC
  • SEM IT infrastructure suited to latest updates
  • Improved cybersecurity and integration to MS OneDrive
Additional upgrades for Verios scanning electron microscopes
Avizo 2D software
  • Advanced image processing, quantification, analysis, and reporting
  • Pre-built workflows and powerful image processing tools to get the desired answers form the imaging data
AutoScript 4 software
  • Powerful Python scripting (API) for DualBeams, allowing repetitive work to be done faster, more accurately, and with fewer mistakes
Maps 3 software
  • Completely automated solution for generating high-quality, multi-scale image of large overview while preserving the context of observations
  • Correlation of images from different microscopes, including imported images from third-party instruments using the integrated Bio-Formats Library
Quattro scanning electron microscope upgrades
Key benefits of Quattro software upgrade (Windows 10)
Key benefits of Quattro software upgrade (Windows 10)
  • Faster performance with new Windows 10 microscope PC
  • SEM IT infrastructure suited to latest updates
  • Improved cybersecurity and integration to MS OneDrive
Additional upgrades for Quattro scanning electron microscopes
ChemiSEM
  • In-depth elemental analysis (EDS), providing deeper insights into otherwise unnoticed elements
  • Live chemical quantification for the shortest time to elemental information
Avizo 2D software
  • Advanced image processing, quantification, analysis, and reporting
  • Pre-built workflows and powerful image processing tools to get the desired answers form the imaging data
AutoScript 4 software
  • Powerful Python scripting (API) for DualBeams, allowing repetitive work to be done faster, more accurately, and with fewer mistakes
Maps 3 software
  • Completely automated solution for generating high-quality, multi-scale image of large overview while preserving the context of observations
  • Correlation of images from different microscopes, including imported images from third-party instruments using the integrated Bio-Formats Library
Axia ChemiSEM upgrades
Key benefits of upgrading ChemiSEM Advance
Key benefits of Axia ChemiSEM upgrade
  • Live quantitative elemental mapping
  • Flexible and easy to use, even for novice users
Additional upgrades for ChemiSEM
Avizo 2D software
  • Advanced image processing, quantification, analysis, and reporting
  • Pre-built workflows and powerful image processing tools to get the desired answers form the imaging data
AutoScript 4 software
  • Powerful Python scripting (API) for DualBeams, allowing repetitive work to be done faster, more accurately, and with fewer mistakes
Maps 3 software
  • Completely automated solution for generating high-quality, multi-scale image of large overview while preserving the context of observations
  • Correlation of images from different microscopes, including imported images from third-party instruments using the integrated Bio-Formats Library
Accessories
CleanConnect Sample Transfer System
CleanConnect Sample Transfer System
  • Sample integrity is preserved by the use of inert gas sample transfer
  • Ergonomic and modular design enables uncomplicated sample handling
  • Seamless, automatic integration to a variety of Thermo Scientific SEM/DualBeams
  • Compact design requiring no changes in system footprint or microscope specifications
  • Compatible with most glove box systems and available with optional direct glove box connectivity
Download datasheet
Cathodoluminescence_380x271
Cathodoluminescence Detector
  • Integrated RGB Cathodoluminescence Detector makes color images available as soon as the beam is scanning
  • Unique asset to multi-user labs requiring CL data and correlation with worry-free operation
Download datasheet
µHeater
µheater
  • Ultra-fast heating solution for in situ high resolution imaging
  • Fully integrated
  • Temperatures up to 1200 °C
Style Sheet for Komodo Tabs
Style Sheet for Global Design System
Contact us
Style Sheet for Support and Service footer
Style Sheet for Fonts
Style Sheet for Cards