Simple, fast, low-throughput Sanger sequencing and fragment analysis system
The Applied Biosystems SeqStudio Genetic Analyzer is a low-throughput, easy-to-use, and convenient benchtop system that delivers gold-standard Sanger sequencing technology and fragment analysis by capillary electrophoresis with a simple click. Just load your samples, click in the innovative all-in-one cartridge, and go.
The SeqStudio instrument is easy to use across a broad range of applications, including de novo, targeted, and plasmid Sanger sequencing, CRISPR-Cas9 genome editing confirmation, cell line authentication, and detection of single nucleotide polymorphisms (SNPs). Setup is simple and you will be ready to go in minutes. The SeqStudio Genetic Analyzer offers data quality, usability, service, and support at an affordable price.
Need higher throughput? Like the SeqStudio instrument, our  SeqStudio Flex Genetic Analyzer is an easy-to-use, flexible system for Sanger sequencing and fragment analysis, but with higher throughput.
SeqStudio 3D Tour
Learn more about financing options and programs. Learn more › 
Features
The SeqStudio Genetic Analyzer is designed for both new and experienced users who need simple and affordable Sanger sequencing and fragment analysis, without compromising performance or quality.
Innovative all-in-one cartridge
Just load your samples, insert the cartridge, and go— no need for technical experts for set-up or maintenance
Fast turnaround
Complete your run in as little as 30 minutes
Optimized end-to-end workflow
Single polymer system allows for Sanger sequencing and fragment analysis samples to be run on the same plate in the same workflow with no reconfiguration
Touchscreen usability
The intuitive touchscreen interface simplifies plate, sample, and run setup and displays results in a choice of formats
Connectivity with Thermo Fisher Connect
Manage your run anytime, anywhere from Thermo Fisher Scientific’s Connect cloud-based platform
Comprehensive analysis software
Primary and secondary analysis software modules are available for download or through Thermo Fisher Connect
Traceability and data security module
Optional security, audit, and electronic signature (SAE) software module offers an electronic data chain of custody to help ensure integrity of your data
SmartStart customer orientation
Every SeqStudio instrument purchase includes a one-day, on-site SmartStart Orientation training course to get you up and running quickly
Learn more about these features:
Applications
Sanger sequencing is the gold standard for sequencing technology, enabling a high degree of accuracy, long-read capability, and the flexibility to support a diverse range of applications in many research areas. Similarly, analysis of DNA fragments enables a multitude of applications, from genotyping to bacteria identification and from plant screening to gene expression profiling.
Below are examples of applications that can be run on the SeqStudio Genetic Analyzer. For more application information, see our SeqStudio Applications page.
De novo Sanger sequencing
Targeted Sanger sequencing
Plasmid sequencing
Oncology research
Microbial species identification
Next-generation sequencing (NGS) confirmation
CRISPR-Cas9 genome editing analysis
Human cell line authentication
Single-nucleotide polymorphism (SNP) genotyping
Analysis of human copy number variation (CNV)
DNA methylation analysis
Low-level somatic variant detection and analysis
Microsatellite instability (MSI) analysis
Repeat DNA expansion
SARS-CoV-2 identification
Learn more about these applications:
Optimized end-to-end workflow
The SeqStudio Genetic Analyzer can perform both Sanger sequencing and fragment analysis runs on the same plate within the same workflow. Gain flexibility in your laboratory schedule and help maximize your time by removing the need to batch samples.
Dual workflow.
The SeqStudio Genetic Analyzer can perform simultaneous Sanger sequencing and fragment analysis runs on the same plate.
Analysis of cell line contamination on the SeqStudio Genetic Analyzer.
We offer a complete product portfolio for every step of the Sanger sequencing and fragment analysis workflows. For Sanger sequencing, this includes amplifying the DNA template, PCR cleanup, cycle sequencing, sequencing cleanup, on‑instrument consumables, and data analysis. For fragment analysis, this includes DNA extraction, PCR amplification, on‑instrument consumables, and data analysis.
Videos, webinars, and demos
Specifications
Attribute
Description
Number of capillaries
4
Number of dyes
6
Sample format
96-well standard plate and standard 8-tube strips
Applications
  • Sanger sequencing (resequencing NGS confirmation, indels, heterozygote detection, minor variant detection, microbial identification, genome editing validation)
  • Fragment analysis (microsatellite analysis, compatible with MLPA, Applied Biosystems SNaPshot applications, cell line authentication
Secondary analysis software included
  • Sequence Analysis Software
  • SeqScape Software
  • Variant Reporter Software
  • GeneMapper Software
  • Minor Variant Finder Software
  • Sequencing Analysis Modules (Quality Check, Variant Analysis, and Next-Generation Confirmation) on Connect platform
  • Fragment Analysis Modules (Peak Scanner and Microsatellite Analysis) on Connect platform
  • SeqScreener Gene Editing App
Warranty
Included (1 year)
In-lab application training
Included
Services
Superior services and support for the SeqStudio Genetic Analyzer
Choose a service plan that’s right for you. Help maximize system uptime, reduce repair costs and turnaround time, extend the life of your instrument, and help keep it running at peak performance with one of our comprehensive service plans. Choose from a variety of service options that balance your budget, productivity, uptime, and regulatory requirements. When it comes to superior services and support for your SeqStudio Genetic Analyzer, we’ve got your back.
SmartStart Orientation enables your success
Every SeqStudio Genetic Analyzer includes a one-day, on-site SmartStart Orientation to get you up and running quickly. Led by a field applications scientist (FAS) at your location, this interactive training provides an overview of instrument operation and maintenance, data collection and analysis software, cloud connectivity, data troubleshooting, and more.
Comprehensive warranty and service plans protect your investment
SeqStudio Genetic Analyzers are protected by a standard one-year factory warranty that covers necessary costs for travel, labor, and parts. Extended-coverage service plans are available at the time of instrument purchase. Keep your laboratory running smoothly with preventive maintenance, proactive instrument monitoring, and fast response time that’s included with one of our superior service plans.
Service plan overview
Service aspect
AB Maintenance (Plus) plan
AB Assurance plan
AB Complete plan
On-site response time
Target 2 business days*
Guaranteed 2 business days*
Guaranteed next business day*
Scheduled on-site planned maintenance
Remote instrument diagnostics
Parts, labor, and travel
10% discount (US only)
Priority access to remote service engineer
Requalification, post-planned maintenance (PM) and critical repairs
Troubleshooting by field application scientist
*Response times vary by region
All service plans include scheduled planned maintenance and the latest software updates, enabling optimum reliability. Service plans help keep operating costs predictable, as parts, labor, and engineer travel are included or discounted.
Qualification services help ensure regulatory compliance
Instrument hardware qualifications for SeqStudio Genetic Analyzers include installation qualification (IQ), operational qualification (OQ), and instrument performance verification (IPV) services. Our qualification specialists will partner with you to deliver timely, cost-effective, and trusted qualification services that include reliable, audit-style documentation to help ensure your SeqStudio Genetic Analyzer meets regulatory requirements.
Resources
Ordering information
Catalog # Name Size Price (USD) Qty
A35644 Each -
A35645 Each -
A33671 Each
2,274.65
Online Exclusive
2,365.00
Save 90.35 (4%)
A41331 Each
2,578.65
Online Exclusive
2,700.00
Save 121.35 (4%)
A33401 Each
162.00
A31923 Each
11.07
A38073 Each
1,869.65
Online Exclusive
1,974.00
Save 104.35 (5%)
For Research Use Only. Not for use in diagnostic procedures.
*This offer is only available to customers in Canada and the United States (excluding Puerto Rico) who acquire manufactured products and service plans from Thermo Fisher Scientific or its affiliates. Some exclusions may apply. Offer will apply to approved applicants and who sign a Thermo Fisher Financial Services contract that is received by Thermo Fisher Financial Services no later than December 31, 2022. Customers must submit a Thermo Fisher Financial Services credit application and be approved for credit. Interest rates vary based on credit rating. Not all buyers may qualify for credit approval or promotional rate. Determination of eligibility is at the sole discretion of Thermo Fisher Financial Services credit committee. Financing shall be subject to contract terms and conditions. Cannot be combined with other discounts or promotions.  Offer void where prohibited, licensed or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Offer subject to change without notice. Other restrictions may apply.
1x1 image pixel for data collection