SeqStudio Flex Genetic Analyzers
Flexibility, usability, connectivity, and serviceability in a mid-throughput genetic analyzer
The Applied Biosystems SeqStudio Flex Series genetic analyzers deliver the high level of Sanger sequencing and fragment analysis data quality and reliable performance that scientists need, while leveraging design improvements and technological advances for increased flexibility, easier operation, enhanced connectivity, and remote serviceability.
With a wide range of applications from simple, targeted sequencing to identification of the latest SARS variant of concern, SeqStudio Flex genetic analyzers help you to achieve freedom and peace of mind, providing extra time in your day to do what you do best: answering the big scientific questions. 
Need lower throughput? Like the SeqStudio Flex instrument, our SeqStudio Genetic Analyzer is an easy-to-use, flexible system for Sanger sequencing and fragment analysis, but designed for lower throughput.
SeqStudio Flex 3D Tour
Learn more about financing options and programs. Learn more › 
Highlights
Available in both 8- and 24-capillary configurations, SeqStudio Flex Series genetic analyzers are designed with scientists in mind.
Sample loading flexibility
Four-plate capacity, continuous plate loading, and urgent sample reprioritization provide both the largest mid-throughput capacity and complete scheduling flexibility, translating into extra time in your day.
Ease of use
Easier capillary array installation, streamlined one-button start up, autocalibration, and an onboard learning center offer users of any skill level greater usability and streamlined training whether they work in a single- or multi-user environment.
Standalone instrument
An integrated touchscreen computer with intuitive software and walk-through wizards for simplfied sample mapping, plate linking, and run setup. 
Remote setup, monitoring, and data transfer
Our Thermo Connect Platform provides enhanced flexibility and simplified collaboration by allowing users to stay connected to their instrument and data, remotely and safely, from anywhere.
Remote troubleshooting
Smart Help and Remote Support capabilities reduce instrument downtime and resolve up to 70% of issues without a service visit using immersive, real-time audio/video collaboration tools and push-button support requests.
Advanced communications and connectivity
Integrating the communication technologies of today, such as voice commands and a wide array of connectivity options, to propel laboratory efficiencies of tomorrow.
Applications
SeqStudio Flex genetic analyzers deliver high-level Sanger sequencing and fragment analysis performance using capillary electrophoresis to support a diverse range of applications, including:
Targeted sequencing
NGS confirmation
Cell line authentication
Minor variant finding
Gene editing confirmation
Multiplexed PCR
Microbe or virus identification
DNA methylation analysis
SNP genotyping
Microsatellite instability (MSI) analysis
dsDNA quality control
Plasmid sequencing
Featured applications
Multiplexed PCR analysis
While quantitative PCR is the mainstay for rapid and concurrent analysis of several genomic sequences, it can be limited by the number of unique fluorophores, and therefore, the number of genomic targets that can be queried. In situations where a high multiplexing capability is desired, standard PCR coupled with fragment analysis by capillary electrophoresis offers a versatile solution to detect multiple targets in a single reaction as it separates amplicons based on both size and fluor in a single capillary allowing a very high degree of multiplexing.
dsDNA QC
It is often necessary to run a quality check (QC) on a starting pool of double-stranded DNA (dsDNA) before initiating genomic analysis or continuing to the next step. For example, DNA extracted from FFPE-preserved tissue may be fragmented and too small to provide useful information, so it’s useful to know the overall size before proceeding.
Optimized end-to-end workflow
Thermo Fisher Scientific is committed to helping you improve data quality and drive laboratory efficiency for all your Sanger sequencing and fragment analysis applications, by providing everything you need for successful capillary electrophoresis (CE) separations from sample preparation through data analysis. We focus on quality and providing fully validated workflow solutions so that you can focus on your research.
Webinars and videos
Targeted Sequence Analysis to Propel Biopharmaceutic Development
Unlocking the Genomics of Cancer Using Sanger Sequencing and Fragment Analysis
From Pathogen identification to disease prevention and cure
Advanced Digital Services and Support Ecosystem for your SeqStudio Flex Genetic Analyzer
Services
Superior services and support for the SeqStudio Flex Genetic Analyzer
Keep advancing with new technology. We’ve got your back.
SeqStudio Flex genetic analyzer with Service logo
Choose a service plan that’s right for you. Maximize system uptime, reduce repair costs and turnaround time, extend the life of your instrument, and help keep it running at peak performance with one of our comprehensive service plans. Choose from a variety of service options that balance your budget, productivity, uptime, and regulatory requirements. When it comes to superior services and support for your SeqStudio Flex Genetic Analyzer, we’ve got your back.
Instrument specifications
Number of capillaries
8 or 24
Number of dyes
8-dye capable
Capacity
4-plate capacity; 96-well plates, 384-well plates, 8-tube strips 
Dimensions (W x D x H)
70 x 67.5 x 86.5 cm (27.6 x 26.6 x 34.1 in.)
Weight
115 kg (253.5 lb)
Power input
100–240 V
Internal hard drive
512 GB solid state drive (SSD)
Excitation source
505 nm solid state laser
On-instrument tracking
Radio frequency identification (RFID), internal barcode reader
Communication interface
Connect cloud-based platform; USB port for Wi-Fi dongle; 3 RJ-45 ethernet ports
Configuration
Standalone; optional desktop or laptop computer available
Warranty
Standard one-year warranty included; extended warranties available
SmartStart Orientation
Includes one day of onsite training by a field application scientist
Remote troubleshooting
Smart Help and Remote Support capabilities enable fast and more effective resolution of instrument issues for reduced instrument downtime
Hands-free operation
Voice command capability to perform basic instrument operations
Resources
Ordering information
For Research Use Only. Not for use in diagnostic procedures.
*This offer is only available to customers in Canada and the United States (excluding Puerto Rico) who acquire manufactured products and service plans from Thermo Fisher Scientific or its affiliates. Some exclusions may apply. Offer will apply to approved applicants and who sign a Thermo Fisher Financial Services contract that is received by Thermo Fisher Financial Services no later than December 31, 2022. Customers must submit a Thermo Fisher Financial Services credit application and be approved for credit. Interest rates vary based on credit rating. Not all buyers may qualify for credit approval or promotional rate. Determination of eligibility is at the sole discretion of Thermo Fisher Financial Services credit committee. Financing shall be subject to contract terms and conditions. Cannot be combined with other discounts or promotions.  Offer void where prohibited, licensed or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Offer subject to change without notice. Other restrictions may apply.