14 Sep 2021
How to Overlay Images Using the Color Wash Module
This short video demonstrates how to use the Color Wash module to overlay label colors onto a greyscale image.
The Color Wash module can be used to overlay two scalar fields. This module can be very useful for simultaneously visualizing a greyscale image and the label field image resulting from its segmentation.
This tutorial shows how to set the Color Wash module to overlay a label field's material colors onto a greyscale image and adjust options to edit the blending of the images.
Additional resources: