Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IQD29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335144.1 | NP_001324495.1 | 2 - 3 | 722 | 72 | |
NM_001335145.1 | NP_001324494.1 | 2 - 3 | 265 | 72 | ||
NM_126334.3 | NP_178382.2 | 2 - 3 | 249 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |