Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CELA2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174714.2 | NP_777139.1 | 4 - 5 | 362 | 59 | |
GenBank mRNA | BC134791.1 | - | 4 - 5 | 375 | 59 | |
X97635.1 | - | 362 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |