Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ifc-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270046.2 | NP_001256975.1 | 6 - 7 | 1507 | 73 | |
NM_001374936.2 | NP_001362085.1 | 19 - 20 | 3606 | 73 | ||
NM_001380893.1 | NP_001367765.1 | 16 - 17 | 3387 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |