Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pik-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268813.4 | NP_001255742.1 | 5 - 6 | 923 | 101 | |
NM_001268814.3 | NP_001255743.1 | 3 - 4 | 720 | 101 | ||
NR_056884.1 | 5 - 6 | 951 | 101 | |||
GenBank mRNA | AF348167.1 | - | 4 - 5 | 867 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |