Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lime1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003503155.2 | XP_003503203.2 | 1 - 2 | 1380 | 68 | |
XM_007643775.1 | XP_007641965.1 | 2 - 3 | 594 | 68 | ||
XM_007643776.1 | XP_007641966.1 | 1 - 2 | 1380 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Probe spans exons |