Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005689335.2 | XP_005689392.1 | 2 - 3 | 437 | 73 | |
XM_013969389.2 | XP_013824843.1 | 2 - 3 | 437 | 73 | ||
GenBank mRNA | JO625511.2 | - | 2 - 3 | 494 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Capra hircus Probe spans exons |