Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cad74A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259885.1 | NP_001246814.1 | 7 - 8 | 4809 | 69 | |
NM_140716.2 | NP_648973.1 | 7 - 8 | 4523 | 69 | ||
NM_168724.2 | NP_730261.1 | 7 - 8 | 4538 | 69 | ||
GenBank mRNA | BT150474.1 | - | 7 - 8 | 4537 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |