Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ftr82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075103.1 | NP_001068571.1 | 5 - 6 | 1263 | 62 | |
GenBank mRNA | BC096901.1 | - | 4 - 5 | 544 | 62 | |
BC124299.1 | - | 5 - 6 | 1263 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |