Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTSO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031129.1 | NP_001026300.1 | 3 - 4 | 414 | 75 | |
XM_015285286.2 | XP_015140772.1 | 3 - 4 | 990 | 75 | ||
GenBank mRNA | AJ719384.1 | - | 3 - 4 | 414 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons |