Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC30B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005573579.2 | XP_005573636.1 | 1 | 2895 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |