Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bckdk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009739.3 | NP_033869.1 | 7 - 8 | 906 | 64 | |
GenBank mRNA | AF043070.1 | - | 6 - 7 | 747 | 64 | |
AK075602.1 | - | 7 - 8 | 832 | 64 | ||
AK145479.1 | - | 7 - 8 | 877 | 64 | ||
AK150607.1 | - | 7 - 8 | 898 | 64 | ||
AK150723.1 | - | 7 - 8 | 898 | 64 | ||
AK167519.1 | - | 7 - 8 | 898 | 64 | ||
BC004077.1 | - | 4 - 5 | 334 | 64 | ||
BC046595.1 | - | 7 - 8 | 876 | 6313106 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |