Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sfxn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053196.3 | NP_444426.3 | 2 - 3 | 273 | 103 | |
GenBank mRNA | AF325261.1 | - | 2 - 3 | 231 | 103 | |
AK034875.1 | - | 273 | 103 | |||
AK149277.1 | - | 242 | 103 | |||
AK150722.1 | - | 2 - 3 | 224 | 103 | ||
AK162407.1 | - | 2 - 3 | 273 | 103 | ||
BC019808.1 | - | 2 - 3 | 271 | 5043343 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |