Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thumpd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008188.2 | NP_032214.1 | 3 - 4 | 427 | 87 | |
GenBank mRNA | AK017246.1 | - | 3 - 4 | 440 | 87 | |
AK142140.1 | - | 3 - 4 | 446 | 87 | ||
BC002024.1 | - | 3 - 4 | 429 | 3591186 | 87 | |
BC012688.1 | - | 2 - 3 | 375 | 3987438 | 87 | |
BC046800.1 | - | 3 - 4 | 427 | 6403880 | 87 | |
U83176.1 | - | 3 - 4 | 421 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |