Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stx12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133887.4 | NP_598648.1 | 3 - 4 | 379 | 59 | |
GenBank mRNA | AB019211.1 | - | 3 - 4 | 322 | 59 | |
AK076390.1 | - | 364 | 59 | |||
AK077678.1 | - | 3 - 4 | 380 | 59 | ||
AK078948.1 | - | 3 - 4 | 339 | 59 | ||
AK146544.1 | - | 3 - 4 | 384 | 59 | ||
AK146735.1 | - | 3 - 4 | 371 | 59 | ||
AK167087.1 | - | 3 - 4 | 293 | 59 | ||
AK168753.1 | - | 3 - 4 | 337 | 59 | ||
AK170203.1 | - | 3 - 4 | 416 | 59 | ||
BC010669.1 | - | 3 - 4 | 366 | 2645010 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |