Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ice1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144837.3 | NP_659086.2 | 17 - 18 | 6457 | 57 | |
GenBank mRNA | AK129244.1 | - | 5 - 6 | 4894 | 57 | |
AK134564.1 | - | 5 - 6 | 2900 | 57 | ||
BC018507.1 | - | 4 - 5 | 552 | 57 | ||
BC026613.1 | - | 5 - 6 | 983 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |