Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc9a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK017792.1 | - | 1 - 2 | 257 | 63 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA 7656 | Mir7656 |
Gm27614, mmu-mir-7656 |
-369.8kb | Sense | Chr. 9 - 94751829 - 94751890 on NCBI Build GRCm38 |
Downstream | carbohydrate sulfotransferase 2 | Chst2 |
AI428561, AW121776, C130041E03Rik, Chts2, GST-2, GlcNAc6ST, Gn6st |
278.9kb | Anti-sense | Chr. 9 - 95400926 - 95407270 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus 1700 Probe spans exons |