Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam20a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153782.1 | NP_722477.1 | 8 - 9 | 1758 | 78 | |
GenBank mRNA | AK142932.1 | - | 8 - 9 | 1783 | 78 | |
BC011274.1 | - | 8 - 9 | 1662 | 78 | ||
BC023310.1 | - | 1 - 2 | 59 | 78 | ||
BC029169.1 | - | 8 - 9 | 1758 | 4947019 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |