Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pmaip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021451.2 | NP_067426.1 | 1 - 2 | 212 | 74 | |
GenBank mRNA | AB041230.1 | - | 1 - 2 | 212 | 74 | |
AK043856.1 | - | 1 - 2 | 243 | 74 | ||
AK088556.1 | - | 1 - 2 | 222 | 74 | ||
AK143990.1 | - | 1 - 2 | 284 | 74 | ||
AK169914.1 | - | 1 - 2 | 256 | 74 | ||
BC050821.1 | - | 1 - 2 | 199 | 6517820 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |