Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudt6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291044.1 | NP_001277973.1 | 2 - 3 | 463 | 155 | |
GenBank mRNA | AF453427.1 | - | 2 - 3 | 572 | 155 | |
AF453428.1 | - | 2 - 3 | 449 | 155 | ||
AK145299.1 | - | 2 - 3 | 455 | 155 | ||
BC146000.1 | - | 2 - 3 | 457 | 40131303 | 155 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |