Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnj14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_170718.1 | NP_733836.1 | 1 - 2 | 1277 | 96 | |
GenBank mRNA | AJ003065.1 | - | 1 - 2 | 1277 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Ion Channel 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |