Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1r16b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191072.1 | NP_001178001.1 | 4 - 5 | 645 | 63 | |
XM_008762383.1 | XP_008760605.1 | 4 - 5 | 729 | 63 | ||
XM_008762384.1 | XP_008760606.1 | 4 - 5 | 1520 | 63 | ||
XM_008762385.1 | XP_008760607.1 | 4 - 5 | 1908 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |