Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: shq1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079148.1 | NP_001072616.1 | 7 - 8 | 891 | 87 | |
XM_012961357.2 | XP_012816811.1 | 8 - 9 | 1031 | 87 | ||
GenBank mRNA | BC123006.1 | - | 7 - 8 | 891 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |