The alkylation of phthalimide with alkyl halides was first reported in 1884, but in 1887 S. Gabriel realized that the process was a general one and developed a synthesis for primary amines. Henceforward, the mild two-step process for the preparation of primary amines from their corresponding alkyl halides, i.e., alkylation followed by hydrolysis, became known as the Gabriel synthesis.
The alkylation step can take place either neat or in the presence of a solvent. If a solvent is used, dimethyl formamide (DMF) is often considered the best choice; however, other solvents can be used including dimethyl sulfoxide (DMSO), Hexamethyl phosporamide (HMPA), acetonitrile, and ethylene glycol.
Several issues limited the use of the original Gabriel synthesis. Firstly, when the reaction of the potassium phthalimide and alkyl halide required high temperatures, heat-sensitive substrates could not be used. Secondly, the hydrolysis step was usually performed in the presence of a strong acid such as sulfuric, hydrobromic, or hydroiodic acids, further preventing the use of substrates that were sensitive to acid conditions. Alternatively, where strong alkali could be used for hydrolysis, base-sensitive functional groups had to be excluded.
In 1926, H.R. Ing and R.H.F. Manske developed a modification using hydrazine hydrate in refluxing ethanol to cleave the N-alkylphthalimide under milder and neutral conditions, which became known as the Ing-Manske procedure.
Since then, several other modifications have been developed, such as the development of novel Gabriel reagents that replace the phthalimide with other nitrogen sources, the addition of catalytic amounts of crown ether or a cryptand to improve yields, and the use of sodium borohydride in isopropyl alcohol (IPA) for exceptionally mild cleavage of the phthalimide.
The total synthesis of peramine, an alkaloid produced by a fungus that protects grasses against grazing by mammals and insects, successfully employed the Gabriel synthesis in the latter stages.
Mechanism of the Gabriel synthesis reaction
Review available Thermo Scientific products for the Gabriel synthesis reaction:
Catalog # Name Size Price (USD)
A12584.36 Phthalimide, 99% Each
41.65
Online Exclusive
45.70
Save 4.05 (9%)
A12584.0E Phthalimide, 99% Each
85.65
Special offer
Online exclusive
Ends: 26-Sep-2025
101.00
Save 15.35 (15%)
A12584.0C Phthalimide, 99% Each
249.65
Special offer
Online exclusive
Ends: 26-Sep-2025
293.00
Save 43.35 (15%)
A11134.22 Potassium phthalimide, 98+% Each
30.65
Special offer
Online exclusive
Ends: 30-Jun-2025
35.80
Save 5.15 (14%)
A11134.36 Potassium phthalimide, 98+% Each
95.65
Online Exclusive
106.00
Save 10.35 (10%)
A11134.0E Potassium phthalimide, 98+% Each
353.65
Special offer
Online exclusive
Ends: 30-Jun-2025
416.00
Save 62.35 (15%)
A14005.18 Hydrazine monohydrate, 98+% Each
A14005.30 Hydrazine monohydrate, 98+% Each
A14005.36 Hydrazine monohydrate, 98+% Each
A14005.0E Hydrazine monohydrate, 98+% Each
16651.36 Each
38788.36
13432.36
88983.36
L20325.06 Di-tert-butyl iminodicarboxylate, 98% Each
98.65
Online Exclusive
109.00
Save 10.35 (9%)
L20325.14 Di-tert-butyl iminodicarboxylate, 98% Each
324.65
Online Exclusive
361.00
Save 36.35 (10%)
Vote for your favorite named reaction
Learn more about the most popular named reactions based on feedback from our customers. Do you have a favorite reaction? Let us know!
Vote now