Hantzsch dihydropyridine synthesis – a heterocycle formation reaction
In 1882, A. Hantzsch condensed two equivalents of ethyl acetoacetate with one of acetaldehyde and ammonia to obtain what he believed at the time to be a 2,3-dihydropyridine but was later found to be a 1,4-dihydropyridine. Since then, the one-pot condensation of a beta-keto ester or a 1,3-dicarbonyl compound with an aldehyde and ammonia to prepare 1,4-dihydropyridines is known as the Hantzsch dihydropyridine synthesis.
Often the 1,4-dihydropyridines spontaneously oxidize to the corresponding substituted pyridines, but in the case of more stable variants, oxidizing agents are used as necessary, e.g., HNO2, HNO3, MnO2, etc.
The original procedure only provided symmetrical products. However, several modifications afford unsymmetrical dihydropyridines; for example, utilizing the Knoevenagel modification, various substituted 1,5-dicarbonyl compounds can be prepared.
Making use of the Hantzsch dihydropyridine synthesis, M. Baley reported the first synthesis of an unsymmetrical 2, 2-6’2”-terpyridine.
This reaction has many applications. Commercially, 1,4-dihydropyridines are an important class of calcium channel blockers that help reduce blood pressure in patients with hypertension, e.g., nifedipine.
Review available Thermo Scientific products for the Hantzsch dihydropyridine synthesis:
Catalog # Name Size Price (USD) Qty
A12544.30 Ethyl acetoacetate, 99+% Each
24.65
Special offer
Online exclusive
Ends: 26-Sep-2025
28.70
Save 4.05 (14%)
A12544.36 Ethyl acetoacetate, 99+% Each
29.65
Special offer
Online exclusive
Ends: 26-Sep-2025
35.10
Save 5.45 (16%)
A12544.0E Ethyl acetoacetate, 99+% Each
84.65
Special offer
Online exclusive
Ends: 26-Sep-2025
98.90
Save 14.25 (14%)
A12544.0C Ethyl acetoacetate, 99+% Each
289.65
Special offer
Online exclusive
Ends: 30-Jun-2025
340.00
Save 50.35 (15%)
L15215.AP Acetaldehyde, 99% Each -
L15215.K2 Acetaldehyde, 99% Each
190.00
L15215.K4 Acetaldehyde, 99% Each -
H27080.AE Ammonia, 2M in methanol Each -
H27080.AU Ammonia, 2M in methanol Each -
H30382.AE Ammonia, 7M in methanol Each -
H30382.AU Ammonia, 7M in methanol Each -
A11629.14 Ethyl 2-methylacetoacetate, 95% Each
43.65
Online Exclusive
48.80
Save 5.15 (11%)
A11629.22 Ethyl 2-methylacetoacetate, 95% Each
113.65
Special offer
Online exclusive
Ends: 30-Jun-2025
133.00
Save 19.35 (15%)
A11629.36 Ethyl 2-methylacetoacetate, 95% Each
382.65
Special offer
Online exclusive
Ends: 30-Jun-2025
450.00
Save 67.35 (15%)
A10765.36 Manganese(IV) oxide, 97% Each
122.65
Online Exclusive
136.00
Save 13.35 (10%)
A10765.0E Manganese(IV) oxide, 97% Each
159.65
Special offer
Online exclusive
Ends: 26-Sep-2025
188.00
Save 28.35 (15%)
A10765.A9 Manganese(IV) oxide, 97% Each
446.65
Special offer
Online exclusive
Ends: 30-Jun-2025
525.00
Save 78.35 (15%)
Vote for your favorite named reaction
Learn more about the most popular named reactions based on feedback from our customers.  Do you have a favorite reaction?  Let us know!