Oncomine-Comp-Assay-Plus-shwocase3
Enabling large genomic rearrangement detection in BRCA 1 and BRCA 2 genes in as little as 24 hours
The Oncomine BRCA Assay uniquely empowers laboratories to detect all classes of mutations in one NGS workflow, removing the need to employ multiple technologies.
Request more information
Download white paper ›        Download flyer ›
The Oncomine BRCA Assay enables robust performance and reliable, rapid and consistent high-quality results from every sample.
  • Based on proven Ion AmpliSeq technology
  • Fully qualified on clinical research samples
  • Requires only 20 ng DNA input
  • Provides 100% exonic coverage, including flanking intronic sequences (average of 64 bases in either direction), superior uniformity, and high reads (average is >600)
  • Detects 5% and below minor-allele frequencies
  • Flexible workflow enables full automation
  • Detect large genomic rearrangement, such as large insertions and deletions (indels), and exon-level duplication and deletion
1557237760999
White Paper: Evaluation of the Oncomine BRCA Research Assay for variant detection by next-generation sequencing
Learn how the Oncomine BRCA Research Assay offers sensitive, specific detection of SNVs, MNVs, indels, and whole-exon, multiple-exon, or entire-gene aberrations.
Download white paper ›
Oncomine BRCA Assay workflow
The Ion Torrent Oncomine BRCA Assay is now available for use on the Ion GeneStudio systems with the Ion Chef Instrument or for use on the Ion Torrent Genexus System.
COL110993-Machines-V4
Figure 1. Ion Torrent Next Generation Sequencing systems
Oncomine BRCA Assay performance
The associated figures demonstrate the superior performance of the Oncomine BRCA Assay. All exons are 100% covered, with an average of 64 bases of flanking sequence into the introns upstream and downstream of each exon, allowing for over 99% confidence of detecting 5% somatic variants. The uniformity and high read  counts ensure high sensitivity and accuracy of both somatic and germline mutation detection, demonstrated with different workflows (and sequencers). The performance has been verified for use on the Ion 530 chips on GeneStudio system and GX5 chip on Genexus system.
Figure 2. 100% exon coverage across both BRCA 1 and BRCA 2 genes, with high uniformity and read counts across all exons, allowing for over 99% confidence of detecting 5% somatic variant.
Control sample variants
Platform
Library
SNV
Indel
% Sensitivity
% PPV
% Sensitivity
% PPV
5% allele frequency
GeneStudio S5 System
Manual
100
98
98
92
Chef
100
92
99
99
Genexus System
Genexus
100
100
98
100
50%, 100% allele frequency
GeneStudio S5 System
Manual
100
100
100
100
Chef
100
100
100
100
Genexus System
Genexus
100
100
94
100
Figure 3. Superior accuracy in detecting somatic and germline variants that is highly consistent and independent of workflow. At 5% allele frequency, >1,000 SNV and >600 indel variants measured. At 50%, 100% allele frequency, >4,000 SNV and >200 indel variants measured. Positive predictive value (PPV) = true positives/total number of positives. Sensitivity = true positive/(true positives + false positives).
BRCA-Figure-4-Chart
Figure 4. Relative abundance of BRCA exons are plotted. The sample has a deletion in BRCA1 (red) of exons 4–9 (green circle). BRCA2 (blue) has no CNV. The green plot indicates the sample ID amplicons used for normalization.z
Ordering Information