c7c8 - Key/Specs/Resources/Appl/Tech/Doc/Contact
Phenom Pro Desktop SEM
The sixth-generation of Thermo Scientific Phenom Pro G6 Desktop SEM fills the gap between light microscopy and floor-model SEM analysis, thus expanding the capabilities of research facilities.
Fast and easy to use, the Phenom Pro G6 Desktop SEM can be used to relieve the burden of routine analysis for common samples from floor-model SEM instruments. Instrument configuration and the sample loading mechanism ensure quick imaging with minimal time spent tuning between experiments.
Facility users of any experience level can quickly begin producing high-quality results with the Phenom Pro G6 Desktop SEM. Its long-lifetime CeB6 source offers high brightness while requiring low maintenance. Additionally, its high stability and small form factor allow the instrument to be used in practically any lab environment; more simply put, it does not require specialized infrastructure or expert oversight.
Key Features
Effortless
The color navigation camera in the Phenom Pro Desktop SEM provides information that helps you make the link between optical and electron optical images. Users are ready to take images after only 10 minutes of basic training. A large variety of sample holders is available to accommodate a range of applications. Sample loading is fast and easy thanks to our patented sample vacuum loading technology.
Phenom ProSuite Software
Thermo Scientific Phenom ProSuite Software is an optional software application platform that has been developed to further enhance the capabilities of the Phenom desktop SEM. Phenom ProSuite Software enables maximum information to be extracted from images obtained on the Phenom Pro Desktop SEM. 
Secondary electron detector
A secondary electron detector (SED) is optionally available on the Phenom Pro Desktop SEM. The SED collects low-energy electrons from the top surface layer of the sample. It is therefore the perfect choice to reveal detailed sample surface information. 
ChemiSEM Technology
Thermo Scientific ChemiSEM Technology revolutionizes and simplifies EDS analysis by fully integrating SEM and EDS functions into a single, cohesive user interface. Based on live quantification and building on decades of expertise in EDS analysis, the technology provides elemental information quickly and easily, guaranteeing reliable results in less time. ChemiSEM Technology now comes with a powerful new feature: ChemiPhase. ChemiPhase identifies unique phases with a big data approach, finding minor and trace elements while eliminating user bias and reducing possible mistakes.
Learn more
Specifications
Style Sheet for Products Table Specifications
Light optical magnification
  • 20-134x
Electron optical magnification range
  • 160-350,000x
Resolution
  • ≤ 6 nm SED and ≤ 8 nm BSD
Digital zoom
  • Max. 12x
Light optical navigation camera
  • Color
Acceleration voltages
  • Default: 5 kV, 10 kV and 15 kV
  • Advanced mode: adjustable range between 4.8 kV and 20.5 kV imaging mode
Vacuum modes
  • High vacuum mode
  • Charge reduction mode via optional low vacuum sample holder
Detector
  • Backscattered electron detector (standard)
  • Energy dispersive X-ray spectroscopy detector (optional)
  • Secondary electron detector (optional)
Sample size
  • Up to 25 mm diameter
  • 32 mm (optional)
Sample height
  • Up to 35 mm
  • 100 mm (optional)
Style Sheet for Komodo Tabs
Resources
Video Player is loading.
Current Time 0:00
Duration 3:07
Loaded: 5.24%
Stream Type LIVE
Remaining Time 3:07
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Webinar: Scanning electron microscopy: selecting the right technology for your needs
    This on-demand webinar has been designed to help you decide which SEM best meets your unique needs. We present an overview of Thermo Fisher Scientific SEM technology for multi-user research labs and focus on how these wide-ranging solutions deliver performance, versatility, in situ dynamics and faster time to results. Watch this webinar if you are interested in:
    • How the needs for different microanalysis modalities are met (EDX, EBSD, WDS, CL, etc.).
    • How samples are characterized in their natural state without the need for sample preparation.
    • How new advanced automation allows researchers to save time and increase productivity.
    Video Player is loading.
    Current Time 0:00
    Duration 3:07
    Loaded: 5.24%
    Stream Type LIVE
    Remaining Time 3:07
    1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
      • en (Main), selected
      Webinar: Scanning electron microscopy: selecting the right technology for your needs
      This on-demand webinar has been designed to help you decide which SEM best meets your unique needs. We present an overview of Thermo Fisher Scientific SEM technology for multi-user research labs and focus on how these wide-ranging solutions deliver performance, versatility, in situ dynamics and faster time to results. Watch this webinar if you are interested in:
      • How the needs for different microanalysis modalities are met (EDX, EBSD, WDS, CL, etc.).
      • How samples are characterized in their natural state without the need for sample preparation.
      • How new advanced automation allows researchers to save time and increase productivity.
      Applications
      Process control using electron microscopy
      Process control using electron microscopy
      Modern industry demands high throughput with superior quality, a balance that is maintained through robust process control. SEM and TEM tools with dedicated automation software provide rapid, multi-scale information for process monitoring and improvement.
      Quality control and failure analysis using electron microscopy
      Quality control and failure analysis
      Quality control and assurance are essential in modern industry. We offer a range of EM and spectroscopy tools for multi-scale and multi-modal analysis of defects, allowing you to make reliable and informed decisions for process control and improvement.
      Fundamental Materials Research_R&D_Thumb_274x180_144DPI
      Fundamental Materials Research
      Novel materials are investigated at increasingly smaller scales for maximum control of their physical and chemical properties. Electron microscopy provides researchers with key insight into a wide variety of material characteristics at the micro- to nano-scale.
      Techniques
      EDS Elemental Analysis
      Thermo Scientific Phenom Elemental Mapping Software provides fast and reliable information on the distribution of chemical elements within a sample.
      Learn more ›
      3D EDS Tomography
      Modern materials research is increasingly reliant on nanoscale analysis in three dimensions. 3D characterization, including compositional data for full chemical and structural context, is possible with 3D EM and energy dispersive X-ray spectroscopy.
      Learn more ›
      Atomic-Scale Elemental Mapping with EDS
      Atomic-resolution EDS provides unparalleled chemical context for materials analysis by differentiating the elemental identity of individual atoms. When combined with high-resolution TEM, it is possible to observe the precise organization of atoms in a sample.
      Learn more ›
      Imaging Hot Samples
      Studying materials in real-world conditions often involves working at high temperatures. The behavior of materials as they recrystallize, melt, deform, or react in the presence of heat can be studied in situ with scanning electron microscopy or DualBeam tools.
      Learn more ›
      In Situ experimentation
      Direct, real-time observation of microstructural changes with electron microscopy is necessary to understand the underlying principles of dynamic processes such as recrystallization, grain growth, and phase transformation during heating, cooling, and wetting.
      Learn more ›
      Multi-scale analysis
      Novel materials must be analyzed at ever higher resolution while retaining the larger context of the sample. Multi-scale analysis allows for the correlation of various imaging tools and modalities such as X-ray microCT, DualBeam, Laser PFIB, SEM and TEM.
      Learn more ›
      EDS Elemental Analysis
      Thermo Scientific Phenom Elemental Mapping Software provides fast and reliable information on the distribution of chemical elements within a sample.
      Learn more ›
      3D EDS Tomography
      Modern materials research is increasingly reliant on nanoscale analysis in three dimensions. 3D characterization, including compositional data for full chemical and structural context, is possible with 3D EM and energy dispersive X-ray spectroscopy.
      Learn more ›
      Atomic-Scale Elemental Mapping with EDS
      Atomic-resolution EDS provides unparalleled chemical context for materials analysis by differentiating the elemental identity of individual atoms. When combined with high-resolution TEM, it is possible to observe the precise organization of atoms in a sample.
      Learn more ›
      Imaging Hot Samples
      Studying materials in real-world conditions often involves working at high temperatures. The behavior of materials as they recrystallize, melt, deform, or react in the presence of heat can be studied in situ with scanning electron microscopy or DualBeam tools.
      Learn more ›
      In Situ experimentation
      Direct, real-time observation of microstructural changes with electron microscopy is necessary to understand the underlying principles of dynamic processes such as recrystallization, grain growth, and phase transformation during heating, cooling, and wetting.
      Learn more ›
      Multi-scale analysis
      Novel materials must be analyzed at ever higher resolution while retaining the larger context of the sample. Multi-scale analysis allows for the correlation of various imaging tools and modalities such as X-ray microCT, DualBeam, Laser PFIB, SEM and TEM.
      Learn more ›
      Style Sheet to change H2 style to p with em-h2-header class
      Style Sheet to change H3 to p with em-h3-header class
      Contact us
      Electron microscopy services for
      the materials science
      To ensure optimal system performance, we provide you access to a world-class network of field service experts, technical support, and certified spare parts.
      Style Sheet for Support and Service footer
      Style Sheet for Fonts
      Style Sheet for Cards