Speciation Analysis by IC-ICP-MS
Trace elemental species separation and detection
IC-ICP-MS is an ideal method for identifying different versions of a molecule, which can be important in determining a sample’s safety or toxicity.
On this page
What is speciation analysis?
Speciation is the process of separating and quantifying different molecular versions of a compound, which can exhibit very different physiochemical properties, including varying toxicities.
Why is speciation important?
Speciation analysis has become important for food, environmental, and pharmaceutical industries, where simply measuring the total amount of an element is insufficient. Identifying the different species and concentrations of those species provides a more informed understanding of the environmental or health related impact associated with a particular sample. The clinical or environmental approach would be very different depending on the concentration of the toxic species. For example, an exposure to toxic inorganic arsenic (arsenite and arsenate) is a much bigger concern than an exposure to the mildly toxic organic arsenic species. In contrast, organic mercury species are more toxic than inorganic mercury species, so an exposure to methylmercury or ethylmercury poses a more severe health risk.
How is speciation analysis performed?
IC-ICP-MS provides a complete inorganic elemental analysis solution for speciation analysis.
  • Metal-free ion chromatography (IC) separates the individual ionic species without contributing trace metal contamination
  • Ion Chromatography-Inductively Coupled Plasma Mass Spectrometry (IC-ICP-MS) performs trace elemental detection and quantification
Metal-free IC (with high resolution ion exchange columns and simple online connectivity) together with high sensitivity ICP-MS and integrated software are a powerful combination for fast and efficient metal speciation.
Speciation Analysis with ICP/MS
Frank Hoefler, Ph.D., talks about the synergy with ion chromatography (IC) and inductively coupled plasma mass spectrometry (ICP/MS). The iCap Q ICP/MS brings speciation analysis to a new level of sensitivity and simplicity.
Video Player is loading.
Current Time 0:00
Duration 2:40
Loaded: 6.16%
Stream Type LIVE
Remaining Time 2:40
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
      1. Now Playing
        Up NextSpeciation Analysis with ICP/MS
        Frank Hoefler, Ph.D., talks about the synergy with ion chromatography (IC) and inductively coupled plasma mass spectrometry (ICP/MS). The iCap Q ICP/MS brings speciation analysis to a new level of sensitivity and simplicity.
      2. Now Playing
        Up NextIC-ICP-MS System Provides PPT Elemental Analysis
        Discussion of the IC-ICP-MS method for detecting trace elements such as organic and inorganic arsenic in apple juice and the advantages of this method.
    What example analytes require speciation analysis?
    • Complete inorganic analysis
    • Arsenic
    • Chromium
    • Iodide
    • Mercury
    Speciation Analysis of As in Organic Brown Rice Syrup Using the Thermo Scientific™ iCAP™ Q ICP-MS
    Discussion on how to analyze for arsenic in organic brown rice syrup using the Thermo Scientific™ iCAP™ Q ICP-MS.
    Video Player is loading.
    Current Time 0:00
    Duration 3:50
    Loaded: 4.27%
    Stream Type LIVE
    Remaining Time 3:50
    1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
      • en (Main), selected
        1. Now Playing
          Up NextSpeciation Analysis of As in Organic Brown Rice Syrup Using the Thermo Scientific™ iCAP™ Q ICP-MS
          Discussion on how to analyze for arsenic in organic brown rice syrup using the Thermo Scientific™ iCAP™ Q ICP-MS.
        2. Now Playing
          Up NextChromium Speciation Analysis in Drinking Water by IC-ICP-MS
          Discussion of using IC-ICP-MS as a sensitive and high-throughput method analyzing chromium 3 and 6 in drinking water
        3. Now Playing
          Up NextAnalysis of Arsenic Species in Apple Juice by IC-ICP-MS
          Discussion of using IC-ICP-MS to analyze for 6 arsenic species in apple juice.
      What systems are available for speciation analysis?
      Any of the Thermo Scientific Dionex IC systems can be used in conjunction with the iCAP RQplus ICP-MS system.
      See all configurations
      Resources
      Style Sheet for Global Design System
      Style Sheet for Komodo Tabs
      CMD SchemaApp code