SOLA SPE Plates and Cartridges
Prevent sample failures in bioanalytical workflows
Use Thermo Scientific SOLA Solid Phase Extraction (SPE) cartridges and plates to help prevent costly reanalysis of bioanalytical samples by preventing blocking, voiding, and channeling during the SPE sample preparation process. Award-winning fritless polymeric technology eliminates the issues encountered in conventional SPE so that your lab can:
  • Remove variability
  • Reduce failure rates
  • Optimize high throughput workflows
  • Prevent sample failures
  • Prevent blocked wells or cartridges
  • Reduce reanalysis
Contact us
Dramatically better SPE in every way
SOLA and SOLAμ SPE is available in a range of phases, formats, and sizes to suit any bioanalysis application. Choose from:
5 phases to choose from
  • HRP: Reversed phase for hydrophobic retention
  • SCX: Mixed mode for retention of weak bases
  • SAX: Mixed mode for retention of weak acids)
  • WCX: Mixed mode for retention of strong bases)
  • WAX: Mixed mode for retention of strong acids)
SOLA SPE bed weights
  • SOLA 10 mg is the flexible go-to size for most applications
  • SOLA 30 mg is ideal when analyte breakthrough or loading capacity is an issue, such as samples with very low target analyte concentrations
96-Well plate & cartridge format
  • Cartridges are ideal for method development
  • Well plates are the optimal choice for high-throughput assays
SOLAμ SPE bed weights
  • For low-sample-volume applications and cost-conscious chromatographers
  • 2 mg/1 mL SOLAµ removable 96-well plate works perfectly to reduce solvent consumption and remove the reconstitution-concentration step, saving on precious time and money in high throughput bioanalytical labs.
SOLA & SOLAμ SPE products
Unsure of your sample clean-up method? Try Solid Phase Extraction – Product and Method Selection Tool
 SOLAµ SPE Well Plates
SOLAµ SPE Well Plates
Low-sample, low-solvent volumes in high throughput workflows
SOLA SPE Well Plates
SOLA SPE Well Plates
Flexibility in sample and solvent volumes in high throughput workflows
SOLA SPE Cartridges
SOLA SPE Cartridges
For method development and flexibility in sample and solvent volumes
HRP: Reversed phase for hydrophobic retentionSOLAµ HRP 96-Well PlatesSOLA HRP 96-Well PlatesSOLA HRP Cartridges
SCX: Mixed mode for retention of weak basesSOLAµ SCX 96-Well PlatesSOLA SCX 96-Well PlatesSOLA SCX Cartridges
SAX: Mixed mode for retention of weak acidsSOLAµ SAX 96-Well PlatesSOLA SAX 96-Well PlatesSOLA SAX Cartridges
WCX: Mixed mode for retention of strong basesSOLAµ WCX 96-Well PlatesSOLA WCX 96-Well PlatesSOLA WCX Cartridges
WAX: Mixed mode for retention of strong basesSOLAµ WAX 96-Well PlatesSOLA WAX 96-Well PlatesSOLA WAX Cartridges
Watch SOLA SPE quick protocols
Quick protocols: SPE Phase Selection on Thermo Scientific SOLA SPE Cartridges and Plates
Learn how to select the best material to optimize your solid phase extraction (SPE) protocols with easy-to-use SOLA SPE cartridges and plates.
Video Player is loading.
Current Time 0:00
Duration 5:38
Loaded: 0%
Stream Type LIVE
Remaining Time 5:38
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
      1. Now Playing
        Up NextQuick protocols: SPE Phase Selection on Thermo Scientific SOLA SPE Cartridges and Plates
        Learn how to select the best material to optimize your solid phase extraction (SPE) protocols with easy-to-use SOLA SPE cartridges and plates.
      2. Now Playing
        Up NextQuick protocols: Biofluid sample pre-treatment for solid phase extraction (SPE)
        Get an overview of several sample pre-treatment protocols of biofluids for robust assays and better SPE results.
      3. Now Playing
        Up NextQuick protocols: Assembly of Thermo Scientific SPE Manifolds
        Learn how to properly assemble the vaccum manifold for several Thermo Scientific SPE plate and cartridge formats
    View our SOLA Solid-Phase Extraction (SPE) application note compendium
    Find out how SOLA SPE technology can revolutionize your bioanalytical sample preparation.
    Download now
    Download our SOLA SPE method development assets
    Try our award-winning fritless technology to eliminate conventional SPE issues like voiding, blocking and channeling.
    Download now
    2022/2023 Chromatography Sample Preparation Catalog
    Access the latest interactive version of our Chromatography Consumables & Columns Catalog. Our enhanced online version features easier access to product information, key application notes and brand-new selector tools.
    Get the Sample Preparation catalog
    Discover live and on-demand webinars that enable you to improve productivity, maintain compliance, and learn about the constantly evolving scientific landscape.
    Watch now
    Resources
    Product literature
    Application notes
    CMD SchemaApp code