Volatile Organic Compounds (VOC) Analysis
What is a VOC?
In an environmental analysis context, the VOC designation, or volatile organic contaminants, generally refers to the analysis of compounds in environmental samples with the following chemical properties:+
  • Low boiling points (below 200°C)
  • Low vapor pressures
  • Low-to-medium water solubility
  • Organic compounds
  • Low molecular weights
  • HLC(H)*10-3–10-5 atm*m3/mol'
*Henry's law constants (H) range defined as volatility from liquid to air
Receive updates  Read the blog
VOCs are human-made contaminants used and produced in the processing of or as paints, adhesives, petroleum products, pharmaceuticals, and refrigerants. This includes emissions from automotive and industrial activity among other sources (USGS).
Compound groups included under this classification have various structural characteristics and chemical properties that include:
  • Halogenated hydrocarbons
  • Aromatics
  • Ketones
  • Nitriles
  • Acrylates
  • Acetates
  • Ethers
  • Sulfides
Many of these compounds contaminate our environment today. Acceptable exposure limits and regulations for the release of VOCs to the environment are provided by the EPA and other regulatory bodies.
Volatile Environmental Sampling
At room temperature the properties of VOC cause these compounds to vaporize, evaporate, or sublimate from the liquid or solid form, allowing them to escape into the atmosphere. Sampling methods and extraction techniques must be designed to limit volatile loss from collections to analysis. Similar to SVOC analysis, the type of sample being analyzed dictates the extraction method required. However, techniques to capture and isolate VOC will differ greatly from semivolatile methods.
Aqueous
  • Drinking water
  • Wastewater
  • Source water
  • Ground water
  • Surface water
Solids & semisolids
  • Soil
  • Sediments
  • Sludge
  • Plant tissue
  • Animal tissue
  • Biosolids
Air
  • Ambient air filters/traps
  • External air filters/traps
  • Chimney exhaust filters
  • Air filters from contaminated sites
  • Dusts
VOC Extraction and Concentration
Extraction and concentration
There are various techniques and strategies used to prepare and transfer VOC to instrumentation for analysis. Analysis of volatiles is dominated by the use of purge-and-trap followed by gas chromatography (GC) or gas chromatography mass spectrometry (GC/MS), although static headspace is also used frequently. Five of the most common methods for volatile extraction are listed below.
  • Static headspace analysis
  • Purge-and-trap concentrator
  • Thermal desorption (to air page)
  • Hexadecane or alternate solvent dilution for direct injection
  • Azeotropic and vacuum distillation (nonpurgeable, water soluble, organics)
Headspace Techniques for VOC analysis
Headspace techniques for VOC analysis
Within a sample vial the unoccupied area above the sample is commonly referred to as the head space. Using static headspace, sealed vials containing sample are gently heated to drive VOC compounds out of the sample matrix into equilibrium with the gas phase. Once stabilized the gas phase within the vial is then collected or directly transferred to the instrument for analysis. This technique is generally suited for compounds with relatively high Henry's law constants including fuel range organics. However, this method may be unsuitable for MTBE or ethylene dibromide (EDB).
Purge and trap (P&T) concentrator for VOC analysis
Perhaps the most commonly used and most sensitive technique for volatile analysis is purge and trap concentration, also known as dynamic headspace sampling.
  • Sample is deposited into the sparging vessel where method analytes are purged from the water using a helium or other inert gas stream to sparge through the sample. As with static headspace heat is sometimes applied to drive VOCs out of liquid phase and into the gas phase.
  • Once liberated from the sample VOC’s in the gas phase are transferred to an adsorbent trap where they will transiently bind.
  • The trap is then heated and backflushed with carrier gas to drive the analytes into a gas chromatographic.
Note: After purging, the trap may be dry purged for a short period to remove water.
Webinar
Gas chromatography for VOC analysis
As stated previously the most common technique used to detect, identify and quantitate VOC is gas chromatography with flame ionization (FID), electron capture (ECD) or mass spectrometry (GC-MS) detection. Selection of a detection method requires consideration of various analytical parameters and regulatory requirements. Common stand-alone GC detectors used in VOC analysis with their affinities are listed in the table below.
GC Detectors for TRACE 1600 series GC systems
Detector typeDetection affinity
iConnect Flame Ionization Detector (FID)Sensitive detection of organic compounds over a wide linear dynamic range
iConnect Electron Capture Detector (ECD)Selectivity and sensitivity for trace-level analysis of compounds with electronegative functional groups (e.g. Chlorinated pesticides)
iConnect Nitrogen Phosphorous Detector (NPD)Selectivity and sensitivity to the determination of organic compounds containing nitrogen or phosphorus
iConnect Thermal Conductivity Detector (TCD)Universal detector responds sensitively to both organic and inorganic compounds
iConnect Flame Photometric Detector (FPD)Sensitivity and response stability for determination of sulfur, phosphorous, or tin-containing compounds
Mass Spectrometry for VOC Analysis
Mass spectrometry for VOC analysis
Mass spectrometry has generally replaced GC stand-alone for detection of vocs due to a higher degree of confidence in compound identification. However, routine monitoring of many volatiles is still accomplished by Gas Chromatography detection today. Using GC- MS methods analytes are identified by comparing the acquired mass spectra and retention times to reference spectra and retention times for calibration standards acquired under identical GC-MS conditions.
EPA Regulatory Methods for VOC Analysis
Regulatory methods for VOC analysis
Regulatory agencies publish guidance for monitoring volatile contaminants in the environment. Regulations often group VOC based on several criteria including detection methods, intended compound purpose use, compound structural attributes classifications and other justifications.
Although the United States Environmental Protection Agency (EPA) publishes specific methods for analyzing VOCs not all regulatory agencies provide method guidance. This European Water Framework Directive (EUWFD) provides VOC monitoring limits without a designated methodology. The analytical method chosen must meet published detection limits and criteria specified by each governing body. Frequently used VOC methods are listed below.
EPA GC-MS methods for VOC analysis
  • US EPA 524.2 - Measurement of purgeable organic compounds in water by capillary column GC-MS
  • US EPA 624 - Methods for organic chemical analysis of municipal and industrial waters. 624 – Purgeables
  • US EPA 8260 - Volatile organic compounds by Gas Chromatography /Mass spec (GC-MS)
UX DATA-TRKR Retired - Admins, feel free to remove this reference component.