Drinking Water Analysis
Analytical solutions to ensure safe drinking water
Drinking water requires testing for over 90 possible contaminants according to U.S. EPA regulations in the Safe Drinking Water Act (SDWA). Using approved analytical methods, scientists in municipal water facilities and contract labs routinely monitor for pollutants such as inorganic anions and cations, metals, radionuclides, and organic contaminants using a range of Thermo Scientific solutions including IC, ICP-OES, ICP-MS, GC, GC-MS, LC, LC-MS, and colorimetry. See below to determine which technique to use. 
Contact us   Download brochure
Quick links
Drinking water analysis categories
Our pH, ISE, conductivity, and dissolved oxygen measurement products are well known for excellence in water and liquid analysis. Thermo Scientific™ Orion™ products include a full range of complementary liquid sensing and measurement.
These portable colorimeters can go where you go for your water analysis needs. Choose from 134 preprogrammed methods to measure the concentrations of sample components.
Take your portable turbidity meter to the source for onsite measurement. Our turbidity meters feature white and infrared light sources, helping you meet regulatory requirements.
Use IC for analysis of inorganic anions in drinking water by EPA Method 300.0 or 300.1, and for anion analysis measuring nitrate, nitrite, fluoride, and disinfection byproducts (bromate, chlorite). 
Use LC-MS/MS for hormones by EPA 539, and for microcystins by EPA Methods 544 and 545. Use the power of Orbitrap-based LC-MS to identify and quantify unknown emerging contaminants.
Our GC and GC-MS systems are tailored to meet regulatory compliance for analysis of organic contaminants in drinking water, including volatiles (VOCs) and semivolatiles (SVOCs).
EPA Method 200.8 uses ICP-MS to analyze certain heavy metal contaminants in drinking water. For regulatory compliance, different metals require the use of different methods. 
U.S. EPA Method 200.7 employs ICP-OES to analyze heavy metal contaminants in drinking water. Different metals require different methods to meet regulatory compliance. 
Our discrete analyzers automate photometric and electrochemical analysis providing fast, reproducible results that allow laboratories to measure multiple analytes simultaneously, while reducing total analysis and operator time.
Rely on these solutions to deliver value and help to you achieve your drinking water safety goals. 
Choose from a complete line of ultraviolet-visible (UV-Vis) spectrophotometers. With award-winning designs and user-friendly software, they help you quantify, assess purity, and more.
Drinking water testing techniques
 Turbidity MeterICGC, GC-MSLC, LC-MSICP-OES, ICP-MS
Water turbidity
Odor and taste
Inorganic anions
Trace metals
Disinfection byproducts
Organic contaminants
Emerging contaminants
Featured videos
Santa Clara Valley Water District Customer Testimonial
Customer Testimonial: Santa Clara Valley Water District
Video Player is loading.
Current Time 0:00
Duration 3:46
Loaded: 0%
Stream Type LIVE
Remaining Time 3:46
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
      1. Now Playing
        Up NextSanta Clara Valley Water District Customer Testimonial
        Customer Testimonial: Santa Clara Valley Water District
      2. Now Playing
        Up NextChromium Speciation Analysis in Drinking Water by IC-ICP-MS
        Discussion of using IC-ICP-MS as a sensitive and high-throughput method analyzing chromium 3 and 6 in drinking water
      3. Now Playing
        Up NextA Better Way to Test Drinking Water
        Eurofins Eaton uses high-resolution accurate-mass mass spectrometry to go beyond conventional techniques for analyzing drinking water, adding the ability to rapidly detect and identify unknown compounds.
    Thermo Fisher Scientific is committed to enhancing the quality of our global water resources. As innovation leaders in ion and liquid chromatography, our analytical instruments are used by governments and industry to provide solutions for environmental water testing for a wide range of regulated and emerging inorganic elements and organic compounds.
    Download compendium
    Access this Toolkit to learn from application and industry experts about workflows and innovations that you can implement to advance your environmental water testing. Explore eBooks, application notes, white papers, case studies, webinars and more to optimize your sample processing and analysis.
    Access Toolkit ›
    High-throughput water and nutrient analysis
    Combining rugged hardware, specifically designed software, and simple operation, Thermo Scientific Gallery and Gallery Plus Aqua Master discrete analyzers are built for high-throughput water and nutrient analysis that meets local regulations.
    Download brochure
    Application notes
    Resources
    Brochures
    Application notes