Search Thermo Fisher Scientific
We offer a wide range of solutions to help you ensure that chemically treated drinking water is free of harmful contaminants by enabling analysis of water samples for disinfection byproducts (chlorate, chlorite, bromate, and haloacetic acids), inorganic cations (alkaline earth metals and ammonium), and inorganic anions (fluoride, chloride, bromide, nitrite, phosphate sulfate, perchlorate, hexavalent chromium). Partner with our team of experts to quickly and confidently meet evolving environmental regulatory requirements and overcome your investigative challenges.
We offer a variety of workflows for performing cost-effective robust and reproducible detection, identification, and analysis of inorganic anions, cations and disinfection byproducts for multiple analytes and types of water.
Identify and quantify low concentrations of nine haloacetic acids, bromate and dalapon in drinking water using a uniquely selective ion exchange column. This IC-MS/MS method, requiring no sample pretreatment, can determine all analytes in water samples in 35 minutes, 39% faster than the original U.S. EPA Method 557. Excellent method precision and accuracy are obtained for the standard solution (DI water), laboratory synthetic sample matrix (LSSM), and municipal drinking water. Full method validation including calibration curves, lowest concentration minimum reporting levels (LCMRL), and method detection limits (DL) for all nine HAAs, bromate, and dalapon are presented per U.S. EPA Method 557 guidelines.
"The original gas chromatography electron capture detector (GC-ECD) method we used for HAA determination was labor-intensive and fraught with recovery issues. Since adopting the new, easy-to-use IC-MS/MS system, we have improved confidence in our results, identifying and quantifying low concentrations of HAAs in drinking water samples. The direct detection eliminates tedious sample preparation steps such as sample acidification, extraction, and derivatization, reducing the overall analysis time."
Gerhard Paluca, Senior Sanitary Chemist, Erie County Public Health Laboratory
This application proof note demonstrates the determination of inorganic anions in drinking water according to U.S. EPA Method 300.1. Seven common inorganic anions (fluoride, chloride, bromide, nitrite, nitrate, phosphate, and sulfate) are separated within 5 minutes using a carbonate/bicarbonate eluent and the Dionex IonPac AS22-Fast-4μm column.
Although the common alkali and alkaline earth cations, and ammonium are not considered primary drinking water contaminants in the U.S., they are routinely monitored in this country and are regulated in Europe and Japan. Ammonium is measured in the U.S. for wastewater discharge compliance according to the criteria, published by the U.S. EPA in 2013, for the protection of aquatic life from the toxic effects of ammonia in freshwater.
Learn more about a simple, low-cost ion chromatography workflow, including an integrated ion chromatography system, autosampler and suppressed conductivity detection, for routine determination of inorganic cations and ammonium in drinking water and wastewater samples.
Learn about the solutions we offer for labs performing environmental water testing for a wide range of regulated and emerging inorganic elements and organic compounds.
Search online by compound, technology, or market specialty for a variety of quick start analytical methods featuring Thermo Scientific products and workflows.
Learn how triple quadrupole and high resolution accurate mass spectrometry can improve your ability to deliver faster, accurate and reliable test results.
We offer a variety of solutions to suit your cost-per-sample threshold, throughput requirements, limits of detection, regulatory compliance needs and analytical challenges with inorganic anions, inorganic cations and disinfection byproduct testing.
Multiple anion or cation contaminants can be analyzed using ion chromatography in a single run with unprecedented separation and sensitivity. Modern chromatography techniques bring in cutting-edge technologies such as Reagent-Free Ion Chromatography (RFIC) and High Pressure Ion Chromatography (HPIC) systems that allow you to achieve high-quality, accurate results. Our comprehensive product portfolio allows environmental scientists to choose the instruments and columns that fit their analysis and cost requirements.
Both triple quadrupole (QqQ) and high resolution accurate mass (HRAM) when it comes to quantitation of analytes in complex matrices. Both QqQ and HRAM technologies offer unique advantages, enabling supreme confidence in the quantitation of a variety of molecule types.
Needs | HRAM | QqQ |
---|---|---|
Untargeted screening | ![]() | ![]() |
Targeted screening | ![]() | ![]() |
Untargeted and targeted quantitation | ![]() | ![]() |
Targeted quantitation only | ![]() | ![]() |
Retrospective analysis | ![]() | ![]() |
![]() | ![]() | ![]() | ![]() | ![]() |
In this paper we outline the latest developments in Triple Quadrupole (QqQ) and Orbitrap-based high-resolution spectrometry, highlighting the suitability of each for varying application demands and challenges.
Discover how the power of Orbitrap mass analyzer technology delivers exceptional peak discrimination at low m/z, fine isotopic distribution, quantitation and unknown identification capabilities.
Make data analysis easier and faster by streamlining your small molecule qualitative and quantitative methods for environmental contaminants. Now that you have acquired comprehensive, high-quality MS and MS2 data, you need to identify and characterize unknowns and transfer discovery knowledge through to everyday testing.
Prof. Izumi Yoshihiro talks about performing metabolomics analyses using IC-MS in his laboratory at Kyushu University in Japan.
Learn how to couple an ion chromatography to a mass spectrometric detector, including the importance of suppressor technology.
Learn why electrolytic supression is critical to coupling ion chromatography with mass spectrometry.
Frank Hoefler - Pittcon 2015 Seminar
Hear how IC-MS is used at Ecocontrol Sud to analyze phytopharmaceuticals.
Hear how Italy's Veritas uses IC-MS to detect highly polar pesticides in water.
Learn how your laboratory can partner with Thermo Fisher Scientific to help you overcome barriers to your lab's success.
The world needs science to move faster. Now more than ever. Which is why when more scientists have access to the latest scientific instrumentation, we all benefit. That is the sole purpose of our Lab-Forward Trade-Up Program. For a limited time, trade in your chromatography and mass spectrometry instruments and save!