Portable design with uncompromised performance
Depend on Thermo Scientific Orion AQUAfast colorimeters for simple, accurate, and portable instruments for your water analysis. Offering a wide range of methods, optimum wavelengths, user-friendly displays, and easy calibration, AQUAfast colorimetry meters are fieldwork essentials.
Speak to specialist
Colorimetry instruments
Go where you need to go for your water analysis needs with any AQUAfast Portable Colorimeter. Choose between four models and 134 preprogrammed methods to measure the concentrations of sample components.
AQUAfast AQ3700 Colorimeter
Advanced reliability when you need it most
  • 90+ pre-programmed powder, tablet, and liquid reagent-based methods
  • 6 LED wavelengths, auto-selected via method
  • Stores 10 user-created methods
  • 1,000 data-point memory
  • Rugged IP-67 (waterproof) construction
  • Meter and reagents are compliant with many global regulated testing methods for water samples, including U.S. EPA methods.
  • Compliant with US EPA and other global testing methods for many drinking water and wastewater parameters.
  • Accessory adapters, sealing ring and meter verification check standards kit available
View AQUAfast AQ3700 Colorimeter Reagents ›
AQUAfast AQ4000 Colorimeter
Sophisticated capability, basic affordability
  • Auto-test barcoded ampoules enable hands-free program selection and test initiation
  • 60+ pre-programmed powder, tablet, and liquid reagent-based methods
  • 4 LED wavelengths, auto-selected via method
  • Stores 10 user-created methods
  • 100 data-point memory
  • Rugged IP-67 (waterproof) construction
  • Meter and reagents are compliant with many global regulated testing methods for water samples, including U.S. EPA methods.
  • Compliant with US EPA and other global testing methods for COD in wastewater
  • Accessory vials, adapters, and zero kit available
View AQUAfast AQ4000 Colorimeter Reagents ›
AQUAfast AQ3170 Colorimeter
Free and total chlorine detection made easy
  • Meter and chlorine reagents are compliant with many global regulated testing methods, including US EPA testing methods for free and total chlorine measurements of drinking water and wastewater samples
  • LED light source at 503 nm
  • Two testing ranges of 0.02 to 2.00 mg/L and 0.1 to 8.0 mg/L (with use of accessory 10 mm short path-length vials)
  • Up to 5,000 measurements in continuous mode
  • 16-data point memory
  • CE certified; rugged IP-67 (waterproof) construction
  • Accessory vials and chlorine standard kits available
View AQUAfast AQ3170 Colorimeter Reagents ›
AQUAfast AQ3140 COD Colorimeter
Exclusive focus on chemical oxygen demand detection
  • 430 nm and 610 nm LED wavelengths
  • Three COD testing ranges: 0 to 150 mg/L; 0 to 1500 mg/L; 0 to 15,000 mg/L
  • Benchtop or hand-held use
  • Rugged IP-67 (waterproof) construction
  • Meter and digestion tube reagents are compatible with most global regulated COD testing methods for wastewater samples, including U.S. EPA methods.
  • Compliant with US EPA and other global testing methods for COD in wastewater
  • Available Thermoreactor and COD accessories
View AQUAfast AQ3140 Colorimeter Reagents ›
Browse all reagents and accessories available for your colorimeter
AQUAfast Reagents for the AQUAfast 3700 Colorimeter
Chemical species
Method Description
Method
Form Factor
Range
Shop
Alkalinity
-M, Acid/Indicator Method, 100 tests
AC2002
Tablet
5 – 200 mg/L
AQUAfast alkalinity-m reagent tablets
-P, Acid/Indicator Method, 100 tests
AC3002P
Tablet
5 – 300 mg/L
AQUAfast alkalinity-p reagent tablets
Aluminum
Eriochrome Cyanine R Method, 50 tests
AC2027
Tablet
0.01 – 0.3 mg/L
AQUAfast aluminum reagent tablets*
Eriochrome Cyanine R Method, 100 tests
AC4P27
Powder
0.01 – 0.25 mg/L
AQUAfast aluminum reagent powder packs*
Ammonia as Nitrogen
High Range, Salicylate Method, 50 tubes
ACR011
Reaction Tube
1 – 50 mg/L
AQUAfast ammonia HR Auto-ID ampoules*
Indophenol Phenate Method, 50 tests
AC2012
Tablet
0.02 – 1 mg/L
AQUAfast ammonia reagent tablets*
Low Range, Salicylate Method, 50 tubes
ACR012
Reaction Tube
0.02 – 2.5 mg/L
AQUAfast ammonia LR Auto-ID ampoules*
Salicylate Method, 100 tests
AC4P12
Powder
0.01 – 0.8 mg/L
AQUAfast ammonia reagent powder packs*
Bromine
DPD Method, 100 tests
AC2035
Tablet
0.05 – 13 mg/L
AQUAfast bromine reagent tablets
Chloride
Silver Nitrate/Turbidity Method, 50 tests
AC2017
Tablet
0.5 – 25 mg/L
AQUAfast chloride reagent tablets
Chlorine Dioxide
DPD Method, 100 tests
AC2099
Tablet
0.02 – 11 mg/L
AQUAfast chlorine dioxide reagent tablets*
Chlorine
Free & Total, DPD Method, 100 tests
AC2070
Tablet
0.01 – 6 mg/L
AQUAfast chlorine (free & total) reagent tablets*
Free, DPD bulk powder dispenser with one vial x 250 tests
AQ250F
Powder
0.02 – 2 mg/L Cl2
AQUAfast free chlorine DPD bulk powder dispenser*
Free, DPD Method, 100 tests
AC2071
Tablet
0.01 – 6 mg/L
AQUAfast chlorine (free) reagent tablets*
Free, DPD Method, 100 tests
AC4P71
Powder
0.02 – 2 mg/L
AQUAfast chlorine (free) reagent powder packs*
Free, DPD powder refill for AQ250F dispenser, 1 vial x 250 tests
AC71P1
Powder
0.02 – 2 mg/L Cl2
AQUAfast free chlorine DPD powder refill for AQ250F dispenser*
Free, DPD powder refill for AQ250F dispenser, 2 vials x 250 tests each
AC71P2
Powder
0.02 – 2 mg/L Cl2
AQUAfast free chlorine DPD powder refill for AQ250F dispenser*
Total, DPD Method, 100 tests
AC2072
Tablet
0.01 – 6mg /L
AQUAfast chlorine (total) reagent tablets*
Total, DPD Method, 100 tests
AC4P72
Powder
0.02 – 2 mg/L
AQUAfast chlorine (total) reagent powder packs*
Total, DPD powder dispenser with one vial x 250 tests
AQ250T
Powder
0.02 – 2 mg/L Cl2
AQUAfast total chlorine DPD powder dispenser with one vial x 250 count*
Total, DPD powder refill for AQ250T dispenser, 1 vial x 250 tests
AC72P1
Powder
0.02 – 2 mg/L Cl2
AQUAfast total chlorine DPD powder refill for AQ250T dispenser*
Total, DPD powder refill for AQ250T dispenser, 2 vials x 250 tests each
AC72P2
Powder
0.02 – 2 mg/L Cl2
AQUAfast free chlorine DPD powder refill for AQ250F dispenser*
Total, High Range, KI/Acid Method, 100 tests
AC3072
Tablet
5 – 200 mg/L
AQUAfast chlorine (total) HR & sodium chlorate reagent tablets
COD
Low Range, Dichromate Closed Reflux Method, 25 Tests
CODL00
Digestion Tube
0 - 150mg/L
AQUAfast COD 0-150 ppm digestion tubes*
Low Range, Dichromate Closed Reflux Method, 150 Tests
CODL150
Digestion Tube
0 to 150mg/L O2
AQUAfast COD 0-150 ppm digestion tubes*
Mid Range, Dichromate Closed Reflux Method, 25 Tests
CODH00
Digestion Tube
0 - 1,500mg/L
AQUAfast COD 0-1500 ppm digestion tubes*
Mid Range,Dichromate Closed Reflux Method, 150 Tests
CODM150
Digestion Tube
0 to 1,500mg/L O2
AQUAfast COD 0-1500 ppm digestion tubes*
High Range, Dichromate Closed Reflux Method, 25 Tests
CODHP0
Digestion Tube
0 - 15,000mg/L
AQUAfast COD 0-15000 ppm digestion tubes*
High Range, Dichromate Closed Reflux Method, 150 Tests
CODH150
Digestion Tube
0 to 15,000mg/L O2
AQUAfast Digestion Tubes, Range: 0 to 15000 mg/L O2, COD High Range*
Copper
Free & Total, Biquinoline, 50 tests
AC2029
Method Tablet L
0.05 - 5mg/
AQUAfast copper (free & total) reagent tablets
Free, Bicinchoninate, 100 tests
AC4P29
Method Powder
0.05 - 5mg/L
AQUAfast copper (free) reagent powder packs*
Cyanuric Acid
Melamine Method, 100 tests
AC2098
Tablet
0 – 160 mg/L
AQUAfast cyanuric acid reagent tablets
DEHA
PPST Method, 100 tests
AC4P88
Digestion Tubes and Liquid
0.02 – 0.5 mg/L
AQUAfast DEHA reagent powder pack & liquid set
Fluoride
SPADNS Kit Method, 50 tests
AC2009
Liquid
0.05 – 2 mg/L
AQUAfast fluoride SPADNS liquid reagent*
Hardness
Calcium, Low Range, Murexide Method, 100 tests
AC3032C2
Tablet
0 – 500 mg/L
AQUAfast hardness (calcium) LR reagent tablets, Murexide Method
Calcium, High Range, Murexide Method, 100 tests
AC3032C
Tablet
50 – 900 mg/L
AQUAfast hardness (calcium) HR reagent tablets, Murexide Method
Total, Metallphthlein Method, 100 tests
AC3032T
Tablet
2 – 50 mg/L
AQUAfast hardness (total) reagent tablets, Metallphthalein Method
Hydrazine
4-(Dimethyl-amino)-Benzaldehyde Method, 30 tests
AC2030
Powder
0.05 - 0.5 mg/L
AQUAfast hydrazine reagent powder kit
Hydrogen Peroxide
DPD/Catalyst Method, 100 tests
AC3069
Tablet
0.03 – 3 mg/L
AQUAfast hydrogen peroxide reagent tablets
Iron
(II & III), PPST Method, 100 tests
AC2078
Tablet
0.02 – 1 mg/L
AQUAfast iron (II & III) reagent tablets
Ferro, 1,10-Phenanthroline Method, 100 packs
AC4P78
Powder
0.02 – 3 mg/L
AQUAfast iron (ferro) reagent powder packs*
Total, TPTZ Method, 100 tests
AC4P79
Powder
0.02 - 1.8 mg/L
AQUAfast iron (total) reagent powder packs
Manganese
Formaldoxime Method, 50 tests
AC2055
Tablet
0.2 – 4 mg/L
AQUAfast manganese reagent tablets
High Range, Periodate Oxidation Method, 100 tests
AC4P55
Powder
0.1 – 18 mg/L
AQUAfast manganese HR reagent powder packs*
Low Range, PAN Method, 100 tests
AC4P54
Digestion Tubes and Liquid
0.01 – 0.7mg/L
AQUAfast manganese LR reagent powder & liquid set
Molybdate / Molybdenum
Mercaptoacetic Acid Method, 100 tests
AC4P42
Powder
0.5 – 66 mg/L
AQUAfast molybdate reagent powder packs
Nitrate as Nitrogen (N)
Chromotropic Acid Method, 50 tests
ACR007
Reaction Tube
1 – 30 mg/L
AQUAfast nitrate reaction tubes
Zinc Reduction Method, 50 tests
AC2007
Tablet
0.08 – 1 mg/L
AQUAfast nitrate reagent tablets*
Diazotization (Azo) Method, 100 tests
AC4P46
Powder
0.01 – 0.3 mg/L
AQUAfast nitrite reagent powder packs*
N-(1-Naphthyl)-Ethylenediamine Method, 100 tests, 50 tests
AC2046
Tablet
0.01 – 0.5 mg/L
AQUAfast nitrite reagent tablets*
Nitrogen
Total, High Range, Persulfate Digestion Method, 50 tests
ACD007
Digestion Tube
5 – 150 mg/L
AQUAfast total nitrogen HR digestion tubes
Total, Low Range, Persulfate Digestion Method, 50 pack
ACD004
Digestion Tube
0.5 – 25 mg/L
AQUAfast total nitrogen LR digestion tubes
pH
pH, Bromocresol Purple Method, 100 tests
AC3001BP
Tablet
5.2 – 6.8 pH
AQUAfast ph LR reagent tablets, bromocresol purple
pH, Phenol Red Method, 100 tests
AC2001
Tablet
6.5 – 8.4 pH
AQUAfast pH reagent tablets
pH, Phenol Red Method, 30 tests
AC3001
Liquid
6.5 – 8.4 pH
AQUAfast pH liquid dropper reagent
pH, Thymol Blue Method, 100 tests
AC3001TB
Tablet
8.0 – 9.6 pH
AQUAfast pH HR reagent tablets, thymol blue
Phosphorous as P
Acid Hydrolyzable, Acid Digestion/Ascorbic Acid Method, 50 tests
ACD095AH
Digestion Tube
0.02 – 1.6 mg/L
AQUAfast phosphate (acid hydrolysable) digestion tubes
Total, Persulfate Digestion/Ascorbic Acid Method, 50 tests
ACD095
Digestion Tube
0.02 – 1 mg/L
AQUAfast phosphate (total) digestion tubes*
Phosphate
Ortho,Phosphomolybdenum/Ascorbic Acid Method, 100 tests
AC4P95
Powder
0.06 – 2.5 mg/L
AQUAfast phosphate (ortho) reagent powder packs*
Ortho, Phosphomolybdenum/ Ascorbic Acid Method, 50 tubes
ACR095
Reaction Tube
0.06 – 5mg/L
AQUAfast phosphate (ortho) reaction tube*
Ortho, Phosphomolybdic Acid/ Ascorbic Acid Method, 50 tests
AC2095-WA
Tablet
0.05 – 4 mg/L
AQUAfast phosphate (ortho) LR reagent tablets*
Ortho, Ascorbic Acid/Vanadomolybdate Method, 50 tests
AC2096
Tablet
1 – 80 mg/L
AQUAfast phosphate (ortho) HR reagent tablets*
Potassium
Tetraphenylborate-Turbidity Method, 100 tests
AC3019
Tablet
0.7 – 12 mg/L
AQUAfast potassium reagent tablets
Silica
High Range, Silicomolybdate Method, 100 tests
AC4P60
Powder
1 – 90 mg/L
AQUAfast silica (high range) reagent powder packs*
Silicomolybdate, Heteropoly Blue Method, 50 tests
AC2060
Tablet
0.05 – 4 mg/L
AQUAfast silica reagent tablets*
Phosphate Removal for AC2060 Method, 100 tablets
AC2061
Tablet
---
AQUAfast silica phosphate removal reagent
Sodium
Hypochlorite, Potassium Iodide Method, 100 tests
AC3072
Tablet
0.2 – 16% NaOCl
AQUAfast chlorine (total) HR & sodium chlorate reagent tablets
Sulfate
Barium Sulfate/Turbidity Method, 100 tests
AC2082
Tablet
5 – 100 mg/L
AQUAfast sulfate reagent tablets*
Barium Sulfate/Turbidity Method, 100 tests
AC4P82
Powder
5 – 100 mg/L
AQUAfast sulfate reagent powder packs*
Sulfide
DPD/Catalyst Method, 50 tests
AC2016
Tablet
0.04 – 0.5 mg/L
AQUAfast sulfide reagent tablets
Zinc
Zincon Method, 50 tests
AC2065
Tablet
0.02 – 1 mg/L
AQUAfast zinc reagent tablets*
*Compliant with US EPA and other global testing methods for COD in wastewater
Accessories for use with the AQUAfast AQ3700 Colorimeter
Cat. No.Description 
AC2V16
Replacement 16mm vials, 10/pack
Shop
AC2V24
Replacement 24mm vials, 12/pack
Shop
AC3VSK
Verification standard kit, set of 6 
Shop
AC3CBR
Vial cleaning brush, 5/pack
Shop
COD165
Digestion methods thermoreactor, pre-set for 100°, 120°, 150°, 160°, and 165°C 
Shop
CODS01
COD standard, 1,000ppm, 475mL 
Shop
CODS01
COD standard, 1,000ppm, 475mL 
Shop
AQUAfast Reagents for the AQUAfast 4000 Colorimeter
Chemical species
Method Description
Method
Form Factor
Range
Shop
Alkalinity
-M, Acid/Indicator Method, 100 tests
AC2002
Tablet
5 – 200 mg/L
AQUAfast alkalinity-m reagent tablets
Aluminum
Eriochrome Cyanine R Method, 50 tests
AC2027
Tablet
0.01 – 0.3 mg/
AQUAfast aluminum reagent tablets*
Ammonia as Nitrogen
High Range, Nesslerization Method, 30 pack
AC4011
Reaction Tube
1 – 14 mg/L
AQUAfast ammonia HR Auto-ID ampoules*
Indophenol Phenate Method, 50 tests
AC2012
Tablet
0.02 – 1 mg/L
AQUAfast ammonia reagent tablets*
Low Range, Nessler Method, 30 pack
AC4012
Reaction Tube
0.5 – 7 mg/L
AQUAfast ammonia LR Auto-ID ampoules*
Salicylate Method, 100 tests
AC4P12
Powder
0.01 – 0.8 mg/L
AQUAfast ammonia reagent powder packs*
Salicylate Method, 30 pack
AC4010
Ampoule
0.2 – 3 mg/L; 2 – 30 mg/L
AQUAfast ammonia ULR Auto-ID ampoules*
Chloride
Ferricyanide (Ferric Thiocyanate) Method, 30 tests
AC4017
Ampoule
2.5 – 40 mg/L
AQUAfast chloride Auto-ID ampoules*
Silver Nitrate/Turbidity Method, 50 tests
AC2017
Tablet
0.5 – 25 mg/L
AQUAfast chloride reagent tablets
Chlorine Dioxide
DPD Method, 30 tests
AC4099
Ampoule
0.8 – 11 mg/L
AQUAfast chlorine dioxide Auto-ID ampoules*
Chlorine
Free & Total, DPD Method, 100 tests
AC2070
Tablet
0.01 – 6 mg/L Cl2
AQUAfast chlorine (free & total) reagent tablets*
Free & Total, DPD Method, 30 tests
AC4070
Ampoule
0.4 – 5 mg/L Cl2
AQUAfast chlorine (free & total) Auto-ID ampoules*
Free, DPD bulk powder dispenser with one vial x 250 tests
AQ250F
Powder
0.02 – 2 mg/L Cl2
AQUAfast free chlorine DPD bulk powder dispenser*
Free, DPD Method
AC2071
Tablet
0.01 – 6 mg/L Cl2
AQUAfast chlorine (free) reagent tablets*
Free, DPD Method
AC4P71
Powder
0.02 – 2 mg/L Cl2
AQUAfast chlorine (free) reagent powder packs*
Free, DPD powder refill for AQ250F dispenser, 1 vial x 250 tests
AC71P1
Powder
0.02 – 2 mg/L Cl2
AQUAfast free chlorine DPD powder refill for AQ250F dispenser*
Free, DPD powder refill for AQ250F dispenser, 2 vials x 250 tests each
AC71P2
Powder
0.02 – 2 mg/L Cl2
AQUAfast free chlorine DPD powder refill for AQ250F dispenser*
Total, DPD Method, 100 tests
AC2072
Tablet
0.01 – 6mg /L Cl2
AQUAfast chlorine (total) reagent tablets*
Total, DPD Method, 100 tests
AC4P72
Powder
0.02 – 2 mg/L Cl2
AQUAfast chlorine (total) reagent powder packs*
Total, DPD powder dispenser with one vial x 250 tests
AQ250T
Powder
0.02 – 2 mg/L Cl2
AQUAfast total chlorine DPD powder dispenser with one vial x 250 count*
Total, DPD powder refill for AQ250T dispenser, 1 vial x 250 tests
AC72P1
Powder
0.02 – 2 mg/L Cl2
AQUAfast total chlorine DPD powder refill for AQ250T dispenser*
Total, DPD powder refill for AQ250T dispenser, 2 vials x 250 tests each
AC72P2
Powder
0.02 – 2 mg/L Cl2
AQUAfast free chlorine DPD powder refill for AQ250F dispenser*
COD
Low Range, Dichromate closed reflux Method, 25 Tests
CODL00
Digestion Tube
0 – 150mg/L
AQUAfast COD 0-150 ppm digestion tubes*
Low Range, Dichromate closed reflux Method, 150 Tests
CODL150
Digestion Tube
0 – 150mg/L O2
AQUAfast COD 0-150 ppm digestion tubes*
Mid Range, Dichromate closed reflux Method, 25 Tests
CODH00
Digestion Tube
0 – 1,500mg/L
AQUAfast COD 0-1500 ppm digestion tubes*
Mid Range, Dichromate closed reflux Method, 150 Tests
CODM150
Digestion Tube
0 – 1,500mg/L O2
AQUAfast COD 0-1500 ppm digestion tubes*
High Range, Dichromate closed reflux Method, 25 Tests
CODHPO
Digestion Tube
0 – 15,000mg/L O2
AQUAfast COD 0-15000 ppm digestion tubes*
High Range, Dichromate closed reflux Method, 150 Tests
CODH150
Digestion Tube
0 – 15,000mg/L O2
AQUAfast Digestion Tubes, Range: 0 to 15000 mg/L O2, COD High Range*
Copper
Free & Total, Biquinoline, 50 tests
AC2029
Tablet
0.05 – 5mg/L
AQUAfast copper (free & total) reagent tablets
Free, Bicinchoninate, 100 tests
AC4P29
Powder
0.05 – 5mg/L
AQUAfast copper (free) reagent powder packs*
Cyanide
Isonicotinic-Barbituric Acid Method, 30 tests
AC4006
Ampoule
0.04 – 0.4mg/L
AQUAfast cyanide (free) auto-id ampoules*
Fluoride
SPADNS Kit Method, 50 tests
AC2009
Liquid
0.05 – 2 mg/L
AQUAfast fluoride SPADNS liquid reagent*
Hydrazine
4-(Dimethyl-amino)-Benzaldehyde Method, 30 tests
AC2030
Powder
0.05 – 0.5 mg/L
AQUAfast hydrazine reagent powder kit
Iron
(II & III), PPST Method, 100 tests
AC2078
Tablet
0.02 – 1 mg/L
AQUAfast iron (II & III) reagent tablets
Ferro, 1,10-Phenanthroline Method, 100 tests
AC4P78
Powder
0.02 – 3 mg/L
AQUAfast iron (ferro) reagent powder packs*
Total & Soluble, Phenanthroline Method, 30 tests
AC4078
Ampoule
0.2 – 6 mg/L
AQUAfast iron (total & soluble) Auto-ID ampoules*
Total, TPTZ Method, 100 tests
AC4P79
Powder
0.02 – 1.8 mg/L
AQUAfast iron (total) reagent powder packs
Manganese
Formaldoxime Method, 50 tests
AC2055
Tablet
0.2 – 4 mg/L
AQUAfast manganese reagent tablets
High Range, Periodate Oxidation Method, 100 tests
AC4P55
Powder
0.1 – 18 mg/L
AQUAfast manganese HR reagent powder packs*
Periodate Method, 30 tests
AC4055
Ampoule
2 – 30 mg/L
AQUAfast manganese Auto-ID ampoules*
Molybdate / Molybdenum
Mercaptoacetic Acid Method, 100 tests
AC4P42
Powder
0.5 – 66 mg/L
AQUAfast molybdate reagent powder packs
Nitrate as Nitrogen (N)
Cadmium Reduction Method, 30 tests
AC4004
Ampoule
0.2 – 1.5 mg/L
AQUAfast nitrate LR Auto-ID ampoules*
Cadmium Reduction Method, 30 tests
AC4005
Ampoule
2.0 – 7.50 mg/L
AQUAfast nitrate MR Auto-ID ampoules*
Zinc Reduction Method, 50 tests
AC2007
Tablet
0.08 – 1 mg/L
AQUAfast nitrate reagent tablets*
Cadmium Reduction Method, 30 tests
AC4007
Ampoule
5 – 50 mg/L
AQUAfast nitrate HR Auto-ID ampoules*
Diazotization (NED) Method
AC4P46
Powder
0.01 – 0.3 mg/L
AQUAfast nitrite reagent powder packs*
Diazotization (NED) Method
AC4046
Tablet
0.08 – 1 mg/L
AQUAfast nitrite Auto-ID ampoules*
Diazotization (NED) Method
AC2046
Tablet
0.01 – 0.5 mg/L
AQUAfast nitrite reagent tablets*
Ozone
DPD Method, 30 tests
AC4048
Ampoule
0.2 – 5 pH
AQUAfast ozone Auto-ID ampoules
pH
Phenol Red Method, 100 tests
AC2001
Tablet
6.5 – 8.4 pH
AQUAfast pH reagent tablets
Phosphate
Phosphate, Ortho, Phosphomolybdenum/Ascorbic Acid Method, 100 tests 
AC4P95
Powder
0.06 – 2.5 mg/L
AQUAfast phosphate (ortho) reagent powder packs*
Phosphate, Ortho, Phosphomolybdic Acid/Ascorbic Acid Method, 50 tests
AC2095-WA
Tablet
0.05 – 4 mg/L
AQUAfast phosphate (ortho) LR reagent tablets*
Ortho, Stannous Chloride Method, 30 tests
g
Reaction Tube
0.3 – 8 mg/L
AQUAfast phosphate (ortho) Auto-ID ampoules
Phosphate, Ortho, Ascorbic Acid/Vanadomolybdate Method, 50 tests
AC2096
Tablet
1 – 80 mg/L
AQUAfast phosphate (ortho) HR reagent tablets*
Silica
Heteropoly Blue Method, 30 tests
AC4060
Ampoule
0.5 – 10 mg/L
AQUAfast silica Auto-ID ampoules*
Phosphate Removal for AC2060 Method, 100 tests
AC2061
Tablet
----
AQUAfast silica phosphate removal reagent*
High Range, Silicomolybdate Method, 100 tests
AC4P60
Powder
1 – 90 mg/L
AQUAfast silica (high range) reagent powder packs*
Silicomolybdate, Heteropoly Blue Method, 50 tests
AC2060
Tablet
0.05 – 4 mg/L
AQUAfast silica reagent tablets*
Sulfate
Barium Sulfate/Turbidity Method, 100 tests
AC2082
Tablet
5 – 100 mg/L
AQUAfast sulfate reagent tablets*
Barium Sulfate/Turbidity Method, 30 tests
AC4082
Ampoule
8 – 100 mg/L
AQUAfast sulfate Auto-ID ampoules*
Barium Sulfate/Turbidity Method, 100 tests
AC4P82
Powder
5 – 100 mg/L
AQUAfast sulfate reagent powder packs*
Sulfide
Acid Soluble, Methylene Blue Method, 30 tests
AC4016
Ampoule
0.2 – 3 mg/L
AQUAfast sulfide (Acid Soluble) Auto-ID ampoule*
DPD/Catalyst Method, 50 tests
AC2016
Tablet
0.04 – 0.5 mg/L
AQUAfast sulfide reagent tablets
Zinc
Zinc, Zincon Method, 50 tests
AC2065
Tablet
0.02 – 1 mg/L
AQUAfast zinc reagent tablets*
Zinc, Zincon Method, 30 pack
AC4065
Auto-ID Ampoule
0.3 – 3 mg/L
AQUAfast zinc Auto-ID ampoules*
*Compliant with US EPA and other global testing methods for COD in wastewater
Accessories for use with the AQUAfast AQ4000 Colorimeter
Cat. No.
Description
AC2V16
Replacement 16mm vials, 10/pack
Shop
AC2V24
Replacement 24mm vials, 12/pack
Shop
AC3CBR
Vial cleaning brush, 5/pack
Shop
AQ4CBL
RS232 cable for data download and software update
Shop
AQ4ZER
AQUAfast Zero Kit
Shop
COD165
Digestion methods thermoreactor, pre-set for 100°, 120°, 150°, 160°, and 165°C 
Shop
CODS01
COD standard, 1000ppm, 475mL 
Shop
CODS10
COD standard, 10000ppm, 475mL 
Shop
CLSK100
Chlorine primary standard, 1.5mg/L ampoule kit
Shop
AC3ROD
Crush and stir rod for tablets and AC2V24 vials
Shop
AQUAfast Reagents for the AQUAfast 3170 Colorimeter
Chemical species
Method Description
Method
Form Factor
Range
Shop
Chlorine
Free, DPD Method, 100 tests
AC4P71
Powder
0.02 – 2 mg/L Cl2; 0.1 – 8 mg/L Cl2+
AQUAfast chlorine (free) reagent powder packs*
Free, DPD bulk powder dispenser with one vial x 250 tests
AQ250F
Powder
0.02 – 2 mg/L Cl2
AQUAfast free chlorine DPD bulk powder dispenser*
Free, DPD powder refill for AQ250F dispenser, 1 vial x 250 tests
AC71P1
Powder
0.02 – 2 mg/L Cl2
AQUAfast free chlorine DPD powder refill for AQ250F dispenser*
Free, DPD powder refill for AQ250F dispenser, 2 vials x 250 tests each
AC71P2
Powder
0.02 – 2 mg/L Cl2
AQUAfast free chlorine DPD powder refill for AQ250F dispenser*
Total, DPD Method, 100 tests
AC4P72
Powder
0.02 – 2 mg/L Cl2; 0.1 – 8 mg/L Cl2+
AQUAfast chlorine (total) reagent powder packs*
Total, Bluk DPD powder dispenser with one vial x 250 tests
AQ250T
Powder
0.02 – 2 mg/L Cl2
AQUAfast total chlorine DPD powder dispenser with one vial x 250 count*
Total, DPD powder refill for AQ250T dispenser, 1 vial x 250 tests
AC72P1
Powder
0.02 – 2 mg/L Cl2
AQUAfast total chlorine DPD powder refill for AQ250T dispenser*
Total, DPD powder refill for AQ250T dispenser, 2 vials x 250 tests each
AC72P2
Powder
0.02 – 2 mg/L Cl2
AQUAfast free chlorine DPD powder refill for AQ250F dispenser*
† Use AC2V10 short path length plastic vials and two powder packs.
*Compliant with US EPA and other global testing methods for COD in wastewater
Accessories for use with the AQUAfast AQ3170 Colorimeter
Cat. No.
Description
AC2V24
Replacement 24mm vials, 12/pack
Shop
AC2V10
AQUAfast 10mm short path length plastic vials for high-level chlorine, 12/pack
Shop
CLSK100
Chlorine primary standard, 1.5mg/L ampoule kit
Shop
CLSK200
Chlorine secondary standards (0.0, 0.2, and 1mg/L)
Shop
AC3CBR
Vial cleaning brush, 5/pack
Shop
AQUAfast Reagents for the AQUAfast 3140 COD Colorimeter
Chemical species
Method Description
Method
Form Factor
Range
Shop
COD
Low Range, Dichromate Digestion Method, 25 Tests
CODL00
Digestion Tube
0 – 150mg/L
AQUAfast COD 0-150 ppm digestion tubes*
Low Range, Dichromate Digestion Method, 150 Tests
CODL150
Digestion Tube
0 – 150mg/L O2
AQUAfast COD 0-150 ppm digestion tubes*
Mid Range, Dichromate Digestion Method, 25 Tests
CODH00
Digestion Tube
0 – 1,500mg/L
AQUAfast COD 0-1500 ppm digestion tubes*
Mid Range,Dichromate Digestion Method, 150 Tests
CODM150
Digestion Tube
0 – 1,500mg/L O2
AQUAfast COD 0-1500 ppm digestion tubes*
High Range, Dichromate Digestion Method, 25 Tests
CODHP0
Digestion Tube
0 – 15,000mg/L
AQUAfast Digestion Tubes, Range: 0-15000 mg/L O2 COD High Range*
High Range, Dichromate Digestion Method, 150 Tests
CODH150
Digestion Tube
0 – 15,000mg/L O2
AQUAfast Digestion Tubes, Range: 0 to 15000 mg/L O2, COD High Range*
*Compliant with US EPA and other global testing methods for COD in wastewater
Accessories for use with the AQUAfast AQ3140 COD Colorimeter
Cat. No.
Description
COD165
Digestion methods thermoreactor, pre-set for 100°, 120°, 150°, 160°, and 165°C 
Shop
CODS01
COD standard, 1000ppm, 475mL 
Shop
CODS10
COD standard, 10000ppm, 475mL 
Shop
AQUAfast Reagents for the AQUAfast 3700 Colorimeter
Chemical species
Method Description
Method
Form Factor
Range
Shop
Alkalinity
-M, Acid/Indicator Method, 100 tests
AC2002
Tablet
5 – 200 mg/L
AQUAfast alkalinity-m reagent tablets
-P, Acid/Indicator Method, 100 tests
AC3002P
Tablet
5 – 300 mg/L
AQUAfast alkalinity-p reagent tablets
Aluminum
Eriochrome Cyanine R Method, 50 tests
AC2027
Tablet
0.01 – 0.3 mg/L
AQUAfast aluminum reagent tablets*
Eriochrome Cyanine R Method, 100 tests
AC4P27
Powder
0.01 – 0.25 mg/L
AQUAfast aluminum reagent powder packs*
Ammonia as Nitrogen
High Range, Salicylate Method, 50 tubes
ACR011
Reaction Tube
1 – 50 mg/L
AQUAfast ammonia HR Auto-ID ampoules*
Indophenol Phenate Method, 50 tests
AC2012
Tablet
0.02 – 1 mg/L
AQUAfast ammonia reagent tablets*
Low Range, Salicylate Method, 50 tubes
ACR012
Reaction Tube
0.02 – 2.5 mg/L
AQUAfast ammonia LR Auto-ID ampoules*
Salicylate Method, 100 tests
AC4P12
Powder
0.01 – 0.8 mg/L
AQUAfast ammonia reagent powder packs*
Bromine
DPD Method, 100 tests
AC2035
Tablet
0.05 – 13 mg/L
AQUAfast bromine reagent tablets
Chloride
Silver Nitrate/Turbidity Method, 50 tests
AC2017
Tablet
0.5 – 25 mg/L
AQUAfast chloride reagent tablets
Chlorine Dioxide
DPD Method, 100 tests
AC2099
Tablet
0.02 – 11 mg/L
AQUAfast chlorine dioxide reagent tablets*
Chlorine
Free & Total, DPD Method, 100 tests
AC2070
Tablet
0.01 – 6 mg/L
AQUAfast chlorine (free & total) reagent tablets*
Free, DPD bulk powder dispenser with one vial x 250 tests
AQ250F
Powder
0.02 – 2 mg/L Cl2
AQUAfast free chlorine DPD bulk powder dispenser*
Free, DPD Method, 100 tests
AC2071
Tablet
0.01 – 6 mg/L
AQUAfast chlorine (free) reagent tablets*
Free, DPD Method, 100 tests
AC4P71
Powder
0.02 – 2 mg/L
AQUAfast chlorine (free) reagent powder packs*
Free, DPD powder refill for AQ250F dispenser, 1 vial x 250 tests
AC71P1
Powder
0.02 – 2 mg/L Cl2
AQUAfast free chlorine DPD powder refill for AQ250F dispenser*
Free, DPD powder refill for AQ250F dispenser, 2 vials x 250 tests each
AC71P2
Powder
0.02 – 2 mg/L Cl2
AQUAfast free chlorine DPD powder refill for AQ250F dispenser*
Total, DPD Method, 100 tests
AC2072
Tablet
0.01 – 6mg /L
AQUAfast chlorine (total) reagent tablets*
Total, DPD Method, 100 tests
AC4P72
Powder
0.02 – 2 mg/L
AQUAfast chlorine (total) reagent powder packs*
Total, DPD powder dispenser with one vial x 250 tests
AQ250T
Powder
0.02 – 2 mg/L Cl2
AQUAfast total chlorine DPD powder dispenser with one vial x 250 count*
Total, DPD powder refill for AQ250T dispenser, 1 vial x 250 tests
AC72P1
Powder
0.02 – 2 mg/L Cl2
AQUAfast total chlorine DPD powder refill for AQ250T dispenser*
Total, DPD powder refill for AQ250T dispenser, 2 vials x 250 tests each
AC72P2
Powder
0.02 – 2 mg/L Cl2
AQUAfast free chlorine DPD powder refill for AQ250F dispenser*
Total, High Range, KI/Acid Method, 100 tests
AC3072
Tablet
5 – 200 mg/L
AQUAfast chlorine (total) HR & sodium chlorate reagent tablets
COD
Low Range, Dichromate Closed Reflux Method, 25 Tests
CODL00
Digestion Tube
0 - 150mg/L
AQUAfast COD 0-150 ppm digestion tubes*
Low Range, Dichromate Closed Reflux Method, 150 Tests
CODL150
Digestion Tube
0 to 150mg/L O2
AQUAfast COD 0-150 ppm digestion tubes*
Mid Range, Dichromate Closed Reflux Method, 25 Tests
CODH00
Digestion Tube
0 - 1,500mg/L
AQUAfast COD 0-1500 ppm digestion tubes*
Mid Range,Dichromate Closed Reflux Method, 150 Tests
CODM150
Digestion Tube
0 to 1,500mg/L O2
AQUAfast COD 0-1500 ppm digestion tubes*
High Range, Dichromate Closed Reflux Method, 25 Tests
CODHP0
Digestion Tube
0 - 15,000mg/L
AQUAfast COD 0-15000 ppm digestion tubes*
High Range, Dichromate Closed Reflux Method, 150 Tests
CODH150
Digestion Tube
0 to 15,000mg/L O2
AQUAfast Digestion Tubes, Range: 0 to 15000 mg/L O2, COD High Range*
Copper
Free & Total, Biquinoline, 50 tests
AC2029
Method Tablet L
0.05 - 5mg/
AQUAfast copper (free & total) reagent tablets
Free, Bicinchoninate, 100 tests
AC4P29
Method Powder
0.05 - 5mg/L
AQUAfast copper (free) reagent powder packs*
Cyanuric Acid
Melamine Method, 100 tests
AC2098
Tablet
0 – 160 mg/L
AQUAfast cyanuric acid reagent tablets
DEHA
PPST Method, 100 tests
AC4P88
Digestion Tubes and Liquid
0.02 – 0.5 mg/L
AQUAfast DEHA reagent powder pack & liquid set
Fluoride
SPADNS Kit Method, 50 tests
AC2009
Liquid
0.05 – 2 mg/L
AQUAfast fluoride SPADNS liquid reagent*
Hardness
Calcium, Low Range, Murexide Method, 100 tests
AC3032C2
Tablet
0 – 500 mg/L
AQUAfast hardness (calcium) LR reagent tablets, Murexide Method
Calcium, High Range, Murexide Method, 100 tests
AC3032C
Tablet
50 – 900 mg/L
AQUAfast hardness (calcium) HR reagent tablets, Murexide Method
Total, Metallphthlein Method, 100 tests
AC3032T
Tablet
2 – 50 mg/L
AQUAfast hardness (total) reagent tablets, Metallphthalein Method
Hydrazine
4-(Dimethyl-amino)-Benzaldehyde Method, 30 tests
AC2030
Powder
0.05 - 0.5 mg/L
AQUAfast hydrazine reagent powder kit
Hydrogen Peroxide
DPD/Catalyst Method, 100 tests
AC3069
Tablet
0.03 – 3 mg/L
AQUAfast hydrogen peroxide reagent tablets
Iron
(II & III), PPST Method, 100 tests
AC2078
Tablet
0.02 – 1 mg/L
AQUAfast iron (II & III) reagent tablets
Ferro, 1,10-Phenanthroline Method, 100 packs
AC4P78
Powder
0.02 – 3 mg/L
AQUAfast iron (ferro) reagent powder packs*
Total, TPTZ Method, 100 tests
AC4P79
Powder
0.02 - 1.8 mg/L
AQUAfast iron (total) reagent powder packs
Manganese
Formaldoxime Method, 50 tests
AC2055
Tablet
0.2 – 4 mg/L
AQUAfast manganese reagent tablets
High Range, Periodate Oxidation Method, 100 tests
AC4P55
Powder
0.1 – 18 mg/L
AQUAfast manganese HR reagent powder packs*
Low Range, PAN Method, 100 tests
AC4P54
Digestion Tubes and Liquid
0.01 – 0.7mg/L
AQUAfast manganese LR reagent powder & liquid set
Molybdate / Molybdenum
Mercaptoacetic Acid Method, 100 tests
AC4P42
Powder
0.5 – 66 mg/L
AQUAfast molybdate reagent powder packs
Nitrate as Nitrogen (N)
Chromotropic Acid Method, 50 tests
ACR007
Reaction Tube
1 – 30 mg/L
AQUAfast nitrate reaction tubes
Zinc Reduction Method, 50 tests
AC2007
Tablet
0.08 – 1 mg/L
AQUAfast nitrate reagent tablets*
Diazotization (Azo) Method, 100 tests
AC4P46
Powder
0.01 – 0.3 mg/L
AQUAfast nitrite reagent powder packs*
N-(1-Naphthyl)-Ethylenediamine Method, 100 tests, 50 tests
AC2046
Tablet
0.01 – 0.5 mg/L
AQUAfast nitrite reagent tablets*
Nitrogen
Total, High Range, Persulfate Digestion Method, 50 tests
ACD007
Digestion Tube
5 – 150 mg/L
AQUAfast total nitrogen HR digestion tubes
Total, Low Range, Persulfate Digestion Method, 50 pack
ACD004
Digestion Tube
0.5 – 25 mg/L
AQUAfast total nitrogen LR digestion tubes
pH
pH, Bromocresol Purple Method, 100 tests
AC3001BP
Tablet
5.2 – 6.8 pH
AQUAfast ph LR reagent tablets, bromocresol purple
pH, Phenol Red Method, 100 tests
AC2001
Tablet
6.5 – 8.4 pH
AQUAfast pH reagent tablets
pH, Phenol Red Method, 30 tests
AC3001
Liquid
6.5 – 8.4 pH
AQUAfast pH liquid dropper reagent
pH, Thymol Blue Method, 100 tests
AC3001TB
Tablet
8.0 – 9.6 pH
AQUAfast pH HR reagent tablets, thymol blue
Phosphorous as P
Acid Hydrolyzable, Acid Digestion/Ascorbic Acid Method, 50 tests
ACD095AH
Digestion Tube
0.02 – 1.6 mg/L
AQUAfast phosphate (acid hydrolysable) digestion tubes
Total, Persulfate Digestion/Ascorbic Acid Method, 50 tests
ACD095
Digestion Tube
0.02 – 1 mg/L
AQUAfast phosphate (total) digestion tubes*
Phosphate
Ortho,Phosphomolybdenum/Ascorbic Acid Method, 100 tests
AC4P95
Powder
0.06 – 2.5 mg/L
AQUAfast phosphate (ortho) reagent powder packs*
Ortho, Phosphomolybdenum/ Ascorbic Acid Method, 50 tubes
ACR095
Reaction Tube
0.06 – 5mg/L
AQUAfast phosphate (ortho) reaction tube*
Ortho, Phosphomolybdic Acid/ Ascorbic Acid Method, 50 tests
AC2095-WA
Tablet
0.05 – 4 mg/L
AQUAfast phosphate (ortho) LR reagent tablets*
Ortho, Ascorbic Acid/Vanadomolybdate Method, 50 tests
AC2096
Tablet
1 – 80 mg/L
AQUAfast phosphate (ortho) HR reagent tablets*
Potassium
Tetraphenylborate-Turbidity Method, 100 tests
AC3019
Tablet
0.7 – 12 mg/L
AQUAfast potassium reagent tablets
Silica
High Range, Silicomolybdate Method, 100 tests
AC4P60
Powder
1 – 90 mg/L
AQUAfast silica (high range) reagent powder packs*
Silicomolybdate, Heteropoly Blue Method, 50 tests
AC2060
Tablet
0.05 – 4 mg/L
AQUAfast silica reagent tablets*
Phosphate Removal for AC2060 Method, 100 tablets
AC2061
Tablet
---
AQUAfast silica phosphate removal reagent
Sodium
Hypochlorite, Potassium Iodide Method, 100 tests
AC3072
Tablet
0.2 – 16% NaOCl
AQUAfast chlorine (total) HR & sodium chlorate reagent tablets
Sulfate
Barium Sulfate/Turbidity Method, 100 tests
AC2082
Tablet
5 – 100 mg/L
AQUAfast sulfate reagent tablets*
Barium Sulfate/Turbidity Method, 100 tests
AC4P82
Powder
5 – 100 mg/L
AQUAfast sulfate reagent powder packs*
Sulfide
DPD/Catalyst Method, 50 tests
AC2016
Tablet
0.04 – 0.5 mg/L
AQUAfast sulfide reagent tablets
Zinc
Zincon Method, 50 tests
AC2065
Tablet
0.02 – 1 mg/L
AQUAfast zinc reagent tablets*
*Compliant with US EPA and other global testing methods for COD in wastewater
Accessories for use with the AQUAfast AQ3700 Colorimeter
Cat. No.Description 
AC2V16
Replacement 16mm vials, 10/pack
Shop
AC2V24
Replacement 24mm vials, 12/pack
Shop
AC3VSK
Verification standard kit, set of 6 
Shop
AC3CBR
Vial cleaning brush, 5/pack
Shop
COD165
Digestion methods thermoreactor, pre-set for 100°, 120°, 150°, 160°, and 165°C 
Shop
CODS01
COD standard, 1,000ppm, 475mL 
Shop
CODS01
COD standard, 1,000ppm, 475mL 
Shop
AQUAfast Reagents for the AQUAfast 4000 Colorimeter
Chemical species
Method Description
Method
Form Factor
Range
Shop
Alkalinity
-M, Acid/Indicator Method, 100 tests
AC2002
Tablet
5 – 200 mg/L
AQUAfast alkalinity-m reagent tablets
Aluminum
Eriochrome Cyanine R Method, 50 tests
AC2027
Tablet
0.01 – 0.3 mg/
AQUAfast aluminum reagent tablets*
Ammonia as Nitrogen
High Range, Nesslerization Method, 30 pack
AC4011
Reaction Tube
1 – 14 mg/L
AQUAfast ammonia HR Auto-ID ampoules*
Indophenol Phenate Method, 50 tests
AC2012
Tablet
0.02 – 1 mg/L
AQUAfast ammonia reagent tablets*
Low Range, Nessler Method, 30 pack
AC4012
Reaction Tube
0.5 – 7 mg/L
AQUAfast ammonia LR Auto-ID ampoules*
Salicylate Method, 100 tests
AC4P12
Powder
0.01 – 0.8 mg/L
AQUAfast ammonia reagent powder packs*
Salicylate Method, 30 pack
AC4010
Ampoule
0.2 – 3 mg/L; 2 – 30 mg/L
AQUAfast ammonia ULR Auto-ID ampoules*
Chloride
Ferricyanide (Ferric Thiocyanate) Method, 30 tests
AC4017
Ampoule
2.5 – 40 mg/L
AQUAfast chloride Auto-ID ampoules*
Silver Nitrate/Turbidity Method, 50 tests
AC2017
Tablet
0.5 – 25 mg/L
AQUAfast chloride reagent tablets
Chlorine Dioxide
DPD Method, 30 tests
AC4099
Ampoule
0.8 – 11 mg/L
AQUAfast chlorine dioxide Auto-ID ampoules*
Chlorine
Free & Total, DPD Method, 100 tests
AC2070
Tablet
0.01 – 6 mg/L Cl2
AQUAfast chlorine (free & total) reagent tablets*
Free & Total, DPD Method, 30 tests
AC4070
Ampoule
0.4 – 5 mg/L Cl2
AQUAfast chlorine (free & total) Auto-ID ampoules*
Free, DPD bulk powder dispenser with one vial x 250 tests
AQ250F
Powder
0.02 – 2 mg/L Cl2
AQUAfast free chlorine DPD bulk powder dispenser*
Free, DPD Method
AC2071
Tablet
0.01 – 6 mg/L Cl2
AQUAfast chlorine (free) reagent tablets*
Free, DPD Method
AC4P71
Powder
0.02 – 2 mg/L Cl2
AQUAfast chlorine (free) reagent powder packs*
Free, DPD powder refill for AQ250F dispenser, 1 vial x 250 tests
AC71P1
Powder
0.02 – 2 mg/L Cl2
AQUAfast free chlorine DPD powder refill for AQ250F dispenser*
Free, DPD powder refill for AQ250F dispenser, 2 vials x 250 tests each
AC71P2
Powder
0.02 – 2 mg/L Cl2
AQUAfast free chlorine DPD powder refill for AQ250F dispenser*
Total, DPD Method, 100 tests
AC2072
Tablet
0.01 – 6mg /L Cl2
AQUAfast chlorine (total) reagent tablets*
Total, DPD Method, 100 tests
AC4P72
Powder
0.02 – 2 mg/L Cl2
AQUAfast chlorine (total) reagent powder packs*
Total, DPD powder dispenser with one vial x 250 tests
AQ250T
Powder
0.02 – 2 mg/L Cl2
AQUAfast total chlorine DPD powder dispenser with one vial x 250 count*
Total, DPD powder refill for AQ250T dispenser, 1 vial x 250 tests
AC72P1
Powder
0.02 – 2 mg/L Cl2
AQUAfast total chlorine DPD powder refill for AQ250T dispenser*
Total, DPD powder refill for AQ250T dispenser, 2 vials x 250 tests each
AC72P2
Powder
0.02 – 2 mg/L Cl2
AQUAfast free chlorine DPD powder refill for AQ250F dispenser*
COD
Low Range, Dichromate closed reflux Method, 25 Tests
CODL00
Digestion Tube
0 – 150mg/L
AQUAfast COD 0-150 ppm digestion tubes*
Low Range, Dichromate closed reflux Method, 150 Tests
CODL150
Digestion Tube
0 – 150mg/L O2
AQUAfast COD 0-150 ppm digestion tubes*
Mid Range, Dichromate closed reflux Method, 25 Tests
CODH00
Digestion Tube
0 – 1,500mg/L
AQUAfast COD 0-1500 ppm digestion tubes*
Mid Range, Dichromate closed reflux Method, 150 Tests
CODM150
Digestion Tube
0 – 1,500mg/L O2
AQUAfast COD 0-1500 ppm digestion tubes*
High Range, Dichromate closed reflux Method, 25 Tests
CODHPO
Digestion Tube
0 – 15,000mg/L O2
AQUAfast COD 0-15000 ppm digestion tubes*
High Range, Dichromate closed reflux Method, 150 Tests
CODH150
Digestion Tube
0 – 15,000mg/L O2
AQUAfast Digestion Tubes, Range: 0 to 15000 mg/L O2, COD High Range*
Copper
Free & Total, Biquinoline, 50 tests
AC2029
Tablet
0.05 – 5mg/L
AQUAfast copper (free & total) reagent tablets
Free, Bicinchoninate, 100 tests
AC4P29
Powder
0.05 – 5mg/L
AQUAfast copper (free) reagent powder packs*
Cyanide
Isonicotinic-Barbituric Acid Method, 30 tests
AC4006
Ampoule
0.04 – 0.4mg/L
AQUAfast cyanide (free) auto-id ampoules*
Fluoride
SPADNS Kit Method, 50 tests
AC2009
Liquid
0.05 – 2 mg/L
AQUAfast fluoride SPADNS liquid reagent*
Hydrazine
4-(Dimethyl-amino)-Benzaldehyde Method, 30 tests
AC2030
Powder
0.05 – 0.5 mg/L
AQUAfast hydrazine reagent powder kit
Iron
(II & III), PPST Method, 100 tests
AC2078
Tablet
0.02 – 1 mg/L
AQUAfast iron (II & III) reagent tablets
Ferro, 1,10-Phenanthroline Method, 100 tests
AC4P78
Powder
0.02 – 3 mg/L
AQUAfast iron (ferro) reagent powder packs*
Total & Soluble, Phenanthroline Method, 30 tests
AC4078
Ampoule
0.2 – 6 mg/L
AQUAfast iron (total & soluble) Auto-ID ampoules*
Total, TPTZ Method, 100 tests
AC4P79
Powder
0.02 – 1.8 mg/L
AQUAfast iron (total) reagent powder packs
Manganese
Formaldoxime Method, 50 tests
AC2055
Tablet
0.2 – 4 mg/L
AQUAfast manganese reagent tablets
High Range, Periodate Oxidation Method, 100 tests
AC4P55
Powder
0.1 – 18 mg/L
AQUAfast manganese HR reagent powder packs*
Periodate Method, 30 tests
AC4055
Ampoule
2 – 30 mg/L
AQUAfast manganese Auto-ID ampoules*
Molybdate / Molybdenum
Mercaptoacetic Acid Method, 100 tests
AC4P42
Powder
0.5 – 66 mg/L
AQUAfast molybdate reagent powder packs
Nitrate as Nitrogen (N)
Cadmium Reduction Method, 30 tests
AC4004
Ampoule
0.2 – 1.5 mg/L
AQUAfast nitrate LR Auto-ID ampoules*
Cadmium Reduction Method, 30 tests
AC4005
Ampoule
2.0 – 7.50 mg/L
AQUAfast nitrate MR Auto-ID ampoules*
Zinc Reduction Method, 50 tests
AC2007
Tablet
0.08 – 1 mg/L
AQUAfast nitrate reagent tablets*
Cadmium Reduction Method, 30 tests
AC4007
Ampoule
5 – 50 mg/L
AQUAfast nitrate HR Auto-ID ampoules*
Diazotization (NED) Method
AC4P46
Powder
0.01 – 0.3 mg/L
AQUAfast nitrite reagent powder packs*
Diazotization (NED) Method
AC4046
Tablet
0.08 – 1 mg/L
AQUAfast nitrite Auto-ID ampoules*
Diazotization (NED) Method
AC2046
Tablet
0.01 – 0.5 mg/L
AQUAfast nitrite reagent tablets*
Ozone
DPD Method, 30 tests
AC4048
Ampoule
0.2 – 5 pH
AQUAfast ozone Auto-ID ampoules
pH
Phenol Red Method, 100 tests
AC2001
Tablet
6.5 – 8.4 pH
AQUAfast pH reagent tablets
Phosphate
Phosphate, Ortho, Phosphomolybdenum/Ascorbic Acid Method, 100 tests 
AC4P95
Powder
0.06 – 2.5 mg/L
AQUAfast phosphate (ortho) reagent powder packs*
Phosphate, Ortho, Phosphomolybdic Acid/Ascorbic Acid Method, 50 tests
AC2095-WA
Tablet
0.05 – 4 mg/L
AQUAfast phosphate (ortho) LR reagent tablets*
Ortho, Stannous Chloride Method, 30 tests
g
Reaction Tube
0.3 – 8 mg/L
AQUAfast phosphate (ortho) Auto-ID ampoules
Phosphate, Ortho, Ascorbic Acid/Vanadomolybdate Method, 50 tests
AC2096
Tablet
1 – 80 mg/L
AQUAfast phosphate (ortho) HR reagent tablets*
Silica
Heteropoly Blue Method, 30 tests
AC4060
Ampoule
0.5 – 10 mg/L
AQUAfast silica Auto-ID ampoules*
Phosphate Removal for AC2060 Method, 100 tests
AC2061
Tablet
----
AQUAfast silica phosphate removal reagent*
High Range, Silicomolybdate Method, 100 tests
AC4P60
Powder
1 – 90 mg/L
AQUAfast silica (high range) reagent powder packs*
Silicomolybdate, Heteropoly Blue Method, 50 tests
AC2060
Tablet
0.05 – 4 mg/L
AQUAfast silica reagent tablets*
Sulfate
Barium Sulfate/Turbidity Method, 100 tests
AC2082
Tablet
5 – 100 mg/L
AQUAfast sulfate reagent tablets*
Barium Sulfate/Turbidity Method, 30 tests
AC4082
Ampoule
8 – 100 mg/L
AQUAfast sulfate Auto-ID ampoules*
Barium Sulfate/Turbidity Method, 100 tests
AC4P82
Powder
5 – 100 mg/L
AQUAfast sulfate reagent powder packs*
Sulfide
Acid Soluble, Methylene Blue Method, 30 tests
AC4016
Ampoule
0.2 – 3 mg/L
AQUAfast sulfide (Acid Soluble) Auto-ID ampoule*
DPD/Catalyst Method, 50 tests
AC2016
Tablet
0.04 – 0.5 mg/L
AQUAfast sulfide reagent tablets
Zinc
Zinc, Zincon Method, 50 tests
AC2065
Tablet
0.02 – 1 mg/L
AQUAfast zinc reagent tablets*
Zinc, Zincon Method, 30 pack
AC4065
Auto-ID Ampoule
0.3 – 3 mg/L
AQUAfast zinc Auto-ID ampoules*
*Compliant with US EPA and other global testing methods for COD in wastewater
Accessories for use with the AQUAfast AQ4000 Colorimeter
Cat. No.
Description
AC2V16
Replacement 16mm vials, 10/pack
Shop
AC2V24
Replacement 24mm vials, 12/pack
Shop
AC3CBR
Vial cleaning brush, 5/pack
Shop
AQ4CBL
RS232 cable for data download and software update
Shop
AQ4ZER
AQUAfast Zero Kit
Shop
COD165
Digestion methods thermoreactor, pre-set for 100°, 120°, 150°, 160°, and 165°C 
Shop
CODS01
COD standard, 1000ppm, 475mL 
Shop
CODS10
COD standard, 10000ppm, 475mL 
Shop
CLSK100
Chlorine primary standard, 1.5mg/L ampoule kit
Shop
AC3ROD
Crush and stir rod for tablets and AC2V24 vials
Shop
AQUAfast Reagents for the AQUAfast 3170 Colorimeter
Chemical species
Method Description
Method
Form Factor
Range
Shop
Chlorine
Free, DPD Method, 100 tests
AC4P71
Powder
0.02 – 2 mg/L Cl2; 0.1 – 8 mg/L Cl2+
AQUAfast chlorine (free) reagent powder packs*
Free, DPD bulk powder dispenser with one vial x 250 tests
AQ250F
Powder
0.02 – 2 mg/L Cl2
AQUAfast free chlorine DPD bulk powder dispenser*
Free, DPD powder refill for AQ250F dispenser, 1 vial x 250 tests
AC71P1
Powder
0.02 – 2 mg/L Cl2
AQUAfast free chlorine DPD powder refill for AQ250F dispenser*
Free, DPD powder refill for AQ250F dispenser, 2 vials x 250 tests each
AC71P2
Powder
0.02 – 2 mg/L Cl2
AQUAfast free chlorine DPD powder refill for AQ250F dispenser*
Total, DPD Method, 100 tests
AC4P72
Powder
0.02 – 2 mg/L Cl2; 0.1 – 8 mg/L Cl2+
AQUAfast chlorine (total) reagent powder packs*
Total, Bluk DPD powder dispenser with one vial x 250 tests
AQ250T
Powder
0.02 – 2 mg/L Cl2
AQUAfast total chlorine DPD powder dispenser with one vial x 250 count*
Total, DPD powder refill for AQ250T dispenser, 1 vial x 250 tests
AC72P1
Powder
0.02 – 2 mg/L Cl2
AQUAfast total chlorine DPD powder refill for AQ250T dispenser*
Total, DPD powder refill for AQ250T dispenser, 2 vials x 250 tests each
AC72P2
Powder
0.02 – 2 mg/L Cl2
AQUAfast free chlorine DPD powder refill for AQ250F dispenser*
† Use AC2V10 short path length plastic vials and two powder packs.
*Compliant with US EPA and other global testing methods for COD in wastewater
Accessories for use with the AQUAfast AQ3170 Colorimeter
Cat. No.
Description
AC2V24
Replacement 24mm vials, 12/pack
Shop
AC2V10
AQUAfast 10mm short path length plastic vials for high-level chlorine, 12/pack
Shop
CLSK100
Chlorine primary standard, 1.5mg/L ampoule kit
Shop
CLSK200
Chlorine secondary standards (0.0, 0.2, and 1mg/L)
Shop
AC3CBR
Vial cleaning brush, 5/pack
Shop
AQUAfast Reagents for the AQUAfast 3140 COD Colorimeter
Chemical species
Method Description
Method
Form Factor
Range
Shop
COD
Low Range, Dichromate Digestion Method, 25 Tests
CODL00
Digestion Tube
0 – 150mg/L
AQUAfast COD 0-150 ppm digestion tubes*
Low Range, Dichromate Digestion Method, 150 Tests
CODL150
Digestion Tube
0 – 150mg/L O2
AQUAfast COD 0-150 ppm digestion tubes*
Mid Range, Dichromate Digestion Method, 25 Tests
CODH00
Digestion Tube
0 – 1,500mg/L
AQUAfast COD 0-1500 ppm digestion tubes*
Mid Range,Dichromate Digestion Method, 150 Tests
CODM150
Digestion Tube
0 – 1,500mg/L O2
AQUAfast COD 0-1500 ppm digestion tubes*
High Range, Dichromate Digestion Method, 25 Tests
CODHP0
Digestion Tube
0 – 15,000mg/L
AQUAfast Digestion Tubes, Range: 0-15000 mg/L O2 COD High Range*
High Range, Dichromate Digestion Method, 150 Tests
CODH150
Digestion Tube
0 – 15,000mg/L O2
AQUAfast Digestion Tubes, Range: 0 to 15000 mg/L O2, COD High Range*
*Compliant with US EPA and other global testing methods for COD in wastewater
Accessories for use with the AQUAfast AQ3140 COD Colorimeter
Cat. No.
Description
COD165
Digestion methods thermoreactor, pre-set for 100°, 120°, 150°, 160°, and 165°C 
Shop
CODS01
COD standard, 1000ppm, 475mL 
Shop
CODS10
COD standard, 10000ppm, 475mL 
Shop
Request a consultation
virtual-demo-image-only
Get answers to your top water analysis and water purification questions and see products in action. Learn about:
·  pH & Multiparameter Benchtop & Portable Meters
·  Electrodes (pH Electrodes, ISE, Conductivity, DO)
·  Potentiometric Titrators
·  Spectrophotometers
·  Water Purification Systems
Make Request