Fast, accurate DNA, RNA, and protein quantification, one sample at a time
The Thermo Scientific NanoDrop One/OneC Spectrophotometer is a compact, stand-alone UV-visible spectrophotometer for measurement of highly concentrated samples across a wide variety of analytes without the need for dilution. A glove-compatible touchscreen offers easy access to preloaded software applications and enables purity and concentration assessments in seconds. Built-in Acclaro Sample Intelligence technology helps identify and correct for the contaminants present. Make informed decisions on sample quality and improve your success in downstream applications with NanoDrop technology.
Choose the NanoDrop One instrument for pedestal-only sampling or the NanoDrop OneC instrument for pedestal or cuvette sampling and an extended dynamic range.
Video Player is loading.
Current Time 0:00
Duration 3:01
Loaded: 0.00%
Stream Type LIVE
Remaining Time 3:01
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Watch the NanoDrop One Spectrophotometer in action
    NanoDrop One and Oneᶜ Spectrophotometer features
    Our flagship NanoDrop One and OneC spectrophotometers share common features with other members of the NanoDrop family:
    Pioneering pedestal technology
    Simply pipet a drop of your DNA, RNA, or protein sample on the pedestal and pull down the arm; results appear in seconds
    Advanced sample intelligence
    By analyzing absorbance across the full spectrum, Acclaro Sample Intelligence software can identify contaminants and correct quantitation results before you use them in downstream applications
    Rapid time to results
    Results appear within seconds of initiating a measurement, and Auto-Blank and Auto-Measure settings can further streamline your workflow
    Tiny sample sizes
    Requires only 1–2 µL of sample without dilution, even for highly concentrated samples
    Differentiates DNA from RNA
    Acclaro software uses full-spectrum data to tease apart DNA from RNA and calculate corrected nucleic acid concentrations
    Your dependable lab partner
    Since their introduction in 2001, NanoDrop spectrophotometers have become indispensable in many labs, accumulating over 55,000 scientific citations
    In addition, the NanoDrop One/OneC Spectrophotometer offers indispensable features that make it a reliable, easy-to-use laboratory workhorse for many sample and assay types, including regulated environments. To learn more, see the sections below.
    Expanded dynamic range
    Measure up to 27,500 ng/µL dsDNA or 400 mg/mL IgG without dilution
    Streamlined multi-sample processing
    With instant Auto-Measure and Auto-Blank capabilities
    Cuvette option
    Choose the NanoDrop One instrument for sample pedestal only or the NanoDrop OneC instrument for both pedestal and cuvette
    Advanced connectivity
    Connect, export, print, and view data remotely
    Optional security software for compliance
    Security Suite software aids compliance with FDA 21 CFR Part 11 electronic recordkeeping regulations
    Wide spectral range
    Measure the full UV-Vis spectrum from 190–850 nm, allowing a range of assays as shown in the table below
    Routine and custom embedded applications
    DNA, RNA, and protein quantification, full spectral data, purity ratios (A260/A280, A260/A230), and more
    Local or PC control software
    Operate your instrument from the glove-compatible touchscreen or your Ethernet-connected PC, including custom methods and data export capabilities
    Onboard integrated Learning Center
    A repository of technical support documents and educational animations at your fingertips
    Sample or analysis type
    Wavelength
    Peptides
    205 nm
    DNA and RNA
    260 nm
    Purified protein
    280 nm
    Toxicology assays and industrial dyes
    490 nm
    Gold nanoparticles
    520 nm
    Colorimetric protein assay: BCA
    562 nm
    Colorimetric protein assay: Bradford
    595 nm
    Colorimetric protein assay: Modified Lowry
    650 nm
    Colorimetric protein assay: Pierce 660
    660 nm
    Optical density measurements
    600 nm
    Powerful, flexible UV-Vis spectrophotometer operating and compliance software
    The high-definition, glove-compatible touchscreen interface of NanoDrop One/OneC spectrophotometers give you complete onboard control of your instrument. The user-friendly interface includes custom methods and data export capabilities. An integrated, onboard Learning Center maintains an archive of technical support documents and educational animations at your fingertips.
    In addition to the onboard touchscreen interface, free NanoDrop One/OneC PC Control software allows user control of the instrument from an Ethernet-connected PC. 
    For regulated labs, optional Thermo Scientific Security Suite for NanoDrop One/OneC allows instrument use in a 21 CFR Part 11 compliant environment. This module helps to maintain the records required for user account access, audit trails, electronic signatures, automated data back-up, and more. 
    NanoDrop One Spectrophotometer offers one-touch access to applications, even with gloved hands
    Onboard touchscreen interface. The ergonomic, standalone NanoDrop One/OneC instruments offer onboard control via a high-definition, glove-compatible touchscreen.
    Acclaro Sample Intelligence Technology
    Thermo Scientific Acclaro Sample Intelligence Technology software (included with NanoDrop One/OneC and NanoDrop Eight spectrophotometers) analyzes absorbance across the whole UV-Vis spectrum, not just wavelengths specific to nucleic acids and proteins. It then applies powerful chemometric algorithms to find and correct for contamination in your samples. You get more reliable downstream assay results and avoid potential rework with this efficient, breakthrough capability.
    Video Player is loading.
    Current Time 0:00
    Duration 3:38
    Loaded: 0.00%
    Stream Type LIVE
    Remaining Time 3:38
    1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
      • en (Main), selected
      Next-generation quality assessment using Acclaro software
      Chemometric algorithms allow NanoDrop instruments to identify and correct for specific contaminants, taking quality assessment to the next level.
      Acclaro Sample Intelligence software can even use full-spectrum data to tease apart DNA from copurified RNA and calculate corrected DNA concentrations.
      Verify sample quality in seconds
      Identify sample contaminants and obtain corrected concentration results
      Identify contaminants that absorb in the analysis region
      Determine absorbances from known contaminants like protein or phenol in dsDNA or RNA samples or nucleic acids or phenol in protein samples 
      Rely on accurate results
      Help ensure measurement integrity with embedded sensor and digital image analysis
      Access information through alerts
      Receive instant feedback about sample quality with on-demand technical support and guided troubleshooting
      Analyze the entire spectrum to expose other contaminants
      Detect the presence of guanidine HCl and guanidinium isothiocyanate in nucleic acid samples, dsDNA in RNA, or RNA in dsDNA
      Advanced connectivity for data storage, printing, and sharing
      Improve your laboratory’s productivity and expand your collaborations with connected laboratory instruments that accelerate your research. With 24/7 access to the Thermo Fisher Connect cloud-based platform using the free NanoDrop Cloud App, you can securely upload, store, view, and share your data anytime, anywhere, from any device. 
      Or, when you want to work at your desk, you can use NanoDrop One PC Control software to view and analyze your data or explore Acclaro support features on your PC. Then, when your results are ready, you can print directly or seamlessly transfer results to a PC or the Connect platform using a USB, *Wi-Fi, or Ethernet connection. 
      Wi-Fi model not available in certain regions; consult your local NanoDrop distributor.
      Choose the pedestal-only or pedestal/cuvette option
      NanoDrop OneC instrument offers built-in cuvette capability
      Although both models in the NanoDrop One family have the pioneering microvolume sample retention capability of our pedestal, the NanoDrop OneC instrument includes a built-in cuvette capability. The cuvette position adds experimental flexibility and increases the dynamic range. Combining the pioneering sample-retention system with cuvette capability accommodates both low and high concentrations (0.2–27,500 ng/µL dsDNA and 0.006–820 mg/mL BSA).
      Use cuvettes to measure dilute samples and optical density of bacterial cultures or to perform kinetics experiments. The cuvette position can be used with the arm up or down and includes temperature control and stirring features.
      NanoDrop One and Oneᶜ Spectrophotometer customer stories
      Video Player is loading.
      Current Time 0:00
      Duration 1:33
      Loaded: 0%
      Stream Type LIVE
      Remaining Time 1:33
      1x
        • Chapters
        • descriptions off, selected
        • captions off, selected
        • en (Main), selected
        NanoDrop One Spectrophotometer - Scientist Feedback, University of Cambridge - Robyn Hardisty
        Watch this NanoDrop One testimonial video with a University of Cambridge scientist who studies the chemical biology of nucleic acids.
        Video Player is loading.
        Current Time 0:00
        Duration 2:51
        Loaded: 0.00%
        Stream Type LIVE
        Remaining Time 2:51
        1x
          • Chapters
          • descriptions off, selected
          • captions off, selected
          • en (Main), selected
          NanoDrop One UV-Vis Spectrophotometer Review – Alyssa Grantham
          See what an experienced, young scientist working on amyotrophic lateral sclerosis (ALS) research has to say about her first impressions of the NanoDrop One instrument.
          Whether you are ...
          starting your career,
          developing your career,
          or progressing your career,
          NanoDrop One UV-Vis spectrophotometers are with you every step of the way.
          Peter Tao – Research Assistant, Neurobiology laboratory, Ocular degeneration research Alyssa Grantham – Research Assistant, Neurobiology laboratory, Amyotrophic lateral sclerosis disease research Angela Botes Ph.D - Postdoctoral Associate, Biology laboratory, Metabolic engineering and synthetic biology research
          Peter Tao Alyssa Grantham Angela Botes
          "The compactness of the instrument most definitely stands out from everything else. Well, a lot of things stand out about it but the compactness is the best part for me because coming from a lab where we don't have that much space we want more of it…” "…the auto measure makes sampling faster, easier…I try to figure out the fastest way to read samples when I'm short on time. So I'll get one sample ready while one sample's reading. So I can just set it up, I don’t have to click a button to set GO, I just put it (the pedestal arm) back down and my next sample is ready. "I tend to forget what the (purity) ratio should be. I have a rough idea of what the ratio with protein contamination should be...and this is very helpful to have that extra feature (educational animations) that will correct the concentration for you so what you think your DNA concentration is, is what (the) actual DNA concentration is. That's gonna save a lot of time.”
          NanoDrop One and Oneᶜ Spectrophotometer specifications
          Category
          Attribute
          Description
          Samples
          Minimum Sample Size
          1 μL
          Sample Number
          1
          Limit of Detection
          Pedestal: 2 ng/μL (dsDNA), 0.06 mg/mL (BSA)
          Cuvette (NanoDrop OneC only): 0.2 ng/μL (dsDNA), 0.006 mg/mL (BSA)
          Maximum Concentration
          Pedestal: 27,500 ng/μL (dsDNA), 820 mg/mL (BSA)
          Cuvette (NanoDrop OneC only): 75 ng/μL (dsDNA)
          Optics
          Pathlength(s)
          0.030 to 1.0 mm automatically adjusting
          Light Source
          Xenon flash lamp
          Detector Type
          2048-element CMOS linear image sensor
          Wavelength Range
          190–850 nm
          Wavelength Accuracy
          ±1 nm
          Spectral Resolution
          ≤1.8 nm (FWHM at Hg 254 nm)
          Typical Measurement Repeatability
          0.002 A (1.0 mm path) or 1% CV, whichever is greater
          Absorbance Accuracy1
          3% (at 0.97 A, 302 nm)
          Absorbance Range (10 mm equivalent)
          Pedestal: 0–550A
          Cuvette (NanoDrop OneC only): 0–1.5A
          Measurement and Data Processing Time
          8 seconds
          21 CFR Part 11 Compliance
          Yes, optional
          Characteristics
          Instrument Control
          Built-in touchscreen or PC Control with free PC software
          Footprint (W × D × H)
          20 × 25.4 × 32.3 cm (8 × 10 × 12.7 in.)
          Weight
          3.6 kg
          Sample Pedestal
          303 stainless steel and quartz fiber
          Cuvette Position
          Optional (with stirring) (NanoDrop OneC only)
          Operating Voltage
          12 V (DC)
          Operating Power Consumption
          12–18 W
          System
          Instrument Control
          Built-in touchscreen
          Applications Support
          Nucleic Acid A260, A260/A280, A260/A230, Labeled Nucleic Acids, Microarray, dsDNA, ssDNA, RNA, Oligo DNA/Oligo RNA, RNA/DNA Deconvolution, Custom Factor; Protein A280 and A205, Protein Pierce 660, Protein Bradford, Protein BCA, Protein Lowry, Proteins & Labels. Protein Editor; Dye Editor, OD600, Kinetics, UV-Vis, and Custom Methods
          Language Support
          Chinese, French, German, Japanese, Korean, Polish, Spanish, English
          Accessory Support
          DYMO® LabelWriter® 550 Turbo printer, Bluetooth keyboard, mouse, and barcode reader
          Software Compatibility
          Standalone, Local Control software
          NanoDrop One PC Control (free download)
          Thermo Scientific Security Suite (purchase separately)
          PC Software: Windows® 10 Professional (64 bit)
          Connectivity
          Three USB-A ports, Ethernet, Bluetooth, and Wi-Fi*
          Advanced Connectivity
          NanoDrop One Cloud Application2
          Thermo Fisher Connect Platform
          1 Absorbance expressed as Abs/mm measured at 25 °C.
          2 Visit www.thermofisher.com/connect for details.
          NanoDrop One and Oneᶜ Spectrophotometer ordering information