Detecting low-abundance antigens with even the best conventional dye conjugates can be a challenge, as photobleaching can make it difficult to effectively observe and record staining. The exceptional photostability of Qdot probes conjugates can provide substantial benefits in the detection of low abundance targets. Due to their narrow and symmetric emission spectra, Qdot probes are also ideal for multicolor, multiplexed fluorescence detection using a single excitation source such as the 405 nm laser.
Qdot streptavidin conjugates and conjugation kits
Qdot streptavidin conjugates combine the unsurpassed photostability of Qdot probes with the highly specific binding properties of streptavidin. These powerful fluorescence detection reagents offer unique performance advantages in a wide variety of tissue labeling and flow cytometry experiments; they are efficiently excited using the 405 nm violet laser, and the Qdot probes fluorescence is extremely resistant to photobleaching.
Qdot streptavidin conjugate contents
These products provide 200 µL of the Qdot streptavidin conjugate and a detailed protocol outlining the use of the product in immunohistochemistry (IHC) applications using fluorescence microscopy. At conventional working concentrations, ~100 staining experiments may be performed
Qdot strepavidin conjugate sampler kit contents
Each Qdot Streptavidin Sampler Kit provides 50 µL of each of six colors of Qdot streptavidin conjugate (525, 565, 585, 605, 655, and 705 nm emission) and a detailed protocol describing the use of the product in immunohistochemistry (IHC) applications using fluorescence microscopy. At conventional working concentrations, ~20 staining experiments with each color may be performed.
Catalog # Name Size Price (USD) Qty
Q20001MP Qdot™ Incubation Buffer Each
157.00
Q10151MP Qdot™ Streptavidin Sampler Kit Each
934.00
Q10141MP Qdot™ 525 Streptavidin Conjugate, 200 μL Each
884.00
Q10143MP Qdot™ 525 Streptavidin Conjugate, 50 μL Each
272.00
Q10041MP Qdot™ 525 ITK™ Streptavidin Conjugate Kit Each
856.00
Q10091MP Qdot™ 545 ITK™ Streptavidin Conjugate Kit Each
882.00
Q10131MP Qdot™ 565 Streptavidin Conjugate, 200 μL Each
848.00
Q10133MP Qdot™ 565 Streptavidin Conjugate, 50 μL Each
262.00
Q10111MP Qdot™ 585 Streptavidin Conjugate, 200 μL Each
860.00
Q10113MP Qdot™ 585 Streptavidin Conjugate, 50 μL Each
261.00
Q10101MP Qdot™ 605 Streptavidin Conjugate, 200 μL Each
882.00
Q10103MP Qdot™ 605 Streptavidin Conjugate, 50 μL Each
265.00
Q10001MP Qdot™ 605 ITK™ Streptavidin Conjugate Kit Each
876.00
Q10301MP Qdot™ 605 Biotin Conjugate Kit Each
880.00
Q10121MP Qdot™ 655 Streptavidin Conjugate, 200 μL Each
890.00
Q10123MP Qdot™ 655 Streptavidin Conjugate, 50 μL Each
264.00
Q10021MP Qdot™ 655 ITK™ Streptavidin Conjugate Kit Each
884.00
Q10321MP Qdot™ 655 Biotin Conjugate Kit Each
880.00
Q10161MP Qdot™ 705 Streptavidin Conjugate, 200 μL Each
882.00
Q10163MP Qdot™ 705 Streptavidin Conjugate, 50 μL Each
271.00
Q10061MP Qdot™ 705 ITK™ Streptavidin Conjugate Kit Each
860.00
Q10171MP Qdot™ 800 Streptavidin Conjugate, 200 μL Each
882.00
Q10173MP Qdot™ 800 Streptavidin Conjugate, 50 μL Each
261.00
Q10071MP Qdot™ 800 ITK™ Streptavidin Conjugate Kit Each
846.00
Qdot primary and secondary antibody conjugates
Qdot probes for custom conjugations
Qdot ITK probes are designed to allow researchers to create custom conjugations. Provided as carboxyl-derivatized, PEG-derivatized, and also as a version provided in decane (i.e., not water soluble)
  • Carboxylate functionalized—for preparing custom conjugates that require high loading of biomolecules; can be coupled to amine groups of proteins and modified oligonucleotides using EDC-mediated condensation; can be conjugated to X-PEG-amine bi-functional linkers for custom reactivity and higher specificity
  • Amine-derivatized PEG functionalized—for preparing custom conjugates by reacting efficiently with isothiocyanates and succinimidyl esters, or with native carboxylic acids using water-soluble carbodiimides such as EDC; amine-derivatized PEG helps prevent non-specific interactions and provides a convenient handle for conjugation
  • Organic (provided in decane, not water soluble)—for preparing conjugates for applications calling for organic solvents such as hexane, toluene, dichloromethane, or chloroform; for researchers who want to use their own methods for transferring Qdot materials into water
Qdot wheat germ agglutinin (WGA) conjugate
Wheat germ agglutinin is a 36,000 molecular weight protein that binds to N-acetylglucosamine and N-acetylneuraminic acid (sialic acid) residues of glycoproteins and glycolipids. There are many uses for WGA conjugates including cell surface labeling and measuring retrograde neuronal transport. Our Qdot 655 wheat germ agglutinin conjugate provides highly sensitive labeling of N-acetylglucosamine residues, with low non-specific binding.
Qtracker cell labeling kits and vascular labels
Qtracker cell labeling kits employ a custom targeting peptide to deliver intensely fluorescent Qdot probes into the cytoplasm of live cells in culture. The fluorescence can be traced through several generations and is not transferred to adjacent cells in a population.
Designed to deliver intense fluorescence, Qtracker vascular labels are non-targeted Qdot probes designed to be injected into the tail vein of mice for vasculature studies.
Catalog # Name Size Price (USD) Qty
Q25041MP Qtracker™ 525 Cell Labeling Kit Each
995.65
Online Exclusive
1,046.00
Save 50.35 (5%)
Q25049 Qtracker™ 525 Cell Labeling Kit (Trial Size) Each
301.00
Q25031MP Qtracker™ 565 Cell Labeling Kit Each
1,028.00
Q21031MP Qtracker™ 565 Vascular Labels Each
764.00
Q25011MP Qtracker™ 585 Cell Labeling Kit Each
1,032.00
Q25001MP Qtracker™ 605 Cell Labeling Kit Each
1,000.00
Q25021MP Qtracker™ 655 Cell Labeling Kit Each
1,048.00
Q25029 Qtracker™ 655 Cell Labeling Kit (Trial Size) Each
298.00
Q21021MP Qtracker™ 655 Vascular Labels Each
790.00
Q25061MP Qtracker™ 705 Cell Labeling Kit Each
988.65
Online Exclusive
1,034.00
Save 45.35 (4%)
Q21061MP Qtracker™ 705 Vascular Labels Each
750.65
Online Exclusive
774.00
Save 23.35 (3%)
Q25071MP Qtracker™ 800 Cell Labeling Kit Each
1,007.65
Online Exclusive
1,048.00
Save 40.35 (4%)
Q21071MP Qtracker™ 800 Vascular Labels Each
788.00
Stylesheet for Classic Wide Template adjustments
Hero banner dark gradient overlay
For Research Use Only. Not for use in diagnostic procedures.