Microscopic views of fluorescence fade in cultured cells after 24 and 36 s exposure
Prevent fluorescence photobleaching
Loss of fluorescence through irreversible photobleaching processes can lead to a significant reduction in sensitivity, particularly when target molecules are of low abundance or when excitation light is of high intensity or long duration. The mechanisms associated with photobleaching are also implicated in phototoxicity, which may adversely impact cell viability and data quality.
To minimize photobleaching of experimental samples, we have developed a series of antifade reagents that increase fluorophore photostability in both live-cell and fixed-cell samples. We offer choices for immediate imaging and for long-term storage, plus formulations that include a nuclear stain to combine mounting and counterstaining in a single step. Reduced photobleaching means longer tracking for time-course experiments, higher sensitivity, and better quantitation from fluorescent signals.
On this page:
How to select the antifade for your imaging experiment
 ProLong GlassProLong DiamondProLong Gold
 Learn more
General specifications
FormReady-to-use hard-setting mountant
Refractive index1.52 (after curing)1.47 (after curing)1.47 (after curing)
What type of imaging am I planning?
Specimen typeAnimal cells/tissue (fixed)
Live-cell imaging   
Fixed-cell, immediate imaging   
Fixed-cell, long-term imaging (up to 10 µm sample thickness)
Checked
Checked
Checked
Fixed-cell, long-term imaging (up to 150 µm sample thickness)
Checked
What type of microscope objective can produce best results? (All objective types are compatible with these reagents)
Oil immersion objective
Checked
Glycerol-corrected objective 
Checked
Checked
Water- or air-corrected objective   
What fluorophores does my sample include?
Alexa Fluor dyes
Checked
Checked
Checked
Traditional dyes
Checked
Checked
Fluorescent proteins
Checked
Checked
Do I want to add counterstain in the mountant?
Counterstain with DAPI
Checked
Checked
Checked
Counterstain with SYTOX Deep Red
Checked
Checked
Checked
No counterstain
Checked
Checked
Checked
 SlowFade GlassSlowFade DiamondSlowFade Gold
 Learn more
General specifications
FormReady-to-use soft-setting mountant
Refractive index1.521.421.42
What type of imaging am I planning?
Specimen typeAnimal cells/tissue (fixed)
Live-cell imaging   
Fixed-cell, immediate imaging
Checked
CheckedChecked
Fixed-cell, long-term imaging (up to 10 µm sample thickness)   
Fixed-cell, long-term imaging (up to 500 µm sample thickness)
Checked
What type of microscope objective can produce best results? (All objective types are compatible with these reagents)
Oil immersion objective   
Glycerol-corrected objective
Checked
CheckedChecked
Water- or air-corrected objective   
What fluorophores does my sample include?
Alexa Fluor dyes
Checked
CheckedChecked
Traditional dyes
Checked
Checked 
Fluorescent proteins
Checked
Checked 
Do I want to add counterstain in the mountant?
Counterstain with DAPI
Checked
CheckedChecked
Counterstain with SYTOX Deep Red
No counterstain
Checked
CheckedChecked
 ProLong Live
 Learn more
General specifications
Form100x concentrated liquid
Refractive index1.3
What type of imaging am I planning?
Specimen typeAnimal cells/tissue (live)
Live-cell imaging
Checked
Fixed-cell, immediate imaging 
Fixed-cell, long-term imaging (up to 10 µm sample thickness) 
Fixed-cell, long-term imaging (up to 150 µm sample thickness) 
What type of microscope objective can produce best results? (All objective types are compatible with these reagents)
Oil immersion objective 
Glycerol-corrected objective
Checked
Water- or air-corrected objectiveChecked
What fluorophores does my sample include?
Alexa Fluor dyes
Checked
Traditional dyes
Checked
Fluorescent proteins
Checked
Do I want to add counterstain in the mountant?
Counterstain with DAPI
Counterstain with SYTOX Deep Red 
No counterstain
Checked
 ProLong Glass
(curing)
SlowFade Glass
(non-curing)
CytoVista Reagents
(non-curing)
 Learn moreLearn moreLearn more
General Specifications
FormReady-to-use, curing (hard-setting) mountant
For immediate imaging of samples up to 150 μm thick
Ready-to-use non-curing (soft-set) mountant
For immediate imaging of samples up to 500 μm thick
Ready-to-use, non-curing (soft-setting) family of reagents
3D Cell Culture clearing and staining kit for immediate imaging of 3D cell culture (organoids and spheroids up to 1 mm thick)
Tissue clearing and staining kit for immediate imaging of thick tissue (up to 10 mm)
Refractive index1.52 (after curing)1.521.48 (organoids/spheroids up to 1 mm thick)
1.53 (tissue samples up to 10 mm thick)
What type of imaging am I planning?
Specimen typeAnimal cells/tissue (fixed)Animal cells/tissue (fixed)Animal cells/tissue (fixed)
Fixed-cell imaging, immediate imaging 
Checked
Checked
Fixed-cell, long-term imaging
Checked
What is the thickness of my sample?
Up to 10 μm
Checked
Up to 150 μm
Checked
Up to 500 μm 
Checked
Up to 1 mm  
Checked
Up to 10 mm  
Checked
What type of microscope objective can produce best results? (All objective types are compatible with these reagents)
Oil immersion objective
Checked
Checked
Checked
Glycerol-corrected objective  
Checked
What fluorophores does my sample include?
Alexa Fluor dyes
Checked
Checked
Checked
Traditional dyes
Checked
Checked
Checked
Fluorescent proteins
Checked
Checked
Checked
Do I want to add counterstain in the mountant?
Counterstain with DAPI
Checked
Checked
Counterstain with SYTOX Deep Red
Checked
No counterstain
Checked
Checked
Checked
 Image-iT Plant Clearing and Mounting Reagents for Imaging
 Learn more
General Specifications
FormReady-to-use reagents for clearing and mounting plant tissue for imaging
Refractive index1.45 (clearing and soft-set mounting option)
1.415 (hard-set mounting option)
What type of imaging am I planning?
Specimen typePlant cells/tissue section/whole tissue (fixed)
Live-cell imaging 
Fixed-cell imaging
Checked
Fixed-cell imaging, long-term imaging (up to 10 μM sample thickness)
Checked
Fixed-cell imaging, long-term imaging (up to 150 μM sample thickness)
Checked
What type of microscope objective can produce best results? (All objective types are compatible with these reagents)
Oil immersion objective 
Glycerol-corrected objective
Checked
Water- or air-corrected objective 
What fluorophores does my sample include?
Most common histological stains
Checked
Fluorescent proteins 
Do I want to add counterstain in the mountant?
Counterstain with DAPI 
Counterstain with SYTOX Deep Red 
No counterstain
Checked
 ProLong GlassProLong DiamondProLong Gold
 Learn more
General specifications
FormReady-to-use hard-setting mountant
Refractive index1.52 (after curing)1.47 (after curing)1.47 (after curing)
What type of imaging am I planning?
Specimen typeAnimal cells/tissue (fixed)
Live-cell imaging   
Fixed-cell, immediate imaging   
Fixed-cell, long-term imaging (up to 10 µm sample thickness)
Checked
Checked
Checked
Fixed-cell, long-term imaging (up to 150 µm sample thickness)
Checked
What type of microscope objective can produce best results? (All objective types are compatible with these reagents)
Oil immersion objective
Checked
Glycerol-corrected objective 
Checked
Checked
Water- or air-corrected objective   
What fluorophores does my sample include?
Alexa Fluor dyes
Checked
Checked
Checked
Traditional dyes
Checked
Checked
Fluorescent proteins
Checked
Checked
Do I want to add counterstain in the mountant?
Counterstain with DAPI
Checked
Checked
Checked
Counterstain with SYTOX Deep Red
Checked
Checked
Checked
No counterstain
Checked
Checked
Checked
 SlowFade GlassSlowFade DiamondSlowFade Gold
 Learn more
General specifications
FormReady-to-use soft-setting mountant
Refractive index1.521.421.42
What type of imaging am I planning?
Specimen typeAnimal cells/tissue (fixed)
Live-cell imaging   
Fixed-cell, immediate imaging
Checked
CheckedChecked
Fixed-cell, long-term imaging (up to 10 µm sample thickness)   
Fixed-cell, long-term imaging (up to 500 µm sample thickness)
Checked
What type of microscope objective can produce best results? (All objective types are compatible with these reagents)
Oil immersion objective   
Glycerol-corrected objective
Checked
CheckedChecked
Water- or air-corrected objective   
What fluorophores does my sample include?
Alexa Fluor dyes
Checked
CheckedChecked
Traditional dyes
Checked
Checked 
Fluorescent proteins
Checked
Checked 
Do I want to add counterstain in the mountant?
Counterstain with DAPI
Checked
CheckedChecked
Counterstain with SYTOX Deep Red
No counterstain
Checked
CheckedChecked
 ProLong Live
 Learn more
General specifications
Form100x concentrated liquid
Refractive index1.3
What type of imaging am I planning?
Specimen typeAnimal cells/tissue (live)
Live-cell imaging
Checked
Fixed-cell, immediate imaging 
Fixed-cell, long-term imaging (up to 10 µm sample thickness) 
Fixed-cell, long-term imaging (up to 150 µm sample thickness) 
What type of microscope objective can produce best results? (All objective types are compatible with these reagents)
Oil immersion objective 
Glycerol-corrected objective
Checked
Water- or air-corrected objectiveChecked
What fluorophores does my sample include?
Alexa Fluor dyes
Checked
Traditional dyes
Checked
Fluorescent proteins
Checked
Do I want to add counterstain in the mountant?
Counterstain with DAPI
Counterstain with SYTOX Deep Red 
No counterstain
Checked
 ProLong Glass
(curing)
SlowFade Glass
(non-curing)
CytoVista Reagents
(non-curing)
 Learn moreLearn moreLearn more
General Specifications
FormReady-to-use, curing (hard-setting) mountant
For immediate imaging of samples up to 150 μm thick
Ready-to-use non-curing (soft-set) mountant
For immediate imaging of samples up to 500 μm thick
Ready-to-use, non-curing (soft-setting) family of reagents
3D Cell Culture clearing and staining kit for immediate imaging of 3D cell culture (organoids and spheroids up to 1 mm thick)
Tissue clearing and staining kit for immediate imaging of thick tissue (up to 10 mm)
Refractive index1.52 (after curing)1.521.48 (organoids/spheroids up to 1 mm thick)
1.53 (tissue samples up to 10 mm thick)
What type of imaging am I planning?
Specimen typeAnimal cells/tissue (fixed)Animal cells/tissue (fixed)Animal cells/tissue (fixed)
Fixed-cell imaging, immediate imaging 
Checked
Checked
Fixed-cell, long-term imaging
Checked
What is the thickness of my sample?
Up to 10 μm
Checked
Up to 150 μm
Checked
Up to 500 μm 
Checked
Up to 1 mm  
Checked
Up to 10 mm  
Checked
What type of microscope objective can produce best results? (All objective types are compatible with these reagents)
Oil immersion objective
Checked
Checked
Checked
Glycerol-corrected objective  
Checked
What fluorophores does my sample include?
Alexa Fluor dyes
Checked
Checked
Checked
Traditional dyes
Checked
Checked
Checked
Fluorescent proteins
Checked
Checked
Checked
Do I want to add counterstain in the mountant?
Counterstain with DAPI
Checked
Checked
Counterstain with SYTOX Deep Red
Checked
No counterstain
Checked
Checked
Checked
 Image-iT Plant Clearing and Mounting Reagents for Imaging
 Learn more
General Specifications
FormReady-to-use reagents for clearing and mounting plant tissue for imaging
Refractive index1.45 (clearing and soft-set mounting option)
1.415 (hard-set mounting option)
What type of imaging am I planning?
Specimen typePlant cells/tissue section/whole tissue (fixed)
Live-cell imaging 
Fixed-cell imaging
Checked
Fixed-cell imaging, long-term imaging (up to 10 μM sample thickness)
Checked
Fixed-cell imaging, long-term imaging (up to 150 μM sample thickness)
Checked
What type of microscope objective can produce best results? (All objective types are compatible with these reagents)
Oil immersion objective 
Glycerol-corrected objective
Checked
Water- or air-corrected objective 
What fluorophores does my sample include?
Most common histological stains
Checked
Fluorescent proteins 
Do I want to add counterstain in the mountant?
Counterstain with DAPI 
Counterstain with SYTOX Deep Red 
No counterstain
Checked
What is photobleaching?
Photobleaching is the degradation of fluorescent signal. Free radicals are generated when photoexcited fluorophores are exposed to oxygen, leading to the loss of signal intensity. Photobleaching can be slowed by reducing the intensity and time of exposure of the fluorophore to light. Antifade reagents were introduced to allow for greater and longer signal by lessening the photobleaching effect.
How to use an antifade to prevent photobleaching
Illustration of microscope slide with sample
Step 1. Place sample on slide or coverslip. Complete all staining prior or after this step.
Illustration of microscope slide with antifade applied
Step 2. Apply antifade mountant over sample. For a hard-setting mount, allow to cure overnight open to air.
Illustration of microscope slide with glycerol applied (hard-setting mountants only)
Step 3. Only with a hard-setting mountant: add a drop of 100% glycerol to the mounted sample and apply a coverslip. Glycerol aids in coverslip adhesion. ProLong Glass antifade mountant fully cures to a final RI of ~1.52 after 24 hours.
What is the difference between a soft- and hard-setting mountant?
Mounting media can be soft- or hard-setting, depending on whether the mountant contains a gelling agent. Soft-setting mountants contain a buffered glycerol solution and should be used when slides need to be imaged immediately. Soft-setting mountants can be washed away and the sample can either be restained or used for downstream purposes like single-cell RNA-sequencing or PCR. Hard-setting mountants contain a polymer that permanently affixes the coverslip to the slide. This is used for extended storage of a sample.
Antifades and mounts ordering information
Catalog # Name Size Price (USD) Qty
P36980 ProLong™ Glass Antifade Mountant, 5 x 2 mL Each
332.65
Online Exclusive
358.00
Save 25.35 (7%)
P36982 ProLong™ Glass Antifade Mountant, 2 mL Each
96.65
Online Exclusive
103.00
Save 6.35 (6%)
P36984 ProLong™ Glass Antifade Mountant, 10 mL Each
296.65
Online Exclusive
323.00
Save 26.35 (8%)
P36981 ProLong™ Glass Antifade Mountant with NucBlue™ Stain, 5 x 2 mL Each
344.65
Online Exclusive
377.00
Save 32.35 (9%)
P36983 ProLong™ Glass Antifade Mountant with NucBlue™ Stain, 2 mL Each
107.65
Online Exclusive
116.00
Save 8.35 (7%)
P36985 ProLong™ Glass Antifade Mountant with NucBlue™ Stain, 10 mL Each
304.65
Online Exclusive
333.00
Save 28.35 (9%)
Catalog # Name Size Price (USD) Qty
P36961 ProLong™ Diamond Antifade Mountant, 5 x 2 mL Each
298.65
Online Exclusive
322.00
Save 23.35 (7%)
P36965 ProLong™ Diamond Antifade Mountant, 2 mL Each
88.65
Online Exclusive
95.00
Save 6.35 (7%)
P36970 ProLong™ Diamond Antifade Mountant, 10 mL Each
268.65
Online Exclusive
291.00
Save 22.35 (8%)
P36962 ProLong™ Diamond Antifade Mountant with DAPI, 5 x 2 mL Each
312.65
Online Exclusive
337.00
Save 24.35 (7%)
P36966 ProLong™ Diamond Antifade Mountant with DAPI, 2 mL Each
95.65
Online Exclusive
103.00
Save 7.35 (7%)
P36971 ProLong™ Diamond Antifade Mountant with DAPI, 10 mL Each
287.65
Online Exclusive
309.00
Save 21.35 (7%)
Catalog # Name Size Price (USD) Qty
P36934 ProLong™ Gold Antifade Mountant, 5 x 2 mL Each
296.65
Online Exclusive
323.00
Save 26.35 (8%)
P36930 ProLong™ Gold Antifade Mountant, 10 mL Each
277.65
Online Exclusive
302.00
Save 24.35 (8%)
P36935 ProLong™ Gold Antifade Mountant with DNA Stains DAPI, 5 x 2 mL Each
307.65
Online Exclusive
332.00
Save 24.35 (7%)
P36931 ProLong™ Gold Antifade Mountant with DNA Stains DAPI, 10 mL Each
290.65
Online Exclusive
313.00
Save 22.35 (7%)
Catalog # Name Size Price (USD) Qty
S36917-5X2ML SlowFade™ Glass Soft-set Antifade Mountant, 5 x 2 mL Each
279.65
Online Exclusive
301.00
Save 21.35 (7%)
S36920-5X2ML SlowFade™ Glass Soft-set Antifade Mountant, with DAPI, 5 x 2 mL Each
314.00
S36920 SlowFade™ Glass Soft-set Antifade Mountant, with DAPI, 2 mL Each
93.50
S36963 SlowFade™ Diamond Antifade Mountant, 5 x 2 mL Each
262.65
Online Exclusive
284.00
Save 21.35 (8%)
S36967 SlowFade™ Diamond Antifade Mountant, 2 mL Each
79.65
Online Exclusive
84.50
Save 4.85 (6%)
S36972 SlowFade™ Diamond Antifade Mountant, 10 mL Each
231.65
Online Exclusive
252.00
Save 20.35 (8%)
S36964 SlowFade™ Diamond Antifade Mountant with DAPI, 5 x 2 mL Each
288.65
Online Exclusive
308.00
Save 19.35 (6%)
S36968 SlowFade™ Diamond Antifade Mountant with DAPI, 2 mL Each
88.65
Online Exclusive
94.75
Save 6.10 (6%)
S36973 SlowFade™ Diamond Antifade Mountant with DAPI, 10 mL Each
246.65
Online Exclusive
269.00
Save 22.35 (8%)
S36937 SlowFade™ Gold Antifade Mountant, 5 x 2 mL Each
266.65
Online Exclusive
288.00
Save 21.35 (7%)
S36936 SlowFade™ Gold Antifade Mountant, 10 mL Each
242.65
Online Exclusive
264.00
Save 21.35 (8%)
S36939 SlowFade™ Gold Antifade Mountant with DAPI, 5 x 2 mL Each
283.65
Online Exclusive
303.00
Save 19.35 (6%)
Catalog # Name Size Price (USD) Qty
V11300 CytoVista™ Tissue Clearing Reagent, 30 mL
502.00
V11324 CytoVista™ Tissue Clearing/Staining Kit 1 kit
952.00
V11325 CytoVista™ 3D Cell Culture Clearing/Staining Kit 1 kit
749.65
Online Exclusive
824.00
Save 74.35 (9%)
Examples of samples mounted with ProLong Antifade Mountants and Slowfade Antifade Mountants
Slowfade Antifade Mountant Example
Slowfade Antifade Mountant Example
Figure 1. 100 micron thick-tissue samples were stained and mounted with SlowFade Glass and Images were acquired a Zeiss LSM 710 confocal microscope. Left) Cryo preserved rat brain tissue section immunostained for GFAP with rabbit anti-GFAP and detected using Alexa Fluor 594 Plus goat anti-rabbit. Nuclei were counter-stained with DAPI. A 63X oil immersion objective was used and the resulting image represented as a maximum intensity projection. Right) Mouse mammary tissue section probed with mouse anti-histone H3 and rabbit anti-pan actin. Targets were detected with Alexa Fluor 488 Goat Anti-Mouse (Cat. No. A11029) and Alexa Fluor 594 Goat Anti-Rabbit (Cat. No. A11012) secondary antibody conjugates. The tissue was counter-stained with SYTOX DR (Cat. No. S11381). A 40X oil immersion objective was used and the resulting image represented as a maximum intensity projection.
ProLong Antifade Mountant Example
ProLong Antifade Mountant Example
Figure 2. Cryo-preserved rat brain sections (100-µm thick) were stained for tubulin (red) with Mouse Anti-Beta3-Tubulin (Cat. No. MA1-118) and GFAP (yellow) Rabbit Anti-GFAP (Cat. No. OPA1-06100). Targets were detected with Alexa Fluor Plus 594 Goat Anti-Mouse (Cat. No. A-11032) and Alexa Fluor Plus 647 Goat Anti-Rabbit (Cat. No. A-32733). Nuclei (cyan) were stained with DAPI (Left, Cat. No. D1306). Slides were mounted with ProLong Glass Antifade Mountant and imaged with a Zeiss LSM 710 confocal microscope using 63×oil Immersion.
Publication showing stripping and FISH reprobing after imaging  with SlowFade Gold Antifade Mountant (Cat. No. S36936)
  •  Sountoulidis A, Liontos A, Nguyen HP, Firsova AB, Fysikopoulos A, Qian X, Seeger W, Sundström E, Nilsson M, Samakovlis C. SCRINSHOT enables spatial mapping of cell states in tissue sections with single-cell resolution. PLoS Biol. 2020 Nov 20;18(11):e3000675. PMID: 33216742.
Imaging Protocol Handbook Cover
Imaging protocol handbook
Protocols that fit your needs in imaging ranging from sample and assay preparation to staining, labeling, and data analysis strategies.
Download now
Stylesheet for Classic Wide Template adjustments
Hero banner dark gradient overlay
For Research Use Only. Not for use in diagnostic procedures.