Powerful, intuitive software
The Attune Cytometric Software is designed to facilitate powerful data acquisition and analysis using an intuitive user-friendly interface. Experiment settings can easily be set up, optimized, customized, and saved for future studies. Voltration and compensation are automated and can be set up using a guide. The software is designed to maximize efficiency in performing data analysis, with fast refresh rates for large data sets (up to 20 million events per sample) with the ability to immediately visualize changes on data plots as you make adjustments.
The software has exceptional tools to simplify experimental setup, including reagent selection using the filter configuration manager. This allows guidance for matching the right reagent to the optimized channel on the instrument by selecting reagents from a drop-down menu of prepopulated or customized reagents, which is then applied to plot labels.
Intuitive software interface
Intuitive software interface
The user interface is divided into four panels. Ribbons and tabs in the top panel enable easy selection of key functions. Collection Panel is where you can easily set parameters to acquire data and see acquisition status. The Experiment Workspace is where you can preview your panel and choose from a variety of plot types and a range of statistics to illustrate your analysis. This workspace is also where you can set Smart gate labeling. The Experiment Explorer makes it easy to manage samples and data. This panel is where you can set up batch processing and instrument and compensation settings for new experiments.
“The Attune NxT software is very user friendly and intuitive, significantly reducing training time required to get new users up-and-running.”
James Hemphill,
Ph.D
Senior Scientist
Kytopen
Image processing with the Attune Cytometric Software version 7.1
The Attune Cytometric Software version 7.1 enables high throughput automated image processing of brightfield images and innovative AI data analysis solutions from data generated on the Attune CytPix flow cytometer. High resolution brightfield images show more descriptive features about cells or particles of interest such as size, complexity, particle interaction, and shape, that have previously not been available in tandem with fluorescence and scatter parameters. The software employs machine learning models to automate image processing and generate large measurement datasets rapidly from thousands of Attune CytPix cytometer images. These datasets are provided to the user as distinct image-derived parameters for plotting against fluorescence or scatter data and back-gating with images. You can choose to gate your samples using these parameters to improve gating strategy, verify sample quality, delineate complex samples and develop new applications. These morphology parameters can be utilized for image similarity and dimensional reduction tools for improved resolution into your populations of interest. See table “Image processing parameters” below for more details.
Our suite of image data analysis tools is the main feature of the Attune Cytometric Software version 7.1. We provide workflow wizards that help users with many levels of experience to access the AI machine learning tools. The Image processing feature is intuitive to use and can be incorporated into user-defined scenarios. You can choose a vendor-supplied model that is pre-trained on leukocytes and beads or refine a model to your specifications using the Train Model feature. The system can process images at speeds up to 1,000 images/second, depending on the size and complexity of images. Image-derived data can be exported into the FCS files. Our Model Explorer allows you to visualize stored models, export to share, and import for later use.
diagram representing the workflow for image processing
Screenshot of software buttons
Screenshot of new features in the Attune Cytometric Software interface.
Software features
Core capabilities
  • Compensation—fully automated and manual compensation
  • Baseline Functional Response (BFR)—automates the detector optimization process
  • Instrument tracking—automated baseline and performance test for all channels and linearity
  • Traceability—21 CFR part 11 option for flow cytometry data security, auditing, and electronic signature capabilities
  • File formats—FCS 3.0 and FCS 3.1
Maintenance
  • Automated maintenance—startup (<15 min), shutdown, rinse, wash, debubble, deep clean, monthly decontamination
  • User account maintenance—administrative and individual accounts with user log
Visualization
  • Image gallery view of imaged events
  • Back-gating of images onto flow plots
  • Visualize images from specific gate
  • Gates—standard, customizable, and derived gates
  • Plot previews—easy workspace setup
  • Zoom filmstrip viewer—easy visualization and navigation of plots to set gates and plot attributes
  • Overlay plot builder
  • Heatmap view with color-coding and statistics 
  • Graphics resolution—publication-quality images (customizable)
Image processing with ACS 7.1 & Attune CytPix flow cytometer
  • Automated image processing using 36 morphology parameters
  • Choice of 3 processing pre-trained models and unlimited user trained models with Train Model Wizard
  • Correlate morphology with standard flow cytometric fluorescence and scatter parameters
Data analysis with ACS 7.1 & Attune CytPix flow cytometer
  • Dimensionality reduction wizard that integrates morphology and fluorescence data
  • Image similarity calculations to quickly find events of similar morphology
  • Down Sampling and Concatenation wizard to manage and merge large datasets efficiently
Compensation tools
Compensation for spectral overlap between fluorescence channels is both rapid and accurate using a guided software system. The software streamlines the compensation process by performing automated voltration (Baseline Functional Response) with Attune Performance Tracking beads, providing starting points for voltages that can still be adjusted if desired. The system adjusts the compensation and applies it to the samples so that you only need to prepare the proper controls and adjust the voltage gates.
Automated compensation supports both negative and universal negative (unstained) controls. The workflow also allows you to set up and collect compensation controls directly from a plate if needed. This automation eliminates trial and error and delivers compensation rapidly and accurately. In addition, on-plot compensation adjustment allows for fine tuning, and compensation can be subsequently modified.
How to use negative gating compensation
How to use negative gating compensation
(1) Negative gating is useful for heterogeneous samples that are often found in tumor and blood samples. (2) Choose fluorophore-labeled antibodies based on the emitted emission spectrum. Dyes should have minimally overlapping spectra. (3) Prepare single-stained controls. An unstained control is not required. (4) Fluorescent compensation beads can also be used to set compensation parameters when there is not enough sample. (5) Use the guided system and select for negative gate. (6) Run sample and adjust the voltages to see the fluorescent label on the plot. Press Apply Compensation.
Baseline Functional Response
One of the key barriers to increased adoption of flow cytometry is its inherent complication—the myriad of instrument settings users must expertly adjust before acquiring data. To obtain high-quality fluorescent flow cytometer data, well-optimized instrument settings are required. Baseline Functional Response automatically sets the optimal voltage for each detector, enabling users to optimize their experiments quickly by suggesting the minimum voltage required for the best signal. 
Schematic showing 5 steps for manual optimization with up to 14 samples compared to Baseline Functional Response software reduction to 3 steps, 1 sample and 1 adjustment
Schematic demonstrating Baseline Functional Response software feature on instrument set-up steps. 
Optional 21 CFR Part 11 Software
The FDA released the Electronic Records and Signatures Rule, known as 21 CFR Part 11 in August 1997.* This rule defines the requirements for use of electronic documents in place of paper documents. The law specifies the system elements, controls, and procedures that are necessary to help ensure the reliability of electronically stored records.
This software license for the Attune acoustic focusing cytometers supports compliance with 21 CFR Part 11 FDA guidelines for Security, Auditing, and Electronic Signatures using a SAE console. The Attune 21 CFR software is available for use only for flow cytometry data on Windows 10 computers.
* https://www.fda.gov/regulatory-information/search-fda-guidance-documents/part-11-electronic-records-electronic-signatures-scope-and-application#i (accessed February 27, 2020)
Advanced data analysis software
In collaboration with De Novo Software, Thermo Fisher Scientific offers FCS Express Flow Cytometry Analysis Software. FCS Express software gets you from raw data to easily understandable, beautifully formatted, presentation-ready results more easily and in less time than any other flow cytometry software. This product consists of a one-year license for FCS Express software obtained via digital download.
FCS Express software features include:
  • Fully integrated analysis, statistics, graphing, and reporting
  • Microsoft Office inspired interface that makes learning easy, while integrated spreadsheets and direct high-resolution exports save you from using multiple software tools to achieve a final, publication-ready result
  • Full support for Attune instruments that helps ensure a seamless transition from instrument acquisition to final result
  • Dedicated and knowledgeable support team with less than 24-hour response times
Additional software licenses