To construct cDNA libraries, start with mRNA isolation with dynabeads
Reusable-cDNA-Libraries1
When you isolate mRNA using Dynabeads, you can also easily construct cDNA libraries for use in cDNA amplification, cDNA cloning, RACE experiments and subtractive hybridization.
Benefits of Dynabeads for constructing cDNA libraries
  • Isolates high-quality and full-length mRNA
  • Superior sensitivity allows you to generate libraries even from a single cell
  • Isolated mRNA is already primed for reverse transcription
  • Enzymatic applications are not inhibited by the Dynabeads
  • Dynabeads can be used with any reverse transcriptase
  • The solid-phase cDNA library is very stable, reusable and can be stored at 2-8°C
  • Optimal template for PCR-amplification, cDNA cloning, RACE and subtractive hybridization!
Solid-Phase cDNA synthesis
When preparing solid-phase cDNA libraries, you first capture the mRNA onto the Dynabeads. The beads are then added directly to the RT-reaction, where the bead-bound oligo-dT residues act as primers for first strand cDNA synthesis. As all operations utilize the solid-phase, buffer changes are fast, simple and effective.
Re-Usable cDNA library
A stable, covalently linked cDNA library is produced and can be used over and over again (e.g for PCR-amplification, RACE, subtractive hybridisation, cDNA cloning etc.). Optimal buffer conditions are achieved by simply magnetic separation and resuspension in the appropriate buffer. Reverse transcription will only be limited by the efficiency of the enzyme used.
The bead-bound probe used both for capture and primer can be oligo-dT or any specific DNA sequence. The patented Dynabeads solid-phase cDNA synthesis technology is compatible with a various commercially available cDNA kits.
The use of Dynabeads Oligo(dT)25 for capture of mRNA for cDNA synthesis and/or subtractive cloning is covered by the patent "Process for producing" cDNA (WO9006044, EPO444119, AT122721T, DE68922743T, DE68922743D, CA2003500, AU627815, US5759820 and JP2960776B2).
Order Dynabeads kits for mRNA purification
Catalog # Name Size Price (USD) Qty
61002 Dynabeads™ Oligo(dT)25, 2 mL 2 mL
771.65
Online Exclusive
808.00
Save 36.35 (4%)
61005 Dynabeads™ Oligo(dT)25, 5 mL Each
1,007.65
Online Exclusive
1,054.00
Save 46.35 (4%)
61006 Dynabeads™ mRNA Purification Kit (for mRNA purification from total RNA preps) Each
924.65
Online Exclusive
982.00
Save 57.35 (6%)
61011 Dynabeads™ mRNA DIRECT™ Purification Kit, 5 mL Each
1,031.65
Online Exclusive
1,092.00
Save 60.35 (6%)
61012 Dynabeads™ mRNA DIRECT™ Purification Kit, 10 mL Each
1,427.65
Online Exclusive
1,558.00
Save 130.35 (8%)
61021 Dynabeads™ mRNA DIRECT™ Micro Purification Kit Each
1,077.65
Online Exclusive
1,138.00
Save 60.35 (5%)
12321D DynaMag™-2 Magnet Each
878.00
12027 DynaMag™-96 Side Skirted Magnet Each
1,174.65
Online Exclusive
1,236.00
Save 61.35 (5%)
Selected References
  1. Jakobsen KS et al. (1994). Direct mRNA Isolation Using Magnetic Oligo (dT) Beads: A Protocol for All Types of Cell Cultures, Animal and Plant Tissues. Advances in biomagnetic separation. Eaton Publishing 61-71.
  2. Karrer EE et al. (1995).In situ isolation of mRNA from individual plant cells: creation of cell specific libraries. Proc.Natl.Acad.Sci.USA 92:3814-3818.
  3. Lambert KN and Williamson VM (1993). cDNA library construction from small amounts of RNA using paramagnetic beads and PCR. Nucleic Acids Research 21(3):775-776.
  4. Raineri I, Moroni C and Senn HP (1991). Improved efficiency for single-sided PCR by creating a reusable pool of first-strand cDNA coupled to a solid phase. Nucleic Acids Res. 19(14):4010.
  5. Fellmann F, Pretet J and Fellmann D (1996). Simplified Protocol of Solid-Phase cDNA libraries for Multiple PCR Amplification. BioTechniques 21(5): 766-770.
  6. Rodriguez IR et al.(1994). Structural Analysis of the Human Hydroxyindole-O-methyltransferase Gene. J.Biol.Chem. 269(50):31969-31977.
  7. Aasheim HC, Logtenberg T and Larsen F. (1996). Subtractive Hybridization for the Isolation of Differentially Expressed Genes Using Magnetic Beads. Methods in Molecular Biology 69:115-128. 
How to use Dynabeads for immunoprecipitation
Video Player is loading.
Current Time 0:00
Duration 5:10
Loaded: 0.00%
Stream Type LIVE
Remaining Time 5:10
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Related Pages
    For Research Use Only. Not for use in diagnostic procedures.