Lab Equipment – CTS Series offers our most advanced technology 
Cell therapy production in cGMP environments require specialized equipment with features that help ensure regulatory compliance can be met at every stage. Thermo Scientific Lab Equipment – CTS Series offer our most advanced technology in each product paired with a document package, and compliance services. These packages support your GMP needs for cell and gene therapy manufacturing. They help you get up and running faster, stay compliant, support regulatory audits, and stay on schedule. 
Explore Thermo Scientific Lab Equipment – CTS Series below to learn more about products to support cell and gene therapy production.
Biological safety cabinets for cell and gene therapy
Ideal for demanding applications that require safe handling of products and protection of personnel and the environment, Herasafe 2030i Biological Safety Cabinets – CTS Series are ideally suited for cell and gene therapy production. 
Smarter protection that never takes a day off
Thermo Scientific Herasafe 2030i Biological Safety Cabinet – CTS Series is designed to maximize sample protection and user safety, with an emphasis on containment, comfort and convenience, all day, every day. The CTS Series combines a comprehensive documentation package saving valuable time and giving piece of mind.
gene therapy bsc
Centrifugation for cell and gene therapy
Thermo Scientific General Purpose Pro Centrifuges – CTS Series are designed to meet the needs of today’s rapid-fire discoveries, with updates to help you get your work done more quickly, consistently, and with powerful reliability.
Powerful at every turn
Thermo Scientific General Purpose Pro Centrifuges – CTS Series include both benchtop and floor models within our 4L range. They meet the need for an essential separation equipment and come with the necessary compliance documentation and validation services for cell and gene therapy production.
cell therapy centrifuge
CO₂ incubators for cell and gene therapy
Thermo Scientific stackable CO2 incubators are suitable for ISO Class 5 and Good Manufacturing Practices (GMP) grade A/B environments. For cleanroom usage, we offer the Heracell Vios CR and Steri-cycle CR CO2 incubators – CTS series. All of our CO2 incubators have the option to be stacked and provide optimal cell growth conditions, enhanced cleaning compatibility and an active particle control system.
Next generation cleanroom confidence
Thermo Scientific Heracell Vios CR and Steri-Cycle CR CO2 incubators – CTS series are the first third-party certified cleanroom compatible CO2 incubators on the market, suitable for use in ISO Class 5 and GMP Grade A/B environments. They are designed to withstand rigorous cleaning protocols and come in multiple sizes and configurations to help support increased production in less space.
cell therapy vios incubator
High-volume culturing
Thermo Scientific Forma Steri-Cult CO₂ Incubator – CTS Series offers the ultimate protection for high-value cultures with every element of the design, from the ISO Class 5 HEPA filtration system to their easy cleaning processes and proven automated sterilization cycles. As the largest capacity stackable CO₂ incubator in the market today, the CTS Series supports cell therapy needs from research to high volume production, providing the documentation ritical to validation every step of the way.
forma steri-cult
Controlled-rate freezers for cell and gene therapy
To maintain cellular viability during the freezing process, certain cells require precise control of the freezing rate. This helps minimize the detrimental effects of undercooling and the heat liberation during the phase change process from water to ice. The Thermo Scientific CryoMed Controlled-Rate Freezers (CRFs) are designed with reliability, protection, and data traceability in mind for your sample preparation and GMP production needs.
Freeze samples with repeatable results
For customers in cell and gene therapy, vaccine production, and biobanking, the Thermo Scientific CryoMed Controlled-Rate Freezer (CRF) provides precise, repeatable freezing results that protect samples from intracellular freezing. The CryoMed CRF features enhanced data traceability via a touch screen user interface and offers customizable freezing profiles while supporting 21 CFR part 11 and GMP needs.
CTS equipment Cryomed Controlled incubator
For Laboratory Use.
1x1 image pixel for data collection