What are controls and why are they needed?
Controls play a vital role in helping to ensure the efficiency and accuracy of each stage within an infectious disease research workflow. By incorporating controls, we can closely monitor and evaluate the performance of the workflow as a whole. Controls enable assurance of reliability by validating the accuracy of the results obtained throughout the workflow, minimizing the risk of false-positive or false-negative outcomes.
  • Process controls:
    • Used in the sample preparation and preamplification step to assess whether extraction and purification steps are effectively isolating and preserving nucleic acids of interest
    •  Used to evaluate the performance of preamplification
  • Amplification controls:
    • Used in the PCR stage to help ensure reliability and accuracy of the assay
Overview of controls available for infectious disease research workflows
Research workflow steps that involve the use of controls
Below, we’ve provided a process overview to demonstrate when to utilize process and amplification controls in a real-time PCR workflow for infectious disease research. While the specific example shown is for an OpenArray real-time PCR workflow, the steps are generally the same for TrueMark plates and cards.
The full real-time PCR workflow goes from specimen processing to sample prep and preamplification, to sample loading, to real-time PCR, and finally, analysis and report. Process controls are used during sample prep and preamplification, and amplification controls are used during the real-time PCR step.
Inventoried controls
Process controls for sample preparation and preamplification
TaqMan Universal Extraction Control Organism
TaqMan Spike in (Xeno) Controls
  • Run as separate sample
  • Goes into sample extraction, or it can be added directly to samples, which is optional
  • (B. atrophaeus), serves as a process control for cell lysis and nucleic acid recovery
  • The control is used with the proprietary TaqMan assay for Bacillus atrophaeus sequences
  • Spike into samples during and after sample extraction
  • Serves as a process control for the purification process and presence of inhibitors
Type of control
Catalog #
Product description
Volume
Extraction control
A39180
TaqMan Universal Extraction Control Organism
1 kit (3 pellets)
Spike-in control
A39175
TaqMan Universal DNA Spike-In Control
1,000 µL
Spike-in control
A39176
TaqMan  Universal  Universal DNA Spike-In Control (5X)
5 x 1,000 µL
Spike-in control
A50384
TaqMan Xeno Control, kanamycin resistance
1,000 uL
Spike-in / Reverse transcription control
A39179
TaqMan Universal RNA Spike In/Reverse Transcription (Xeno) Control
5 x 200 µL
Amplification controls
Run amplification controls as a separate sample during the qPCR step to confirm assay performance at a known concentration. Amplification controls are available by application. If the application of interest is not listed in the table below, we also offer custom options to match the content of your panel.
Application
Catalog #
Product description
Volume
Vaginal microbiota
A32040
TaqMan Vaginal Microbiota Amplification Control
20 uL
Vaginal microbiota
A32913
TaqMan Vaginal Microbiota Amplification Control
250 uL
Urinary tract
A39174
TaqMan Urinary Tract Microbiota Amplification Control
5 x 50 uL
Respiratory tract
A39178
TaqMan Respiratory Tract Microbiota Amplification Control
5 x 50 uL
Respiratory tract
A48101
TrueMark Respiratory Panel 2.0 Amplification Control
250 uL
All
A50382
TrueMark Comprehensive Microbiota Control
1,000 uL
Custom real-time PCR amplification controls
Synthetic amplification controls are available to match the exact content of your custom TrueMark real-time PCR panel. Our TaqMan Custom DNA Controls contain a linearized, multi-target plasmid pool used for microbial research and detection using TaqMan OpenArray Plates or TaqMan Array Cards.
  • Controls can be included as a positive control for panel-specific amplification
  • High concentration products (5 x 107 copies/µL) can also be used to determine parameters necessary for analytical validation (e.g., limit of detection and Ct cut-off values)
  • During real-time PCR, the control can be used as a stand-alone sample to verify assay performance and help with troubleshooting
Catalog #
Product description
No. of targets
Concentration (copies/µL)
Volume (µL)
A50319
TrueMark Custom DNA Control
≤20
1 x 105
1,000
A50375
TrueMark Custom DNA Control
21 to 30
1 x 105
1,000
A50376
TrueMark Custom DNA Control
31 to 50
1 x 105
1,000
A50377
TrueMark Custom DNA Control
>50
1 x 105
1,000
A50378
TrueMark Custom DNA Control
≤20
5 x 107
50
A50379
TrueMark Custom DNA Control
21 to 30
5 x 107
50
A50380
TrueMark Custom DNA Control
31 to 50
5 x 107
50
A50381
TrueMark Custom DNA Control
>50
5 x 107
50
All TaqMan Custom DNA Control products should be stored at –15°C to –25°C. Please allow four weeks for your TaqMan Custom DNA Control product to be manufactured.
Ordering information
Ready to move forward with custom TrueMark real-time PCR solutions?
Complete this form to request a quote or consultation, or to place an order. A sales representative will reach out to walk you through the process of designing and implementing your new infectious disease research workflow.
For Research Use Only. Not for use in diagnostic procedures.